Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13388263-13388761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD10
Entrez Gene ID
84280
Gene Name
BTB domain containing 10
Gene Aliases
GMRP-1, GMRP1
UniGene
Hs.332382
Gene Chromosome Location
Chr.11: 13387996-13463297 on Build GRCh38
Transcripts
NM_001297741.1 NM_001297742.1 NM_032320.6 XM_017018405.1 XM_017018406.1 XM_017018407.1 XM_017018408.1 XM_017018409.1 XM_017018410.1 XM_017018411.1

SNP Detail

SNP ID Chromosome Location
rs952136704 Chr.11: 13388486-13388486 on Build GRCh38
rs966103594 Chr.11: 13388523-13388523 on Build GRCh38
rs190543646 Chr.11: 13388299-13388299 on Build GRCh38
rs767365124 Chr.11: 13388511-13388511 on Build GRCh38
rs1042457552 Chr.11: 13388384-13388385 on Build GRCh38
rs377374895 Chr.11: 13388630-13388630 on Build GRCh38
rs922566909 Chr.11: 13388554-13388555 on Build GRCh38
rs537274590 Chr.11: 13388588-13388588 on Build GRCh38
rs573638303 Chr.11: 13388670-13388670 on Build GRCh38
rs757009560 Chr.11: 13388397-13388397 on Build GRCh38
rs138959637 Chr.11: 13388661-13388661 on Build GRCh38
rs946453220 Chr.11: 13388660-13388660 on Build GRCh38
rs1056751768 Chr.11: 13388430-13388430 on Build GRCh38
rs182380118 Chr.11: 13388698-13388698 on Build GRCh38
rs1044431 Chr.11: 13388296-13388296 on Build GRCh38
rs1049221576 Chr.11: 13388717-13388717 on Build GRCh38
rs374929380 Chr.11: 13388624-13388624 on Build GRCh38
rs749997805 Chr.11: 13388731-13388731 on Build GRCh38
rs914961668 Chr.11: 13388606-13388606 on Build GRCh38
rs893581146 Chr.11: 13388352-13388352 on Build GRCh38
rs1015335513 Chr.11: 13388521-13388521 on Build GRCh38
rs939059604 Chr.11: 13388675-13388675 on Build GRCh38
rs550716795 Chr.11: 13388577-13388577 on Build GRCh38
rs923596398 Chr.11: 13388661-13388661 on Build GRCh38
rs977058611 Chr.11: 13388539-13388539 on Build GRCh38
rs149291328 Chr.11: 13388700-13388700 on Build GRCh38
rs757435740 Chr.11: 13388689-13388690 on Build GRCh38
rs779651849 Chr.11: 13388726-13388726 on Build GRCh38
rs576022722 Chr.11: 13388477-13388477 on Build GRCh38
rs1027665907 Chr.11: 13388477-13388478 on Build GRCh38
rs895306336 Chr.11: 13388434-13388436 on Build GRCh38
rs902725410 Chr.11: 13388406-13388406 on Build GRCh38
rs766997853 Chr.11: 13388660-13388661 on Build GRCh38
rs1012388175 Chr.11: 13388443-13388443 on Build GRCh38
rs138607787 Chr.11: 13388378-13388378 on Build GRCh38
rs755997543 Chr.11: 13388701-13388701 on Build GRCh38
rs985306709 Chr.11: 13388605-13388605 on Build GRCh38
rs557821260 Chr.11: 13388419-13388419 on Build GRCh38

View Full Product Details