Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13388993-13389425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD10
Entrez Gene ID
84280
Gene Name
BTB domain containing 10
Gene Aliases
GMRP-1, GMRP1
UniGene
Hs.332382
Gene Chromosome Location
Chr.11: 13387996-13463297 on Build GRCh38
Transcripts
NM_001297741.1 NM_001297742.1 NM_032320.6 XM_017018405.1 XM_017018406.1 XM_017018407.1 XM_017018408.1 XM_017018409.1 XM_017018410.1 XM_017018411.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
458686 c.1205G>A p.R402Q Chr.11: 13389054-13389054 on Build GRCh38
1627876 c.1118-6_1118-3delTTTC p.? Chr.11: 13389144-13389147 on Build GRCh38
6224028 c.1162C>T p.R388C Chr.11: 13389097-13389097 on Build GRCh38
6613296 c.1156G>A p.G386R Chr.11: 13389103-13389103 on Build GRCh38
7104778 c.1163G>T p.R388L Chr.11: 13389096-13389096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147243996 Chr.11: 13389118-13389118 on Build GRCh38
rs3833472 Chr.11: 13389144-13389147 on Build GRCh38
rs1021875357 Chr.11: 13389201-13389201 on Build GRCh38
rs755916930 Chr.11: 13389057-13389057 on Build GRCh38
rs764047272 Chr.11: 13389062-13389062 on Build GRCh38
rs547293901 Chr.11: 13389400-13389400 on Build GRCh38
rs766656420 Chr.11: 13389045-13389045 on Build GRCh38
rs759997243 Chr.11: 13389151-13389152 on Build GRCh38
rs753752344 Chr.11: 13389096-13389096 on Build GRCh38
rs977970258 Chr.11: 13389265-13389265 on Build GRCh38
rs375824034 Chr.11: 13389154-13389157 on Build GRCh38
rs563825040 Chr.11: 13389102-13389102 on Build GRCh38
rs980783151 Chr.11: 13389231-13389231 on Build GRCh38
rs778015480 Chr.11: 13389157-13389157 on Build GRCh38
rs992554551 Chr.11: 13389320-13389320 on Build GRCh38
rs992356286 Chr.11: 13389356-13389356 on Build GRCh38
rs748571680 Chr.11: 13389188-13389188 on Build GRCh38
rs746454873 Chr.11: 13389388-13389388 on Build GRCh38
rs956138191 Chr.11: 13389173-13389173 on Build GRCh38
rs373249163 Chr.11: 13389381-13389381 on Build GRCh38
rs75019394 Chr.11: 13389054-13389054 on Build GRCh38
rs769163052 Chr.11: 13389164-13389164 on Build GRCh38
rs1007917541 Chr.11: 13389155-13389155 on Build GRCh38
rs940262306 Chr.11: 13389336-13389336 on Build GRCh38
rs528844903 Chr.11: 13389325-13389325 on Build GRCh38
rs752423002 Chr.11: 13389055-13389055 on Build GRCh38
rs758725248 Chr.11: 13389154-13389154 on Build GRCh38
rs960664804 Chr.11: 13389251-13389251 on Build GRCh38
rs555820989 Chr.11: 13389328-13389328 on Build GRCh38
rs948304085 Chr.11: 13389070-13389070 on Build GRCh38
rs1038651509 Chr.11: 13389340-13389340 on Build GRCh38
rs144205292 Chr.11: 13389302-13389302 on Build GRCh38
rs750003509 Chr.11: 13389116-13389116 on Build GRCh38
rs753180265 Chr.11: 13389153-13389153 on Build GRCh38
rs112455409 Chr.11: 13389216-13389216 on Build GRCh38
rs781693674 Chr.11: 13389177-13389177 on Build GRCh38
rs765763753 Chr.11: 13389162-13389162 on Build GRCh38
rs561317873 Chr.11: 13389272-13389272 on Build GRCh38
rs1036829779 Chr.11: 13389198-13389198 on Build GRCh38
rs144141685 Chr.11: 13389032-13389032 on Build GRCh38
rs1006805521 Chr.11: 13389251-13389253 on Build GRCh38
rs56274863 Chr.11: 13389103-13389103 on Build GRCh38
rs543213291 Chr.11: 13389234-13389234 on Build GRCh38
rs916239731 Chr.11: 13389368-13389368 on Build GRCh38
rs529830585 Chr.11: 13389382-13389387 on Build GRCh38
rs575183642 Chr.11: 13389334-13389336 on Build GRCh38
rs753011483 Chr.11: 13389175-13389176 on Build GRCh38
rs990107334 Chr.11: 13389176-13389176 on Build GRCh38
rs773380514 Chr.11: 13389023-13389023 on Build GRCh38
rs984988871 Chr.11: 13389382-13389382 on Build GRCh38
rs1022251707 Chr.11: 13389251-13389254 on Build GRCh38
rs771772666 Chr.11: 13389382-13389384 on Build GRCh38
rs889162361 Chr.11: 13389244-13389244 on Build GRCh38
rs920059762 Chr.11: 13389383-13389383 on Build GRCh38

View Full Product Details