Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 13694610-13695108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAR1
Entrez Gene ID
84188
Gene Name
fatty acyl-CoA reductase 1
Gene Aliases
MLSTD2, PFCRD, SDR10E1
UniGene
Hs.501991
Gene Chromosome Location
Chr.11: 13668659-13732346 on Build GRCh38
Transcripts
NM_032228.5 XM_011520400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6268372 c.75G>T p.V25V Chr.11: 13694840-13694840 on Build GRCh38
1162940 c.75G>A p.V25V Chr.11: 13694840-13694840 on Build GRCh38
353736 c.141A>T p.K47N Chr.11: 13694906-13694906 on Build GRCh38
2090533 c.189G>T p.K63N Chr.11: 13694954-13694954 on Build GRCh38
7121808 c.33G>T p.K11N Chr.11: 13694798-13694798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535559662 Chr.11: 13694696-13694696 on Build GRCh38
rs779466538 Chr.11: 13694915-13694915 on Build GRCh38
rs202010733 Chr.11: 13694901-13694901 on Build GRCh38
rs777996465 Chr.11: 13694727-13694727 on Build GRCh38
rs772019928 Chr.11: 13694761-13694761 on Build GRCh38
rs376190176 Chr.11: 13694942-13694942 on Build GRCh38
rs549284066 Chr.11: 13695084-13695084 on Build GRCh38
rs760159969 Chr.11: 13694746-13694746 on Build GRCh38
rs771576871 Chr.11: 13694973-13694974 on Build GRCh38
rs774938787 Chr.11: 13694995-13694995 on Build GRCh38
rs750699930 Chr.11: 13694941-13694941 on Build GRCh38
rs755663785 Chr.11: 13694883-13694883 on Build GRCh38
rs775362138 Chr.11: 13694925-13694925 on Build GRCh38
rs559635265 Chr.11: 13694984-13694984 on Build GRCh38
rs372922085 Chr.11: 13694732-13694732 on Build GRCh38
rs200113117 Chr.11: 13694744-13694744 on Build GRCh38
rs563599248 Chr.11: 13695038-13695038 on Build GRCh38
rs766142190 Chr.11: 13694967-13694967 on Build GRCh38
rs116695081 Chr.11: 13694706-13694706 on Build GRCh38
rs7130943 Chr.11: 13694983-13694983 on Build GRCh38
rs12282039 Chr.11: 13694635-13694635 on Build GRCh38
rs369516382 Chr.11: 13694716-13694716 on Build GRCh38
rs769544089 Chr.11: 13694726-13694726 on Build GRCh38
rs768003518 Chr.11: 13695072-13695073 on Build GRCh38
rs779413918 Chr.11: 13695000-13695000 on Build GRCh38
rs140104758 Chr.11: 13694940-13694940 on Build GRCh38
rs749001926 Chr.11: 13695028-13695028 on Build GRCh38
rs573784971 Chr.11: 13694962-13694962 on Build GRCh38
rs565669518 Chr.11: 13694714-13694714 on Build GRCh38
rs143770401 Chr.11: 13695067-13695068 on Build GRCh38
rs755681545 Chr.11: 13694986-13694986 on Build GRCh38
rs750130905 Chr.11: 13694802-13694802 on Build GRCh38
rs757025174 Chr.11: 13694997-13694997 on Build GRCh38
rs780555760 Chr.11: 13694853-13694853 on Build GRCh38
rs781618056 Chr.11: 13694841-13694841 on Build GRCh38
rs771515994 Chr.11: 13694734-13694734 on Build GRCh38
rs71482916 Chr.11: 13695072-13695072 on Build GRCh38
rs142969007 Chr.11: 13694829-13694829 on Build GRCh38
rs61741588 Chr.11: 13694801-13694801 on Build GRCh38
rs766940262 Chr.11: 13694982-13694982 on Build GRCh38
rs371461855 Chr.11: 13695003-13695003 on Build GRCh38
rs762762822 Chr.11: 13694819-13694819 on Build GRCh38
rs545006269 Chr.11: 13695023-13695023 on Build GRCh38
rs752320111 Chr.11: 13694876-13694876 on Build GRCh38
rs749628706 Chr.11: 13694900-13694900 on Build GRCh38
rs772556363 Chr.11: 13694987-13694987 on Build GRCh38
rs117395380 Chr.11: 13694777-13694777 on Build GRCh38
rs200144475 Chr.11: 13695072-13695072 on Build GRCh38
rs776871908 Chr.11: 13694947-13694947 on Build GRCh38
rs760799339 Chr.11: 13694789-13694789 on Build GRCh38
rs777503835 Chr.11: 13694889-13694889 on Build GRCh38
rs112423747 Chr.11: 13695062-13695062 on Build GRCh38
rs947261604 Chr.11: 13694646-13694646 on Build GRCh38
rs751715597 Chr.11: 13694712-13694712 on Build GRCh38
rs538072123 Chr.11: 13694994-13694994 on Build GRCh38
rs139363221 Chr.11: 13694822-13694822 on Build GRCh38
rs772698486 Chr.11: 13694924-13694924 on Build GRCh38
rs369660345 Chr.11: 13694974-13694974 on Build GRCh38

View Full Product Details