Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14877890-14878390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2R1
Entrez Gene ID
120227
Gene Name
cytochrome P450 family 2 subfamily R member 1
Gene Aliases
-
UniGene
Hs.371427
Gene Chromosome Location
Chr.11: 14877436-14898913 on Build GRCh38
Transcripts
NM_024514.4 XM_005252788.1 XM_005252789.2 XM_011519895.1 XM_011519898.2 XM_017017190.1 XM_017017191.1 XM_017017192.1 XM_017017193.1 XM_017017194.1 XM_017017195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
221265 c.1501C>G p.R501G Chr.11: 14878127-14878127 on Build GRCh38
4980451 c.1372A>G p.M458V Chr.11: 14878256-14878256 on Build GRCh38
5372136 c.1468C>T p.Q490* Chr.11: 14878160-14878160 on Build GRCh38
1352844 c.1390G>A p.A464T Chr.11: 14878238-14878238 on Build GRCh38
5894931 c.1331-1G>A p.? Chr.11: 14878298-14878298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374603644 Chr.11: 14878102-14878102 on Build GRCh38
rs782144463 Chr.11: 14878090-14878090 on Build GRCh38
rs782314673 Chr.11: 14878084-14878084 on Build GRCh38
rs977081819 Chr.11: 14878010-14878010 on Build GRCh38
rs542334082 Chr.11: 14877999-14877999 on Build GRCh38
rs782044143 Chr.11: 14878242-14878242 on Build GRCh38
rs146366698 Chr.11: 14878265-14878265 on Build GRCh38
rs782411780 Chr.11: 14878339-14878339 on Build GRCh38
rs781941135 Chr.11: 14878088-14878088 on Build GRCh38
rs772865661 Chr.11: 14878028-14878028 on Build GRCh38
rs782237999 Chr.11: 14878082-14878082 on Build GRCh38
rs781801893 Chr.11: 14878310-14878311 on Build GRCh38
rs782256827 Chr.11: 14878193-14878193 on Build GRCh38
rs782670089 Chr.11: 14878288-14878288 on Build GRCh38
rs781788203 Chr.11: 14878105-14878105 on Build GRCh38
rs2291781 Chr.11: 14877962-14877962 on Build GRCh38
rs782665218 Chr.11: 14878139-14878139 on Build GRCh38
rs543339931 Chr.11: 14878056-14878056 on Build GRCh38
rs200649142 Chr.11: 14878149-14878149 on Build GRCh38
rs782447018 Chr.11: 14878110-14878110 on Build GRCh38
rs781828535 Chr.11: 14878143-14878143 on Build GRCh38
rs1046098132 Chr.11: 14878203-14878203 on Build GRCh38
rs782459427 Chr.11: 14878287-14878287 on Build GRCh38
rs369802952 Chr.11: 14878316-14878316 on Build GRCh38
rs782091409 Chr.11: 14878253-14878254 on Build GRCh38
rs375059732 Chr.11: 14877965-14877965 on Build GRCh38
rs782443739 Chr.11: 14877957-14877957 on Build GRCh38
rs782819635 Chr.11: 14878097-14878097 on Build GRCh38
rs782488944 Chr.11: 14878128-14878128 on Build GRCh38
rs369501032 Chr.11: 14878230-14878230 on Build GRCh38
rs782013800 Chr.11: 14878254-14878254 on Build GRCh38
rs1030564176 Chr.11: 14877992-14877992 on Build GRCh38
rs782321028 Chr.11: 14878250-14878250 on Build GRCh38
rs782159981 Chr.11: 14878255-14878255 on Build GRCh38
rs781794360 Chr.11: 14878282-14878282 on Build GRCh38
rs782220639 Chr.11: 14878315-14878315 on Build GRCh38
rs782494351 Chr.11: 14878146-14878146 on Build GRCh38
rs201959570 Chr.11: 14878308-14878308 on Build GRCh38
rs191125686 Chr.11: 14878008-14878008 on Build GRCh38
rs530266065 Chr.11: 14877987-14877987 on Build GRCh38
rs375045934 Chr.11: 14878103-14878103 on Build GRCh38
rs782792215 Chr.11: 14878126-14878126 on Build GRCh38
rs373226890 Chr.11: 14878289-14878289 on Build GRCh38
rs781897345 Chr.11: 14878127-14878127 on Build GRCh38
rs782741911 Chr.11: 14878277-14878277 on Build GRCh38
rs957096811 Chr.11: 14878029-14878029 on Build GRCh38
rs782272712 Chr.11: 14878337-14878337 on Build GRCh38
rs782127135 Chr.11: 14878349-14878349 on Build GRCh38
rs915776714 Chr.11: 14878047-14878047 on Build GRCh38
rs375350522 Chr.11: 14878298-14878298 on Build GRCh38
rs201344073 Chr.11: 14878152-14878152 on Build GRCh38
rs545401539 Chr.11: 14878261-14878261 on Build GRCh38
rs781978068 Chr.11: 14878347-14878347 on Build GRCh38
rs148414061 Chr.11: 14877955-14877955 on Build GRCh38
rs781961850 Chr.11: 14878080-14878080 on Build GRCh38
rs782336661 Chr.11: 14878216-14878216 on Build GRCh38
rs887879653 Chr.11: 14878314-14878314 on Build GRCh38
rs782508410 Chr.11: 14878295-14878295 on Build GRCh38
rs782770082 Chr.11: 14878119-14878119 on Build GRCh38
rs782675680 Chr.11: 14878176-14878176 on Build GRCh38
rs781875625 Chr.11: 14878264-14878264 on Build GRCh38
rs540425350 Chr.11: 14877924-14877924 on Build GRCh38
rs989956990 Chr.11: 14878030-14878030 on Build GRCh38

View Full Product Details