Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14885605-14886141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2R1
Entrez Gene ID
120227
Gene Name
cytochrome P450 family 2 subfamily R member 1
Gene Aliases
-
UniGene
Hs.371427
Gene Chromosome Location
Chr.11: 14877436-14898913 on Build GRCh38
Transcripts
NM_024514.4 XM_005252788.1 XM_005252789.2 XM_011519895.1 XM_011519898.2 XM_017017190.1 XM_017017191.1 XM_017017192.1 XM_017017193.1 XM_017017194.1 XM_017017195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6461666 c.342A>T p.L114F Chr.11: 14885801-14885801 on Build GRCh38
6031833 c.324C>T p.D108D Chr.11: 14885819-14885819 on Build GRCh38
274722 c.310G>A p.E104K Chr.11: 14885833-14885833 on Build GRCh38
3700027 c.279T>C p.D93D Chr.11: 14885864-14885864 on Build GRCh38
6344182 c.329C>A p.P110Q Chr.11: 14885814-14885814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782799552 Chr.11: 14885773-14885773 on Build GRCh38
rs201553303 Chr.11: 14885844-14885844 on Build GRCh38
rs781829367 Chr.11: 14886008-14886008 on Build GRCh38
rs781844096 Chr.11: 14886008-14886008 on Build GRCh38
rs909596957 Chr.11: 14886085-14886085 on Build GRCh38
rs375085420 Chr.11: 14885791-14885791 on Build GRCh38
rs778653093 Chr.11: 14886068-14886068 on Build GRCh38
rs782596005 Chr.11: 14885954-14885954 on Build GRCh38
rs782730969 Chr.11: 14885779-14885779 on Build GRCh38
rs111824082 Chr.11: 14885834-14885834 on Build GRCh38
rs371686308 Chr.11: 14885747-14885747 on Build GRCh38
rs567512092 Chr.11: 14886022-14886022 on Build GRCh38
rs138945380 Chr.11: 14885853-14885853 on Build GRCh38
rs192940523 Chr.11: 14885918-14885918 on Build GRCh38
rs782784041 Chr.11: 14886005-14886005 on Build GRCh38
rs782776278 Chr.11: 14885908-14885908 on Build GRCh38
rs782065675 Chr.11: 14885995-14885995 on Build GRCh38
rs782523665 Chr.11: 14885776-14885776 on Build GRCh38
rs61744566 Chr.11: 14885820-14885820 on Build GRCh38
rs782025990 Chr.11: 14885889-14885889 on Build GRCh38
rs781969896 Chr.11: 14885818-14885818 on Build GRCh38
rs138438430 Chr.11: 14885813-14885813 on Build GRCh38
rs376832292 Chr.11: 14885957-14885957 on Build GRCh38
rs202011621 Chr.11: 14885775-14885775 on Build GRCh38
rs782344364 Chr.11: 14885859-14885859 on Build GRCh38
rs782211370 Chr.11: 14885992-14885992 on Build GRCh38
rs182378927 Chr.11: 14886035-14886035 on Build GRCh38
rs78383885 Chr.11: 14885976-14885976 on Build GRCh38
rs753817846 Chr.11: 14885993-14885993 on Build GRCh38
rs970384363 Chr.11: 14885706-14885706 on Build GRCh38
rs998161173 Chr.11: 14885858-14885858 on Build GRCh38
rs782286410 Chr.11: 14885731-14885731 on Build GRCh38
rs372298377 Chr.11: 14885932-14885932 on Build GRCh38
rs782031601 Chr.11: 14886000-14886000 on Build GRCh38
rs782746132 Chr.11: 14885754-14885754 on Build GRCh38
rs782428542 Chr.11: 14885971-14885971 on Build GRCh38
rs112205170 Chr.11: 14886021-14886021 on Build GRCh38
rs61744571 Chr.11: 14885833-14885833 on Build GRCh38
rs782115359 Chr.11: 14885871-14885871 on Build GRCh38
rs782087797 Chr.11: 14885911-14885911 on Build GRCh38
rs756231680 Chr.11: 14885632-14885632 on Build GRCh38
rs782522679 Chr.11: 14885941-14885941 on Build GRCh38
rs1000712687 Chr.11: 14885890-14885890 on Build GRCh38
rs782549131 Chr.11: 14885841-14885841 on Build GRCh38
rs575781482 Chr.11: 14885677-14885677 on Build GRCh38
rs782809468 Chr.11: 14885939-14885939 on Build GRCh38
rs942532663 Chr.11: 14886089-14886089 on Build GRCh38
rs375785354 Chr.11: 14886001-14886001 on Build GRCh38
rs781855914 Chr.11: 14885940-14885940 on Build GRCh38
rs992190998 Chr.11: 14885646-14885646 on Build GRCh38
rs782372996 Chr.11: 14885735-14885735 on Build GRCh38
rs903179615 Chr.11: 14885884-14885884 on Build GRCh38
rs565274924 Chr.11: 14885878-14885878 on Build GRCh38
rs542981986 Chr.11: 14885763-14885763 on Build GRCh38
rs782287559 Chr.11: 14885960-14885960 on Build GRCh38
rs782381294 Chr.11: 14885893-14885893 on Build GRCh38
rs186671646 Chr.11: 14886113-14886113 on Build GRCh38
rs782400829 Chr.11: 14885997-14885997 on Build GRCh38
rs782068748 Chr.11: 14885762-14885762 on Build GRCh38
rs61495246 Chr.11: 14885847-14885847 on Build GRCh38
rs942538897 Chr.11: 14885837-14885837 on Build GRCh38
rs781928193 Chr.11: 14885761-14885761 on Build GRCh38
rs782610438 Chr.11: 14885868-14885868 on Build GRCh38
rs917472675 Chr.11: 14885653-14885653 on Build GRCh38
rs782027522 Chr.11: 14885768-14885768 on Build GRCh38
rs766412473 Chr.11: 14885654-14885657 on Build GRCh38
rs528079027 Chr.11: 14886024-14886024 on Build GRCh38
rs782698706 Chr.11: 14885927-14885927 on Build GRCh38
rs782149683 Chr.11: 14885933-14885933 on Build GRCh38
rs782004608 Chr.11: 14885739-14885739 on Build GRCh38
rs782433035 Chr.11: 14885958-14885958 on Build GRCh38

View Full Product Details