Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 15078407-15078944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCB
Entrez Gene ID
797
Gene Name
calcitonin related polypeptide beta
Gene Aliases
CALC2, CGRP-II, CGRP2
UniGene
Hs.534305
Gene Chromosome Location
Chr.11: 15073600-15078631 on Build GRCh38
Transcripts
NM_000728.3

SNP Detail

SNP ID Chromosome Location
rs565007597 Chr.11: 15078844-15078844 on Build GRCh38
rs1003030638 Chr.11: 15078685-15078685 on Build GRCh38
rs139497288 Chr.11: 15078590-15078590 on Build GRCh38
rs767650234 Chr.11: 15078493-15078493 on Build GRCh38
rs990487706 Chr.11: 15078613-15078613 on Build GRCh38
rs149306227 Chr.11: 15078592-15078592 on Build GRCh38
rs960440566 Chr.11: 15078698-15078698 on Build GRCh38
rs528069203 Chr.11: 15078736-15078736 on Build GRCh38
rs559029538 Chr.11: 15078516-15078516 on Build GRCh38
rs980906726 Chr.11: 15078444-15078444 on Build GRCh38
rs1042885 Chr.11: 15078505-15078505 on Build GRCh38
rs986649703 Chr.11: 15078848-15078848 on Build GRCh38
rs891195361 Chr.11: 15078594-15078594 on Build GRCh38
rs1036291969 Chr.11: 15078687-15078687 on Build GRCh38
rs1041564595 Chr.11: 15078752-15078752 on Build GRCh38
rs960473219 Chr.11: 15078488-15078488 on Build GRCh38
rs952207353 Chr.11: 15078562-15078562 on Build GRCh38
rs571576025 Chr.11: 15078605-15078605 on Build GRCh38
rs921055076 Chr.11: 15078574-15078574 on Build GRCh38
rs1023235699 Chr.11: 15078640-15078640 on Build GRCh38
rs761991751 Chr.11: 15078449-15078449 on Build GRCh38
rs373005764 Chr.11: 15078610-15078610 on Build GRCh38
rs1011745587 Chr.11: 15078637-15078637 on Build GRCh38
rs921931737 Chr.11: 15078608-15078610 on Build GRCh38
rs1053042186 Chr.11: 15078607-15078607 on Build GRCh38
rs962372115 Chr.11: 15078759-15078759 on Build GRCh38
rs546580146 Chr.11: 15078604-15078604 on Build GRCh38
rs538759949 Chr.11: 15078786-15078786 on Build GRCh38
rs560902687 Chr.11: 15078717-15078717 on Build GRCh38
rs774677427 Chr.11: 15078436-15078436 on Build GRCh38
rs754724166 Chr.11: 15078679-15078679 on Build GRCh38
rs938434926 Chr.11: 15078895-15078895 on Build GRCh38
rs932586712 Chr.11: 15078803-15078803 on Build GRCh38
rs750459722 Chr.11: 15078539-15078539 on Build GRCh38
rs755289242 Chr.11: 15078753-15078753 on Build GRCh38
rs1540140 Chr.11: 15078660-15078660 on Build GRCh38
rs921202490 Chr.11: 15078789-15078789 on Build GRCh38
rs928382666 Chr.11: 15078892-15078892 on Build GRCh38
rs1015002443 Chr.11: 15078579-15078579 on Build GRCh38
rs532069903 Chr.11: 15078846-15078846 on Build GRCh38
rs12274834 Chr.11: 15078507-15078507 on Build GRCh38
rs75237899 Chr.11: 15078610-15078610 on Build GRCh38
rs974371299 Chr.11: 15078774-15078774 on Build GRCh38
rs554743388 Chr.11: 15078593-15078595 on Build GRCh38
rs970254687 Chr.11: 15078429-15078429 on Build GRCh38
rs982424731 Chr.11: 15078511-15078511 on Build GRCh38
rs1038296212 Chr.11: 15078564-15078564 on Build GRCh38

View Full Product Details