Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 18704538-18705037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF22
Entrez Gene ID
283284
Gene Name
immunoglobulin superfamily member 22
Gene Aliases
IGFN2
UniGene
Hs.434152
Gene Chromosome Location
Chr.11: 18704305-18726939 on Build GRCh38
Transcripts
NM_173588.3 XM_011520030.2 XM_011520031.2 XM_017017607.1 XM_017017608.1 NM_153347.2

Gene Symbol
TMEM86A
Entrez Gene ID
144110
Gene Name
transmembrane protein 86A
Gene Aliases
-
UniGene
Hs.502100
Gene Chromosome Location
Chr.11: 18698735-18704785 on Build GRCh38
Transcripts
NM_173588.3 XM_011520030.2 XM_011520031.2 XM_017017607.1 XM_017017608.1 NM_153347.2

SNP Detail

SNP ID Chromosome Location
rs542364977 Chr.11: 18704872-18704872 on Build GRCh38
rs996062233 Chr.11: 18704704-18704704 on Build GRCh38
rs1016630383 Chr.11: 18704983-18704983 on Build GRCh38
rs1042103094 Chr.11: 18704592-18704592 on Build GRCh38
rs1048987732 Chr.11: 18704997-18704997 on Build GRCh38
rs543078198 Chr.11: 18704607-18704607 on Build GRCh38
rs751056944 Chr.11: 18704685-18704685 on Build GRCh38
rs949630848 Chr.11: 18704947-18704947 on Build GRCh38
rs898222049 Chr.11: 18704986-18704986 on Build GRCh38
rs146162324 Chr.11: 18704979-18704979 on Build GRCh38
rs902206190 Chr.11: 18704605-18704605 on Build GRCh38
rs951084174 Chr.11: 18704717-18704717 on Build GRCh38
rs977964363 Chr.11: 18704880-18704880 on Build GRCh38
rs368378451 Chr.11: 18704633-18704633 on Build GRCh38
rs114137280 Chr.11: 18704757-18704757 on Build GRCh38
rs552946698 Chr.11: 18704562-18704562 on Build GRCh38
rs139558433 Chr.11: 18704613-18704613 on Build GRCh38
rs547524255 Chr.11: 18704752-18704752 on Build GRCh38
rs558072843 Chr.11: 18704610-18704610 on Build GRCh38
rs925370864 Chr.11: 18704893-18704893 on Build GRCh38
rs533411437 Chr.11: 18704829-18704829 on Build GRCh38
rs146332021 Chr.11: 18704572-18704572 on Build GRCh38
rs938136503 Chr.11: 18704896-18704896 on Build GRCh38
rs760510327 Chr.11: 18704584-18704584 on Build GRCh38
rs1027521081 Chr.11: 18704724-18704724 on Build GRCh38
rs1055682844 Chr.11: 18704903-18704903 on Build GRCh38
rs559701039 Chr.11: 18704820-18704820 on Build GRCh38
rs530701300 Chr.11: 18704973-18704973 on Build GRCh38
rs887605692 Chr.11: 18704725-18704725 on Build GRCh38
rs377545207 Chr.11: 18704581-18704581 on Build GRCh38
rs372174668 Chr.11: 18704710-18704710 on Build GRCh38
rs186013835 Chr.11: 18704785-18704785 on Build GRCh38
rs757741389 Chr.11: 18704933-18704933 on Build GRCh38
rs1043793277 Chr.11: 18704873-18704873 on Build GRCh38
rs971858847 Chr.11: 18704759-18704759 on Build GRCh38
rs975639288 Chr.11: 18704764-18704764 on Build GRCh38
rs931923696 Chr.11: 18704993-18704993 on Build GRCh38
rs759508959 Chr.11: 18704694-18704694 on Build GRCh38
rs896697005 Chr.11: 18704932-18704932 on Build GRCh38
rs34874178 Chr.11: 18704798-18704798 on Build GRCh38
rs370998366 Chr.11: 18704818-18704818 on Build GRCh38
rs923734402 Chr.11: 18704974-18704974 on Build GRCh38
rs7106356 Chr.11: 18704977-18704977 on Build GRCh38
rs1028154402 Chr.11: 18705012-18705012 on Build GRCh38
rs373109786 Chr.11: 18704854-18704854 on Build GRCh38
rs78847254 Chr.11: 18704856-18704856 on Build GRCh38
rs1004992602 Chr.11: 18704756-18704756 on Build GRCh38
rs543639922 Chr.11: 18704655-18704655 on Build GRCh38
rs900158209 Chr.11: 18704992-18704992 on Build GRCh38

View Full Product Details