Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 28023361-28023863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18A
Entrez Gene ID
81930
Gene Name
kinesin family member 18A
Gene Aliases
MS-KIF18A, PPP1R99
UniGene
Hs.301052
Gene Chromosome Location
Chr.11: 28020616-28108199 on Build GRCh38
Transcripts
NM_031217.3 XM_017018379.1 XM_017018380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
71337 c.2517C>G p.N839K Chr.11: 28023838-28023838 on Build GRCh38
4453130 c.2572A>C p.N858H Chr.11: 28023783-28023783 on Build GRCh38
926155 c.2522C>T p.S841L Chr.11: 28023833-28023833 on Build GRCh38
5838390 c.2533G>C p.D845H Chr.11: 28023822-28023822 on Build GRCh38
926154 c.2543C>A p.S848Y Chr.11: 28023812-28023812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767693064 Chr.11: 28023721-28023721 on Build GRCh38
rs770203259 Chr.11: 28023703-28023703 on Build GRCh38
rs55963710 Chr.11: 28023395-28023395 on Build GRCh38
rs748835733 Chr.11: 28023676-28023676 on Build GRCh38
rs1026888647 Chr.11: 28023512-28023512 on Build GRCh38
rs111742563 Chr.11: 28023605-28023605 on Build GRCh38
rs545863232 Chr.11: 28023524-28023524 on Build GRCh38
rs751776880 Chr.11: 28023716-28023716 on Build GRCh38
rs949096002 Chr.11: 28023580-28023580 on Build GRCh38
rs773462468 Chr.11: 28023609-28023609 on Build GRCh38
rs982247812 Chr.11: 28023593-28023593 on Build GRCh38
rs890421345 Chr.11: 28023689-28023689 on Build GRCh38
rs965263762 Chr.11: 28023486-28023486 on Build GRCh38
rs916200775 Chr.11: 28023568-28023568 on Build GRCh38
rs1053916268 Chr.11: 28023623-28023623 on Build GRCh38
rs940119469 Chr.11: 28023608-28023608 on Build GRCh38
rs773412995 Chr.11: 28023708-28023708 on Build GRCh38
rs952639670 Chr.11: 28023520-28023520 on Build GRCh38
rs917254938 Chr.11: 28023625-28023625 on Build GRCh38
rs567761460 Chr.11: 28023592-28023592 on Build GRCh38
rs369809315 Chr.11: 28023833-28023833 on Build GRCh38
rs935490744 Chr.11: 28023620-28023620 on Build GRCh38
rs748687672 Chr.11: 28023819-28023819 on Build GRCh38
rs538307388 Chr.11: 28023610-28023610 on Build GRCh38
rs555888608 Chr.11: 28023565-28023565 on Build GRCh38
rs368276152 Chr.11: 28023711-28023711 on Build GRCh38
rs773750645 Chr.11: 28023826-28023826 on Build GRCh38
rs972583028 Chr.11: 28023476-28023476 on Build GRCh38
rs370591664 Chr.11: 28023657-28023657 on Build GRCh38
rs567165490 Chr.11: 28023417-28023417 on Build GRCh38
rs909954057 Chr.11: 28023606-28023606 on Build GRCh38
rs977078059 Chr.11: 28023551-28023551 on Build GRCh38
rs534262396 Chr.11: 28023563-28023563 on Build GRCh38
rs1009176126 Chr.11: 28023442-28023442 on Build GRCh38
rs143263342 Chr.11: 28023640-28023640 on Build GRCh38
rs906010064 Chr.11: 28023686-28023686 on Build GRCh38
rs921071891 Chr.11: 28023552-28023552 on Build GRCh38
rs1020612265 Chr.11: 28023477-28023477 on Build GRCh38
rs950244377 Chr.11: 28023652-28023652 on Build GRCh38
rs777372169 Chr.11: 28023490-28023490 on Build GRCh38
rs746647574 Chr.11: 28023807-28023807 on Build GRCh38
rs906032205 Chr.11: 28023409-28023409 on Build GRCh38
rs894060204 Chr.11: 28023628-28023628 on Build GRCh38
rs778788771 Chr.11: 28023762-28023762 on Build GRCh38
rs573052783 Chr.11: 28023802-28023802 on Build GRCh38
rs143135596 Chr.11: 28023795-28023795 on Build GRCh38
rs577743377 Chr.11: 28023646-28023646 on Build GRCh38
rs1008915884 Chr.11: 28023690-28023690 on Build GRCh38
rs952585171 Chr.11: 28023797-28023797 on Build GRCh38
rs540161804 Chr.11: 28023810-28023810 on Build GRCh38
rs993992882 Chr.11: 28023717-28023717 on Build GRCh38
rs762246509 Chr.11: 28023700-28023700 on Build GRCh38
rs1054661171 Chr.11: 28023776-28023776 on Build GRCh38
rs33937991 Chr.11: 28023713-28023713 on Build GRCh38
rs757220711 Chr.11: 28023752-28023752 on Build GRCh38
rs199512613 Chr.11: 28023816-28023816 on Build GRCh38
rs759756095 Chr.11: 28023719-28023719 on Build GRCh38
rs112609277 Chr.11: 28023748-28023748 on Build GRCh38
rs957809234 Chr.11: 28023437-28023437 on Build GRCh38
rs1032527429 Chr.11: 28023427-28023427 on Build GRCh38
rs1044529238 Chr.11: 28023400-28023400 on Build GRCh38
rs750232861 Chr.11: 28023785-28023785 on Build GRCh38
rs777176373 Chr.11: 28023691-28023691 on Build GRCh38
rs753830666 Chr.11: 28023747-28023747 on Build GRCh38
rs35440657 Chr.11: 28023820-28023820 on Build GRCh38

View Full Product Details