Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 28036502-28037006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18A
Entrez Gene ID
81930
Gene Name
kinesin family member 18A
Gene Aliases
MS-KIF18A, PPP1R99
UniGene
Hs.301052
Gene Chromosome Location
Chr.11: 28020616-28108199 on Build GRCh38
Transcripts
NM_031217.3 XM_017018379.1 XM_017018380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3732188 c.2050T>G p.S684A Chr.11: 28036563-28036563 on Build GRCh38
4622049 c.2041A>T p.T681S Chr.11: 28036572-28036572 on Build GRCh38
4614664 c.1996_1997insA p.R666fs*29 Chr.11: 28036616-28036617 on Build GRCh38
1193750 c.2057C>T p.P686L Chr.11: 28036556-28036556 on Build GRCh38
6694869 c.2002C>T p.R668W Chr.11: 28036611-28036611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773695088 Chr.11: 28036590-28036590 on Build GRCh38
rs765377739 Chr.11: 28036643-28036643 on Build GRCh38
rs1033753130 Chr.11: 28036785-28036785 on Build GRCh38
rs945308556 Chr.11: 28036861-28036861 on Build GRCh38
rs932395238 Chr.11: 28036868-28036868 on Build GRCh38
rs775988893 Chr.11: 28036697-28036697 on Build GRCh38
rs1004879551 Chr.11: 28036752-28036752 on Build GRCh38
rs1053674658 Chr.11: 28036611-28036611 on Build GRCh38
rs759894610 Chr.11: 28036536-28036536 on Build GRCh38
rs775653842 Chr.11: 28036814-28036814 on Build GRCh38
rs753725745 Chr.11: 28036671-28036671 on Build GRCh38
rs529814655 Chr.11: 28036817-28036817 on Build GRCh38
rs959263608 Chr.11: 28036847-28036847 on Build GRCh38
rs1017103584 Chr.11: 28036736-28036736 on Build GRCh38
rs192667273 Chr.11: 28036825-28036825 on Build GRCh38
rs769910036 Chr.11: 28036813-28036813 on Build GRCh38
rs1042283060 Chr.11: 28036626-28036626 on Build GRCh38
rs778606809 Chr.11: 28036692-28036692 on Build GRCh38
rs957050628 Chr.11: 28036851-28036851 on Build GRCh38
rs770479385 Chr.11: 28036583-28036583 on Build GRCh38
rs986952960 Chr.11: 28036897-28036897 on Build GRCh38
rs781250611 Chr.11: 28036564-28036564 on Build GRCh38
rs770442076 Chr.11: 28036688-28036688 on Build GRCh38
rs754063353 Chr.11: 28036629-28036629 on Build GRCh38
rs752671024 Chr.11: 28036667-28036667 on Build GRCh38
rs372157089 Chr.11: 28036610-28036610 on Build GRCh38
rs922448944 Chr.11: 28036870-28036870 on Build GRCh38
rs931349087 Chr.11: 28036922-28036922 on Build GRCh38
rs781348142 Chr.11: 28036661-28036661 on Build GRCh38
rs1005560538 Chr.11: 28036720-28036720 on Build GRCh38
rs368827514 Chr.11: 28036695-28036695 on Build GRCh38
rs915158220 Chr.11: 28036852-28036852 on Build GRCh38
rs570376134 Chr.11: 28036912-28036912 on Build GRCh38
rs915169658 Chr.11: 28036613-28036613 on Build GRCh38
rs374870078 Chr.11: 28036534-28036534 on Build GRCh38
rs772507170 Chr.11: 28036596-28036596 on Build GRCh38
rs898517620 Chr.11: 28036762-28036762 on Build GRCh38
rs921060402 Chr.11: 28036867-28036867 on Build GRCh38
rs552106673 Chr.11: 28036907-28036907 on Build GRCh38
rs747999642 Chr.11: 28036569-28036569 on Build GRCh38
rs1037266703 Chr.11: 28036915-28036915 on Build GRCh38
rs755989917 Chr.11: 28036668-28036668 on Build GRCh38
rs759814640 Chr.11: 28036694-28036696 on Build GRCh38
rs745412515 Chr.11: 28036693-28036693 on Build GRCh38
rs1024457523 Chr.11: 28036774-28036774 on Build GRCh38
rs367934453 Chr.11: 28036551-28036551 on Build GRCh38
rs1055135659 Chr.11: 28036931-28036931 on Build GRCh38
rs769711180 Chr.11: 28036577-28036577 on Build GRCh38
rs926405862 Chr.11: 28036824-28036824 on Build GRCh38
rs761084790 Chr.11: 28036602-28036602 on Build GRCh38
rs748895322 Chr.11: 28036581-28036581 on Build GRCh38
rs889881472 Chr.11: 28036900-28036900 on Build GRCh38
rs760112585 Chr.11: 28036592-28036592 on Build GRCh38
rs532406376 Chr.11: 28036703-28036703 on Build GRCh38
rs756450221 Chr.11: 28036540-28036540 on Build GRCh38
rs569405148 Chr.11: 28036820-28036820 on Build GRCh38
rs758471172 Chr.11: 28036652-28036652 on Build GRCh38
rs933855331 Chr.11: 28036871-28036871 on Build GRCh38
rs958778335 Chr.11: 28036802-28036802 on Build GRCh38
rs187894327 Chr.11: 28036765-28036765 on Build GRCh38
rs548030337 Chr.11: 28036818-28036818 on Build GRCh38
rs749001814 Chr.11: 28036678-28036678 on Build GRCh38
rs762621215 Chr.11: 28036527-28036527 on Build GRCh38
rs898486527 Chr.11: 28036917-28036917 on Build GRCh38
rs889758596 Chr.11: 28036713-28036713 on Build GRCh38
rs765797738 Chr.11: 28036532-28036532 on Build GRCh38
rs750514567 Chr.11: 28036646-28036646 on Build GRCh38
rs144753657 Chr.11: 28036537-28036537 on Build GRCh38
rs764366000 Chr.11: 28036541-28036541 on Build GRCh38
rs777649400 Chr.11: 28036670-28036670 on Build GRCh38
rs754820459 Chr.11: 28036560-28036560 on Build GRCh38
rs945150959 Chr.11: 28036614-28036616 on Build GRCh38
rs775970379 Chr.11: 28036597-28036597 on Build GRCh38
rs989887120 Chr.11: 28036860-28036860 on Build GRCh38
rs777630970 Chr.11: 28036580-28036580 on Build GRCh38
rs953149220 Chr.11: 28036694-28036694 on Build GRCh38
rs757370775 Chr.11: 28036546-28036546 on Build GRCh38
rs113524981 Chr.11: 28036637-28036637 on Build GRCh38
rs145665161 Chr.11: 28036753-28036753 on Build GRCh38
rs761897304 Chr.11: 28036634-28036634 on Build GRCh38
rs761103357 Chr.11: 28036699-28036699 on Build GRCh38
rs1012792950 Chr.11: 28036804-28036804 on Build GRCh38
rs994402601 Chr.11: 28036943-28036943 on Build GRCh38
rs746995037 Chr.11: 28036556-28036556 on Build GRCh38
rs117698845 Chr.11: 28036968-28036968 on Build GRCh38

View Full Product Details