Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65340884-65341386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPF2
Entrez Gene ID
5977
Gene Name
double PHD fingers 2
Gene Aliases
REQ, UBID4, ubi-d4
UniGene
Hs.13495
Gene Chromosome Location
Chr.11: 65333754-65352980 on Build GRCh38
Transcripts
NM_006268.4 XM_005274149.2 XM_017018101.1 XM_017018102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4574654 c.219C>T p.S73S Chr.11: 65340991-65340991 on Build GRCh38
70523 c.227C>A p.A76D Chr.11: 65340999-65340999 on Build GRCh38
1241098 c.240_241insA p.R83fs*7 Chr.11: 65341012-65341013 on Build GRCh38
5823295 c.266A>T p.E89V Chr.11: 65341038-65341038 on Build GRCh38
4127485 c.194-1G>A p.? Chr.11: 65340965-65340965 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192636470 Chr.11: 65341331-65341331 on Build GRCh38
rs758102145 Chr.11: 65341195-65341195 on Build GRCh38
rs756830866 Chr.11: 65341083-65341083 on Build GRCh38
rs371177361 Chr.11: 65341102-65341102 on Build GRCh38
rs138433278 Chr.11: 65341255-65341255 on Build GRCh38
rs761898514 Chr.11: 65341054-65341054 on Build GRCh38
rs769153526 Chr.11: 65341106-65341106 on Build GRCh38
rs910017996 Chr.11: 65341170-65341170 on Build GRCh38
rs763561629 Chr.11: 65341032-65341032 on Build GRCh38
rs555684892 Chr.11: 65341200-65341200 on Build GRCh38
rs750677830 Chr.11: 65341063-65341063 on Build GRCh38
rs369045124 Chr.11: 65341209-65341209 on Build GRCh38
rs753577088 Chr.11: 65340966-65340966 on Build GRCh38
rs573587858 Chr.11: 65341167-65341167 on Build GRCh38
rs758428310 Chr.11: 65341077-65341077 on Build GRCh38
rs918559661 Chr.11: 65340910-65340910 on Build GRCh38
rs986429658 Chr.11: 65341347-65341347 on Build GRCh38
rs975839793 Chr.11: 65341284-65341284 on Build GRCh38
rs4647580 Chr.11: 65341273-65341273 on Build GRCh38
rs772164259 Chr.11: 65341361-65341361 on Build GRCh38
rs374991419 Chr.11: 65341199-65341199 on Build GRCh38
rs540770158 Chr.11: 65341092-65341092 on Build GRCh38
rs778752988 Chr.11: 65340976-65340976 on Build GRCh38
rs774728257 Chr.11: 65341114-65341114 on Build GRCh38
rs757826187 Chr.11: 65340977-65340977 on Build GRCh38
rs773189453 Chr.11: 65341292-65341292 on Build GRCh38
rs766136251 Chr.11: 65340958-65340958 on Build GRCh38
rs2307265 Chr.11: 65341189-65341189 on Build GRCh38
rs888586244 Chr.11: 65341210-65341210 on Build GRCh38
rs4647579 Chr.11: 65341240-65341247 on Build GRCh38
rs770311911 Chr.11: 65341019-65341019 on Build GRCh38
rs777134002 Chr.11: 65341078-65341078 on Build GRCh38
rs373761595 Chr.11: 65341118-65341118 on Build GRCh38
rs774551990 Chr.11: 65341052-65341052 on Build GRCh38
rs764046003 Chr.11: 65341033-65341033 on Build GRCh38
rs762876735 Chr.11: 65340939-65340939 on Build GRCh38
rs562819064 Chr.11: 65341324-65341324 on Build GRCh38
rs1038990519 Chr.11: 65340968-65340968 on Build GRCh38
rs539245948 Chr.11: 65341242-65341247 on Build GRCh38
rs373233186 Chr.11: 65340947-65340947 on Build GRCh38
rs144611863 Chr.11: 65340981-65340981 on Build GRCh38
rs545359271 Chr.11: 65341342-65341342 on Build GRCh38
rs746637789 Chr.11: 65340998-65340998 on Build GRCh38
rs1000077930 Chr.11: 65341010-65341010 on Build GRCh38
rs754720218 Chr.11: 65341064-65341064 on Build GRCh38
rs1004327117 Chr.11: 65341247-65341247 on Build GRCh38
rs11553935 Chr.11: 65340995-65340995 on Build GRCh38
rs746166076 Chr.11: 65341005-65341005 on Build GRCh38
rs546806064 Chr.11: 65341240-65341240 on Build GRCh38
rs576435120 Chr.11: 65340921-65340921 on Build GRCh38
rs770763995 Chr.11: 65341002-65341002 on Build GRCh38
rs961800521 Chr.11: 65340914-65340914 on Build GRCh38
rs199801105 Chr.11: 65341020-65341020 on Build GRCh38
rs757571169 Chr.11: 65340951-65340951 on Build GRCh38
rs777383722 Chr.11: 65340988-65340988 on Build GRCh38
rs754795615 Chr.11: 65340967-65340967 on Build GRCh38
rs2307267 Chr.11: 65341076-65341076 on Build GRCh38
rs776097797 Chr.11: 65340929-65340929 on Build GRCh38
rs745535715 Chr.11: 65341089-65341089 on Build GRCh38
rs201807910 Chr.11: 65341080-65341080 on Build GRCh38
rs933979262 Chr.11: 65341240-65341246 on Build GRCh38
rs147915994 Chr.11: 65341058-65341058 on Build GRCh38
rs765130489 Chr.11: 65341073-65341073 on Build GRCh38
rs955665927 Chr.11: 65341057-65341057 on Build GRCh38
rs764250607 Chr.11: 65340946-65340946 on Build GRCh38
rs1031353549 Chr.11: 65341021-65341021 on Build GRCh38
rs1007797566 Chr.11: 65340942-65340942 on Build GRCh38
rs1029921172 Chr.11: 65341290-65341290 on Build GRCh38
rs377750990 Chr.11: 65341101-65341101 on Build GRCh38
rs776435445 Chr.11: 65341004-65341004 on Build GRCh38
rs377683764 Chr.11: 65340920-65340920 on Build GRCh38

View Full Product Details