Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65345848-65346346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPF2
Entrez Gene ID
5977
Gene Name
double PHD fingers 2
Gene Aliases
REQ, UBID4, ubi-d4
UniGene
Hs.13495
Gene Chromosome Location
Chr.11: 65333754-65352980 on Build GRCh38
Transcripts
NM_006268.4 XM_005274149.2 XM_017018101.1 XM_017018102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5012368 c.808A>C p.N270H Chr.11: 65345962-65345962 on Build GRCh38
1238724 c.857C>T p.T286M Chr.11: 65346011-65346011 on Build GRCh38
930383 c.959A>G p.Y320C Chr.11: 65346301-65346301 on Build GRCh38
5341674 c.820G>A p.D274N Chr.11: 65345974-65345974 on Build GRCh38
3720479 c.902C>T p.S301L Chr.11: 65346056-65346056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781004664 Chr.11: 65346260-65346260 on Build GRCh38
rs774528841 Chr.11: 65346208-65346208 on Build GRCh38
rs371888353 Chr.11: 65346305-65346305 on Build GRCh38
rs200480588 Chr.11: 65346266-65346266 on Build GRCh38
rs922053318 Chr.11: 65346159-65346159 on Build GRCh38
rs746856930 Chr.11: 65346206-65346206 on Build GRCh38
rs749275482 Chr.11: 65345949-65345949 on Build GRCh38
rs778732858 Chr.11: 65346096-65346096 on Build GRCh38
rs923884322 Chr.11: 65345896-65345896 on Build GRCh38
rs761898626 Chr.11: 65346302-65346302 on Build GRCh38
rs747879173 Chr.11: 65345900-65345900 on Build GRCh38
rs773774048 Chr.11: 65345901-65345901 on Build GRCh38
rs984543629 Chr.11: 65346233-65346233 on Build GRCh38
rs4647589 Chr.11: 65346136-65346136 on Build GRCh38
rs932353789 Chr.11: 65345872-65345872 on Build GRCh38
rs140259644 Chr.11: 65346287-65346287 on Build GRCh38
rs61736579 Chr.11: 65345988-65345988 on Build GRCh38
rs750608775 Chr.11: 65346197-65346198 on Build GRCh38
rs757235832 Chr.11: 65346224-65346224 on Build GRCh38
rs370360359 Chr.11: 65345887-65345887 on Build GRCh38
rs200364248 Chr.11: 65346290-65346290 on Build GRCh38
rs376580739 Chr.11: 65346307-65346307 on Build GRCh38
rs780053425 Chr.11: 65346224-65346227 on Build GRCh38
rs201776355 Chr.11: 65345882-65345882 on Build GRCh38
rs924546294 Chr.11: 65345955-65345955 on Build GRCh38
rs975030996 Chr.11: 65346153-65346153 on Build GRCh38
rs768721684 Chr.11: 65346296-65346296 on Build GRCh38
rs771389355 Chr.11: 65346200-65346200 on Build GRCh38
rs774473883 Chr.11: 65346299-65346299 on Build GRCh38
rs201162180 Chr.11: 65346243-65346243 on Build GRCh38
rs778555341 Chr.11: 65345963-65345963 on Build GRCh38
rs773339078 Chr.11: 65346323-65346323 on Build GRCh38
rs762952357 Chr.11: 65346070-65346070 on Build GRCh38
rs757092533 Chr.11: 65346083-65346084 on Build GRCh38
rs755004702 Chr.11: 65346092-65346092 on Build GRCh38
rs772219515 Chr.11: 65346304-65346304 on Build GRCh38
rs755653992 Chr.11: 65346125-65346139 on Build GRCh38
rs767749748 Chr.11: 65346110-65346110 on Build GRCh38
rs149073109 Chr.11: 65345936-65345936 on Build GRCh38
rs777520682 Chr.11: 65346107-65346107 on Build GRCh38
rs1056149597 Chr.11: 65345970-65345970 on Build GRCh38
rs372536184 Chr.11: 65346254-65346254 on Build GRCh38
rs748726296 Chr.11: 65345886-65345886 on Build GRCh38
rs747337649 Chr.11: 65346012-65346012 on Build GRCh38
rs367850661 Chr.11: 65346110-65346110 on Build GRCh38
rs766152777 Chr.11: 65346213-65346213 on Build GRCh38
rs753973411 Chr.11: 65345933-65345933 on Build GRCh38
rs1027709780 Chr.11: 65346098-65346098 on Build GRCh38
rs915107368 Chr.11: 65345876-65345876 on Build GRCh38
rs770082049 Chr.11: 65346275-65346275 on Build GRCh38
rs752828892 Chr.11: 65345921-65345921 on Build GRCh38
rs373762591 Chr.11: 65346018-65346018 on Build GRCh38
rs780522954 Chr.11: 65346227-65346227 on Build GRCh38
rs761135699 Chr.11: 65345902-65345902 on Build GRCh38
rs202228737 Chr.11: 65346205-65346205 on Build GRCh38
rs751250323 Chr.11: 65346250-65346250 on Build GRCh38
rs765406677 Chr.11: 65345918-65345918 on Build GRCh38
rs1021715939 Chr.11: 65346174-65346174 on Build GRCh38
rs752185421 Chr.11: 65346100-65346100 on Build GRCh38
rs753773852 Chr.11: 65346088-65346088 on Build GRCh38
rs767199700 Chr.11: 65345923-65345923 on Build GRCh38
rs753615132 Chr.11: 65346214-65346214 on Build GRCh38
rs781732459 Chr.11: 65346201-65346201 on Build GRCh38
rs200015058 Chr.11: 65346021-65346021 on Build GRCh38
rs766206157 Chr.11: 65346084-65346084 on Build GRCh38
rs543058250 Chr.11: 65346150-65346150 on Build GRCh38
rs775695232 Chr.11: 65346054-65346054 on Build GRCh38
rs779922590 Chr.11: 65345938-65345938 on Build GRCh38
rs4647588 Chr.11: 65346126-65346126 on Build GRCh38
rs773005168 Chr.11: 65345997-65345997 on Build GRCh38
rs759638525 Chr.11: 65345915-65345915 on Build GRCh38
rs905707004 Chr.11: 65346083-65346083 on Build GRCh38
rs773856468 Chr.11: 65345899-65345899 on Build GRCh38
rs374651671 Chr.11: 65346109-65346109 on Build GRCh38
rs757960859 Chr.11: 65346241-65346241 on Build GRCh38
rs370206123 Chr.11: 65346071-65346071 on Build GRCh38
rs151219453 Chr.11: 65346015-65346015 on Build GRCh38
rs994858962 Chr.11: 65346095-65346095 on Build GRCh38
rs367559374 Chr.11: 65346039-65346039 on Build GRCh38
rs766928686 Chr.11: 65345905-65345905 on Build GRCh38
rs149432821 Chr.11: 65345922-65345922 on Build GRCh38
rs569277444 Chr.11: 65346103-65346103 on Build GRCh38
rs756194613 Chr.11: 65346220-65346221 on Build GRCh38
rs759255085 Chr.11: 65346220-65346220 on Build GRCh38
rs747745445 Chr.11: 65345983-65345983 on Build GRCh38
rs998146229 Chr.11: 65346104-65346104 on Build GRCh38
rs375672962 Chr.11: 65346082-65346082 on Build GRCh38
rs185419600 Chr.11: 65345911-65345911 on Build GRCh38

View Full Product Details