Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65348771-65349269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPF2
Entrez Gene ID
5977
Gene Name
double PHD fingers 2
Gene Aliases
REQ, UBID4, ubi-d4
UniGene
Hs.13495
Gene Chromosome Location
Chr.11: 65333754-65352980 on Build GRCh38
Transcripts
NM_006268.4 XM_005274149.2 XM_017018101.1 XM_017018102.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235477 c.1067G>C p.C356S Chr.11: 65348899-65348899 on Build GRCh38
930384 c.1048C>T p.R350C Chr.11: 65348880-65348880 on Build GRCh38
6238755 c.1045G>T p.D349Y Chr.11: 65348877-65348877 on Build GRCh38
194209 c.1077G>A p.P359P Chr.11: 65348909-65348909 on Build GRCh38
5360873 c.1067G>A p.C356Y Chr.11: 65348899-65348899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767480539 Chr.11: 65348851-65348851 on Build GRCh38
rs188678655 Chr.11: 65348797-65348797 on Build GRCh38
rs922804945 Chr.11: 65348905-65348905 on Build GRCh38
rs552043364 Chr.11: 65348985-65348985 on Build GRCh38
rs568036040 Chr.11: 65349074-65349074 on Build GRCh38
rs747810698 Chr.11: 65349203-65349203 on Build GRCh38
rs961693554 Chr.11: 65348836-65348836 on Build GRCh38
rs1016972091 Chr.11: 65349098-65349098 on Build GRCh38
rs1001776988 Chr.11: 65348827-65348827 on Build GRCh38
rs200674111 Chr.11: 65349180-65349186 on Build GRCh38
rs879152709 Chr.11: 65349012-65349012 on Build GRCh38
rs1042579349 Chr.11: 65349013-65349013 on Build GRCh38
rs756132079 Chr.11: 65348924-65348924 on Build GRCh38
rs2307266 Chr.11: 65349020-65349020 on Build GRCh38
rs964531733 Chr.11: 65349049-65349049 on Build GRCh38
rs959472661 Chr.11: 65349081-65349081 on Build GRCh38
rs766070534 Chr.11: 65348876-65348876 on Build GRCh38
rs770999788 Chr.11: 65348955-65348955 on Build GRCh38
rs755370914 Chr.11: 65348888-65348888 on Build GRCh38
rs547798630 Chr.11: 65349062-65349062 on Build GRCh38
rs778602589 Chr.11: 65349172-65349172 on Build GRCh38
rs748703811 Chr.11: 65348927-65348927 on Build GRCh38
rs749048277 Chr.11: 65348975-65348975 on Build GRCh38
rs532265156 Chr.11: 65348939-65348939 on Build GRCh38
rs777783511 Chr.11: 65348948-65348948 on Build GRCh38
rs973014153 Chr.11: 65349173-65349173 on Build GRCh38
rs776916116 Chr.11: 65348963-65348963 on Build GRCh38
rs397843675 Chr.11: 65349093-65349094 on Build GRCh38
rs985652205 Chr.11: 65349213-65349213 on Build GRCh38
rs755784167 Chr.11: 65348855-65348855 on Build GRCh38
rs570223845 Chr.11: 65349209-65349209 on Build GRCh38
rs766982957 Chr.11: 65348843-65348843 on Build GRCh38
rs749244157 Chr.11: 65348970-65348973 on Build GRCh38
rs764055932 Chr.11: 65348829-65348829 on Build GRCh38
rs762855033 Chr.11: 65348826-65348826 on Build GRCh38
rs538912420 Chr.11: 65349212-65349212 on Build GRCh38
rs4647592 Chr.11: 65349091-65349092 on Build GRCh38
rs747240416 Chr.11: 65348953-65348953 on Build GRCh38
rs750317661 Chr.11: 65348909-65348909 on Build GRCh38
rs373186765 Chr.11: 65348877-65348877 on Build GRCh38
rs35405661 Chr.11: 65348922-65348922 on Build GRCh38
rs988514123 Chr.11: 65349109-65349109 on Build GRCh38
rs376807610 Chr.11: 65348808-65348808 on Build GRCh38
rs758475381 Chr.11: 65348944-65348944 on Build GRCh38
rs765171222 Chr.11: 65348997-65348997 on Build GRCh38
rs559569168 Chr.11: 65348814-65348814 on Build GRCh38
rs775204450 Chr.11: 65348821-65348821 on Build GRCh38
rs550067165 Chr.11: 65349117-65349117 on Build GRCh38
rs774458671 Chr.11: 65348834-65348834 on Build GRCh38
rs369824078 Chr.11: 65348817-65348817 on Build GRCh38
rs761354272 Chr.11: 65348841-65348841 on Build GRCh38
rs559269423 Chr.11: 65348998-65348998 on Build GRCh38
rs1018817721 Chr.11: 65349026-65349026 on Build GRCh38
rs1053469725 Chr.11: 65348996-65348996 on Build GRCh38
rs180836536 Chr.11: 65349089-65349089 on Build GRCh38

View Full Product Details