Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65917787-65918283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf68
Entrez Gene ID
83638
Gene Name
chromosome 11 open reading frame 68
Gene Aliases
BLES03, P5326
UniGene
Hs.433573
Gene Chromosome Location
Chr.11: 65916810-65919060 on Build GRCh38
Transcripts
NM_001135635.1 NM_031450.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5721735 c.269C>T p.S90F Chr.11: 65917946-65917946 on Build GRCh38
6454311 c.549C>T p.H183H Chr.11: 65917792-65917792 on Build GRCh38
6245319 c.422C>A p.A141D Chr.11: 65917919-65917919 on Build GRCh38
7285574 c.335C>T p.P112L Chr.11: 65917880-65917880 on Build GRCh38
4987092 c.203C>G p.P68R Chr.11: 65918012-65918012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907223202 Chr.11: 65917945-65917945 on Build GRCh38
rs747341715 Chr.11: 65917879-65917879 on Build GRCh38
rs961495072 Chr.11: 65918214-65918214 on Build GRCh38
rs762690740 Chr.11: 65918159-65918159 on Build GRCh38
rs142998377 Chr.11: 65917815-65917815 on Build GRCh38
rs150568483 Chr.11: 65917999-65917999 on Build GRCh38
rs769726744 Chr.11: 65918152-65918152 on Build GRCh38
rs748221899 Chr.11: 65918140-65918140 on Build GRCh38
rs768480845 Chr.11: 65918182-65918182 on Build GRCh38
rs753540562 Chr.11: 65917852-65917852 on Build GRCh38
rs754063393 Chr.11: 65918114-65918114 on Build GRCh38
rs757498619 Chr.11: 65917992-65917992 on Build GRCh38
rs764843542 Chr.11: 65917966-65917966 on Build GRCh38
rs139058880 Chr.11: 65918024-65918024 on Build GRCh38
rs745631066 Chr.11: 65918149-65918149 on Build GRCh38
rs758948370 Chr.11: 65917872-65917872 on Build GRCh38
rs764650988 Chr.11: 65917982-65917982 on Build GRCh38
rs766199663 Chr.11: 65917944-65917944 on Build GRCh38
rs572025542 Chr.11: 65918185-65918185 on Build GRCh38
rs750770390 Chr.11: 65918090-65918090 on Build GRCh38
rs761032572 Chr.11: 65918226-65918226 on Build GRCh38
rs985069815 Chr.11: 65918253-65918253 on Build GRCh38
rs752309095 Chr.11: 65917869-65917869 on Build GRCh38
rs762368596 Chr.11: 65918086-65918086 on Build GRCh38
rs1025362061 Chr.11: 65918088-65918088 on Build GRCh38
rs565007272 Chr.11: 65918110-65918110 on Build GRCh38
rs368301483 Chr.11: 65917844-65917844 on Build GRCh38
rs776462496 Chr.11: 65918052-65918052 on Build GRCh38
rs749419997 Chr.11: 65917925-65917925 on Build GRCh38
rs767960379 Chr.11: 65918087-65918087 on Build GRCh38
rs765481469 Chr.11: 65918251-65918251 on Build GRCh38
rs759287565 Chr.11: 65918057-65918057 on Build GRCh38
rs551517220 Chr.11: 65917861-65917861 on Build GRCh38
rs574139902 Chr.11: 65917995-65917995 on Build GRCh38
rs763705751 Chr.11: 65918081-65918081 on Build GRCh38
rs775627350 Chr.11: 65917919-65917919 on Build GRCh38
rs773978540 Chr.11: 65918190-65918190 on Build GRCh38
rs752220956 Chr.11: 65917967-65917967 on Build GRCh38
rs536451862 Chr.11: 65918017-65918017 on Build GRCh38
rs775219397 Chr.11: 65918158-65918158 on Build GRCh38
rs755166518 Chr.11: 65918249-65918249 on Build GRCh38
rs186200069 Chr.11: 65917918-65917918 on Build GRCh38
rs1024451140 Chr.11: 65917996-65917996 on Build GRCh38
rs925199702 Chr.11: 65918244-65918244 on Build GRCh38
rs754376904 Chr.11: 65917860-65917860 on Build GRCh38
rs759209496 Chr.11: 65917962-65917962 on Build GRCh38
rs375631477 Chr.11: 65918221-65918221 on Build GRCh38
rs778946994 Chr.11: 65918137-65918137 on Build GRCh38
rs35868735 Chr.11: 65918174-65918174 on Build GRCh38
rs781019646 Chr.11: 65918148-65918148 on Build GRCh38
rs755258694 Chr.11: 65918131-65918131 on Build GRCh38
rs775307507 Chr.11: 65918080-65918080 on Build GRCh38
rs757665382 Chr.11: 65917880-65917880 on Build GRCh38
rs770867726 Chr.11: 65918143-65918143 on Build GRCh38
rs374355629 Chr.11: 65917837-65917837 on Build GRCh38
rs955315431 Chr.11: 65918179-65918179 on Build GRCh38
rs754255448 Chr.11: 65918236-65918236 on Build GRCh38
rs191917397 Chr.11: 65918108-65918108 on Build GRCh38
rs779731203 Chr.11: 65918162-65918185 on Build GRCh38
rs73496433 Chr.11: 65918255-65918255 on Build GRCh38
rs962365468 Chr.11: 65918049-65918049 on Build GRCh38
rs776945105 Chr.11: 65917810-65917810 on Build GRCh38
rs771955929 Chr.11: 65917939-65917939 on Build GRCh38
rs750905797 Chr.11: 65918224-65918224 on Build GRCh38
rs760404266 Chr.11: 65917941-65917941 on Build GRCh38
rs752129327 Chr.11: 65917990-65917990 on Build GRCh38
rs767500837 Chr.11: 65917842-65917842 on Build GRCh38
rs758015989 Chr.11: 65917970-65917970 on Build GRCh38
rs1016551148 Chr.11: 65918043-65918043 on Build GRCh38
rs371675950 Chr.11: 65918031-65918031 on Build GRCh38
rs745890262 Chr.11: 65917811-65917811 on Build GRCh38
rs571594456 Chr.11: 65917881-65917881 on Build GRCh38
rs776377534 Chr.11: 65917954-65917954 on Build GRCh38
rs778468116 Chr.11: 65917877-65917877 on Build GRCh38
rs143889365 Chr.11: 65917898-65917898 on Build GRCh38
rs771807885 Chr.11: 65918212-65918212 on Build GRCh38
rs7947619 Chr.11: 65917822-65917822 on Build GRCh38
rs11542177 Chr.11: 65917846-65917846 on Build GRCh38
rs368300054 Chr.11: 65918232-65918232 on Build GRCh38
rs765896274 Chr.11: 65917849-65917849 on Build GRCh38
rs768736153 Chr.11: 65918026-65918026 on Build GRCh38
rs774091393 Chr.11: 65917834-65917834 on Build GRCh38
rs777619552 Chr.11: 65918030-65918030 on Build GRCh38
rs780284825 Chr.11: 65918022-65918022 on Build GRCh38
rs769451839 Chr.11: 65918064-65918064 on Build GRCh38
rs868288399 Chr.11: 65918156-65918156 on Build GRCh38
rs139550883 Chr.11: 65918038-65918038 on Build GRCh38
rs751182740 Chr.11: 65918082-65918082 on Build GRCh38

View Full Product Details