Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66564475-66564985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSF
Entrez Gene ID
8722
Gene Name
cathepsin F
Gene Aliases
CATSF, CLN13
UniGene
Hs.11590
Gene Chromosome Location
Chr.11: 66563463-66568576 on Build GRCh38
Transcripts
NM_003793.3 XM_011545328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4035751 c.1095C>A p.S365S Chr.11: 66564957-66564957 on Build GRCh38
7100054 c.1164G>C p.Q388H Chr.11: 66564888-66564888 on Build GRCh38
169726 c.1252C>T p.R418C Chr.11: 66564627-66564627 on Build GRCh38
7100053 c.1188G>C p.K396N Chr.11: 66564784-66564784 on Build GRCh38
7100832 c.1166-1G>A p.? Chr.11: 66564807-66564807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005776027 Chr.11: 66564517-66564517 on Build GRCh38
rs754874145 Chr.11: 66564795-66564795 on Build GRCh38
rs755154475 Chr.11: 66564566-66564566 on Build GRCh38
rs1055863767 Chr.11: 66564936-66564936 on Build GRCh38
rs770422849 Chr.11: 66564847-66564847 on Build GRCh38
rs200426008 Chr.11: 66564636-66564636 on Build GRCh38
rs889550300 Chr.11: 66564800-66564800 on Build GRCh38
rs764163045 Chr.11: 66564903-66564903 on Build GRCh38
rs752911567 Chr.11: 66564655-66564655 on Build GRCh38
rs768570314 Chr.11: 66564676-66564676 on Build GRCh38
rs201295932 Chr.11: 66564617-66564617 on Build GRCh38
rs199830949 Chr.11: 66564661-66564661 on Build GRCh38
rs746640955 Chr.11: 66564960-66564960 on Build GRCh38
rs148080813 Chr.11: 66564919-66564919 on Build GRCh38
rs778969360 Chr.11: 66564583-66564583 on Build GRCh38
rs368422217 Chr.11: 66564549-66564549 on Build GRCh38
rs780211076 Chr.11: 66564774-66564774 on Build GRCh38
rs781257663 Chr.11: 66564600-66564600 on Build GRCh38
rs770919773 Chr.11: 66564682-66564684 on Build GRCh38
rs534376817 Chr.11: 66564557-66564557 on Build GRCh38
rs778856523 Chr.11: 66564935-66564935 on Build GRCh38
rs985016814 Chr.11: 66564532-66564532 on Build GRCh38
rs772558572 Chr.11: 66564693-66564693 on Build GRCh38
rs745759723 Chr.11: 66564603-66564603 on Build GRCh38
rs748168574 Chr.11: 66564584-66564584 on Build GRCh38
rs937618113 Chr.11: 66564934-66564934 on Build GRCh38
rs199885470 Chr.11: 66564844-66564844 on Build GRCh38
rs775373144 Chr.11: 66564613-66564613 on Build GRCh38
rs28464796 Chr.11: 66564618-66564618 on Build GRCh38
rs990285672 Chr.11: 66564851-66564851 on Build GRCh38
rs758200986 Chr.11: 66564948-66564948 on Build GRCh38
rs751789480 Chr.11: 66564905-66564905 on Build GRCh38
rs1030911295 Chr.11: 66564869-66564869 on Build GRCh38
rs143674429 Chr.11: 66564912-66564912 on Build GRCh38
rs113541570 Chr.11: 66564675-66564675 on Build GRCh38
rs760013945 Chr.11: 66564873-66564873 on Build GRCh38
rs367661681 Chr.11: 66564692-66564692 on Build GRCh38
rs761251097 Chr.11: 66564703-66564703 on Build GRCh38
rs955332400 Chr.11: 66564784-66564784 on Build GRCh38
rs762721365 Chr.11: 66564614-66564614 on Build GRCh38
rs375050229 Chr.11: 66564522-66564522 on Build GRCh38
rs1034531584 Chr.11: 66564513-66564513 on Build GRCh38
rs572277115 Chr.11: 66564729-66564729 on Build GRCh38
rs549857580 Chr.11: 66564838-66564838 on Build GRCh38
rs149687246 Chr.11: 66564565-66564565 on Build GRCh38
rs1055013673 Chr.11: 66564634-66564634 on Build GRCh38
rs768353764 Chr.11: 66564616-66564616 on Build GRCh38
rs139027846 Chr.11: 66564769-66564769 on Build GRCh38
rs200788870 Chr.11: 66564836-66564836 on Build GRCh38
rs372771218 Chr.11: 66564704-66564704 on Build GRCh38
rs779996406 Chr.11: 66564672-66564672 on Build GRCh38
rs776552555 Chr.11: 66564712-66564717 on Build GRCh38
rs778805546 Chr.11: 66564805-66564805 on Build GRCh38
rs116329758 Chr.11: 66564894-66564894 on Build GRCh38
rs749210204 Chr.11: 66564790-66564790 on Build GRCh38
rs747839565 Chr.11: 66564943-66564943 on Build GRCh38
rs150922871 Chr.11: 66564558-66564558 on Build GRCh38
rs745744669 Chr.11: 66564780-66564781 on Build GRCh38
rs754331066 Chr.11: 66564719-66564719 on Build GRCh38
rs961878630 Chr.11: 66564735-66564735 on Build GRCh38
rs201500574 Chr.11: 66564952-66564952 on Build GRCh38
rs779164010 Chr.11: 66564642-66564642 on Build GRCh38
rs777763116 Chr.11: 66564659-66564661 on Build GRCh38
rs758499333 Chr.11: 66564754-66564754 on Build GRCh38
rs571987848 Chr.11: 66564502-66564502 on Build GRCh38
rs867917295 Chr.11: 66564570-66564570 on Build GRCh38
rs985854450 Chr.11: 66564825-66564825 on Build GRCh38
rs747078300 Chr.11: 66564668-66564668 on Build GRCh38
rs759093942 Chr.11: 66564637-66564637 on Build GRCh38
rs752964534 Chr.11: 66564745-66564745 on Build GRCh38
rs1021882738 Chr.11: 66564576-66564577 on Build GRCh38
rs761039140 Chr.11: 66564627-66564627 on Build GRCh38
rs141345438 Chr.11: 66564626-66564626 on Build GRCh38
rs370667878 Chr.11: 66564663-66564663 on Build GRCh38
rs758542993 Chr.11: 66564656-66564656 on Build GRCh38
rs933983229 Chr.11: 66564596-66564596 on Build GRCh38
rs769678799 Chr.11: 66564610-66564610 on Build GRCh38
rs373027690 Chr.11: 66564641-66564642 on Build GRCh38
rs201693863 Chr.11: 66564816-66564816 on Build GRCh38
rs772030326 Chr.11: 66564593-66564593 on Build GRCh38
rs145087378 Chr.11: 66564641-66564641 on Build GRCh38
rs373451370 Chr.11: 66564848-66564848 on Build GRCh38
rs999428377 Chr.11: 66564861-66564862 on Build GRCh38
rs768131664 Chr.11: 66564540-66564540 on Build GRCh38
rs757455465 Chr.11: 66564911-66564911 on Build GRCh38
rs761351900 Chr.11: 66564685-66564685 on Build GRCh38
rs1001199791 Chr.11: 66564511-66564511 on Build GRCh38
rs765612253 Chr.11: 66564885-66564885 on Build GRCh38
rs764262853 Chr.11: 66564758-66564758 on Build GRCh38
rs140795906 Chr.11: 66564564-66564564 on Build GRCh38
rs980109275 Chr.11: 66564662-66564662 on Build GRCh38
rs752410509 Chr.11: 66564664-66564664 on Build GRCh38
rs769495722 Chr.11: 66564801-66564801 on Build GRCh38
rs960864612 Chr.11: 66564876-66564876 on Build GRCh38
rs375891471 Chr.11: 66564768-66564768 on Build GRCh38
rs759960583 Chr.11: 66564728-66564728 on Build GRCh38
rs775310814 Chr.11: 66564878-66564879 on Build GRCh38
rs913357286 Chr.11: 66564861-66564861 on Build GRCh38
rs200760567 Chr.11: 66564961-66564961 on Build GRCh38
rs774192403 Chr.11: 66564684-66564684 on Build GRCh38
rs553298368 Chr.11: 66564629-66564629 on Build GRCh38

View Full Product Details