Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67119663-67120166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
UniGene
Hs.124147
Gene Chromosome Location
Chr.11: 67119210-67258082 on Build GRCh38
Transcripts
NM_012308.2 NR_027473.1 XM_011544860.2

SNP Detail

SNP ID Chromosome Location
rs943582662 Chr.11: 67120048-67120048 on Build GRCh38
rs1019248727 Chr.11: 67119787-67119787 on Build GRCh38
rs1009147664 Chr.11: 67119786-67119786 on Build GRCh38
rs551712101 Chr.11: 67119912-67119912 on Build GRCh38
rs1033608858 Chr.11: 67119830-67119830 on Build GRCh38
rs909667966 Chr.11: 67119985-67119985 on Build GRCh38
rs201622499 Chr.11: 67120072-67120072 on Build GRCh38
rs570899139 Chr.11: 67119919-67119919 on Build GRCh38
rs370928737 Chr.11: 67119804-67119804 on Build GRCh38
rs932243760 Chr.11: 67120126-67120126 on Build GRCh38
rs747094559 Chr.11: 67119978-67119978 on Build GRCh38
rs547225440 Chr.11: 67119810-67119810 on Build GRCh38
rs988863144 Chr.11: 67119838-67119838 on Build GRCh38
rs547107231 Chr.11: 67120024-67120024 on Build GRCh38
rs144920685 Chr.11: 67120060-67120060 on Build GRCh38
rs749689326 Chr.11: 67119859-67119859 on Build GRCh38
rs140960382 Chr.11: 67120017-67120017 on Build GRCh38
rs1003180441 Chr.11: 67120090-67120090 on Build GRCh38
rs956370003 Chr.11: 67119750-67119762 on Build GRCh38
rs974407212 Chr.11: 67120059-67120059 on Build GRCh38
rs957805019 Chr.11: 67120007-67120007 on Build GRCh38
rs763323431 Chr.11: 67120034-67120035 on Build GRCh38
rs532039802 Chr.11: 67119932-67119932 on Build GRCh38
rs982511412 Chr.11: 67120064-67120064 on Build GRCh38
rs970401441 Chr.11: 67119949-67119949 on Build GRCh38
rs1038180703 Chr.11: 67119903-67119903 on Build GRCh38
rs910817785 Chr.11: 67120045-67120045 on Build GRCh38
rs568292835 Chr.11: 67120025-67120025 on Build GRCh38
rs777929754 Chr.11: 67119972-67119972 on Build GRCh38
rs754182468 Chr.11: 67119858-67119858 on Build GRCh38
rs1033274298 Chr.11: 67120103-67120103 on Build GRCh38
rs547238122 Chr.11: 67119753-67119753 on Build GRCh38
rs984862336 Chr.11: 67119737-67119737 on Build GRCh38
rs528593391 Chr.11: 67120015-67120015 on Build GRCh38
rs994344335 Chr.11: 67119767-67119767 on Build GRCh38
rs1054333246 Chr.11: 67119918-67119918 on Build GRCh38
rs996388610 Chr.11: 67119819-67119819 on Build GRCh38
rs945556911 Chr.11: 67120067-67120067 on Build GRCh38
rs981304721 Chr.11: 67119852-67119852 on Build GRCh38
rs895071056 Chr.11: 67120132-67120132 on Build GRCh38
rs1003147936 Chr.11: 67119955-67119955 on Build GRCh38
rs894002221 Chr.11: 67120065-67120065 on Build GRCh38
rs958612375 Chr.11: 67120057-67120057 on Build GRCh38
rs565900476 Chr.11: 67119871-67119871 on Build GRCh38
rs568862860 Chr.11: 67119908-67119908 on Build GRCh38
rs1033147748 Chr.11: 67120004-67120004 on Build GRCh38
rs957437916 Chr.11: 67119836-67119836 on Build GRCh38
rs374579802 Chr.11: 67119992-67119992 on Build GRCh38
rs539699491 Chr.11: 67119892-67119892 on Build GRCh38
rs1007752603 Chr.11: 67119905-67119905 on Build GRCh38
rs910387344 Chr.11: 67119802-67119802 on Build GRCh38
rs912344471 Chr.11: 67120068-67120068 on Build GRCh38
rs913226990 Chr.11: 67119840-67119840 on Build GRCh38
rs1046727321 Chr.11: 67119775-67119775 on Build GRCh38
rs1052048660 Chr.11: 67119876-67119876 on Build GRCh38
rs906272844 Chr.11: 67120071-67120071 on Build GRCh38
rs547878162 Chr.11: 67119698-67119698 on Build GRCh38
rs374396229 Chr.11: 67120062-67120062 on Build GRCh38
rs919298052 Chr.11: 67119869-67119869 on Build GRCh38
rs375954845 Chr.11: 67120069-67120069 on Build GRCh38
rs937215101 Chr.11: 67119744-67119744 on Build GRCh38
rs899505379 Chr.11: 67120070-67120070 on Build GRCh38
rs987447241 Chr.11: 67120034-67120034 on Build GRCh38
rs755267187 Chr.11: 67119862-67119862 on Build GRCh38
rs748303853 Chr.11: 67119898-67119898 on Build GRCh38
rs900725461 Chr.11: 67119965-67119967 on Build GRCh38
rs933332811 Chr.11: 67120033-67120033 on Build GRCh38
rs1009689984 Chr.11: 67119941-67119941 on Build GRCh38
rs929709459 Chr.11: 67119907-67119907 on Build GRCh38
rs890805012 Chr.11: 67119781-67119781 on Build GRCh38
rs1020699869 Chr.11: 67119942-67119942 on Build GRCh38
rs1033579875 Chr.11: 67119962-67119962 on Build GRCh38
rs927127891 Chr.11: 67119692-67119692 on Build GRCh38

View Full Product Details