Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67266695-67267197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK2
Entrez Gene ID
156
Gene Name
G protein-coupled receptor kinase 2
Gene Aliases
ADRBK1, BARK1, BETA-ARK1
UniGene
Hs.83636
Gene Chromosome Location
Chr.11: 67266420-67286558 on Build GRCh38
Transcripts
NM_001619.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3980606 c.89A>G p.K30R Chr.11: 67266788-67266788 on Build GRCh38
3752678 c.96C>A p.I32I Chr.11: 67266795-67266795 on Build GRCh38
6374598 c.71C>T p.P24L Chr.11: 67266770-67266770 on Build GRCh38
6391361 c.61A>G p.K21E Chr.11: 67266760-67266760 on Build GRCh38
5373361 c.31G>T p.V11L Chr.11: 67266730-67266730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047999888 Chr.11: 67267158-67267158 on Build GRCh38
rs549807985 Chr.11: 67267050-67267050 on Build GRCh38
rs1008684744 Chr.11: 67266908-67266908 on Build GRCh38
rs368782127 Chr.11: 67266832-67266832 on Build GRCh38
rs535612287 Chr.11: 67267123-67267123 on Build GRCh38
rs1010588179 Chr.11: 67266999-67266999 on Build GRCh38
rs1030770779 Chr.11: 67266883-67266883 on Build GRCh38
rs1037737654 Chr.11: 67267119-67267119 on Build GRCh38
rs1017361457 Chr.11: 67266931-67266931 on Build GRCh38
rs775578042 Chr.11: 67266799-67266799 on Build GRCh38
rs999381808 Chr.11: 67266881-67266881 on Build GRCh38
rs889818768 Chr.11: 67266840-67266840 on Build GRCh38
rs758993559 Chr.11: 67266726-67266726 on Build GRCh38
rs916016254 Chr.11: 67267093-67267093 on Build GRCh38
rs910121977 Chr.11: 67266781-67266781 on Build GRCh38
rs868320916 Chr.11: 67266764-67266764 on Build GRCh38
rs749318434 Chr.11: 67266825-67266825 on Build GRCh38
rs778398889 Chr.11: 67266738-67266738 on Build GRCh38
rs920576844 Chr.11: 67267138-67267138 on Build GRCh38
rs977958972 Chr.11: 67267075-67267075 on Build GRCh38
rs772047949 Chr.11: 67266829-67266829 on Build GRCh38
rs963259198 Chr.11: 67267040-67267040 on Build GRCh38
rs12805999 Chr.11: 67266986-67266986 on Build GRCh38
rs781538949 Chr.11: 67266753-67266753 on Build GRCh38
rs1022347780 Chr.11: 67267008-67267008 on Build GRCh38
rs747444658 Chr.11: 67266741-67266741 on Build GRCh38
rs537446782 Chr.11: 67267001-67267001 on Build GRCh38
rs934132170 Chr.11: 67267052-67267052 on Build GRCh38
rs991024234 Chr.11: 67267087-67267087 on Build GRCh38
rs960511478 Chr.11: 67266895-67266895 on Build GRCh38
rs2228418 Chr.11: 67266795-67266795 on Build GRCh38
rs890676965 Chr.11: 67267162-67267162 on Build GRCh38
rs780957979 Chr.11: 67267084-67267084 on Build GRCh38
rs763738031 Chr.11: 67266833-67266854 on Build GRCh38
rs1037573191 Chr.11: 67266826-67266826 on Build GRCh38
rs771198173 Chr.11: 67266747-67266747 on Build GRCh38
rs370301772 Chr.11: 67266721-67266721 on Build GRCh38
rs2228418 Chr.11: 67266795-67266795 on Build GRCh38
rs1007292740 Chr.11: 67266857-67266857 on Build GRCh38
rs757163712 Chr.11: 67267056-67267056 on Build GRCh38
rs986916050 Chr.11: 67267053-67267053 on Build GRCh38
rs927837295 Chr.11: 67267082-67267082 on Build GRCh38
rs903736576 Chr.11: 67266878-67266878 on Build GRCh38
rs972943948 Chr.11: 67267045-67267045 on Build GRCh38
rs867482919 Chr.11: 67266722-67266722 on Build GRCh38
rs752062920 Chr.11: 67267033-67267033 on Build GRCh38
rs746094488 Chr.11: 67266788-67266788 on Build GRCh38
rs910141346 Chr.11: 67266961-67266961 on Build GRCh38
rs964776281 Chr.11: 67266987-67266987 on Build GRCh38
rs918688318 Chr.11: 67266830-67266830 on Build GRCh38
rs929582738 Chr.11: 67267157-67267157 on Build GRCh38
rs969121084 Chr.11: 67266933-67266933 on Build GRCh38
rs568079394 Chr.11: 67267083-67267083 on Build GRCh38
rs762658327 Chr.11: 67266831-67266858 on Build GRCh38
rs942726940 Chr.11: 67267060-67267060 on Build GRCh38
rs948786066 Chr.11: 67267106-67267106 on Build GRCh38
rs1043653166 Chr.11: 67266875-67266875 on Build GRCh38
rs570109600 Chr.11: 67266848-67266848 on Build GRCh38
rs984822742 Chr.11: 67266952-67266952 on Build GRCh38

View Full Product Details