Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104998950-104999521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP5
Entrez Gene ID
838
Gene Name
caspase 5
Gene Aliases
ICE(rel)III, ICEREL-III, ICH-3
UniGene
Hs.213327
Gene Chromosome Location
Chr.11: 104994240-105023902 on Build GRCh38
Transcripts
NM_001136109.1 NM_001136110.1 NM_001136112.1 NM_004347.3 NR_024239.1 NR_036562.1 XM_011543021.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6486723 c.905-1G>A p.? Chr.11: 104999029-104999029 on Build GRCh38
685724 c.975G>A p.E325E Chr.11: 104998958-104998958 on Build GRCh38
6486722 c.992-1G>A p.? Chr.11: 104999029-104999029 on Build GRCh38
1146541 c.1062G>A p.E354E Chr.11: 104998958-104998958 on Build GRCh38
139927 c.959C>T p.S320F Chr.11: 104998974-104998974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375504198 Chr.11: 104999059-104999059 on Build GRCh38
rs1025301160 Chr.11: 104999351-104999351 on Build GRCh38
rs1039617408 Chr.11: 104999243-104999243 on Build GRCh38
rs908306385 Chr.11: 104999249-104999250 on Build GRCh38
rs1041836604 Chr.11: 104999081-104999081 on Build GRCh38
rs369278753 Chr.11: 104999058-104999058 on Build GRCh38
rs1021350188 Chr.11: 104999447-104999447 on Build GRCh38
rs947386153 Chr.11: 104999210-104999210 on Build GRCh38
rs766075695 Chr.11: 104999414-104999414 on Build GRCh38
rs776564419 Chr.11: 104999055-104999055 on Build GRCh38
rs751238801 Chr.11: 104999418-104999418 on Build GRCh38
rs535175391 Chr.11: 104999370-104999370 on Build GRCh38
rs986667757 Chr.11: 104999193-104999193 on Build GRCh38
rs560668207 Chr.11: 104999094-104999094 on Build GRCh38
rs949183355 Chr.11: 104999308-104999308 on Build GRCh38
rs523104 Chr.11: 104998981-104998981 on Build GRCh38
rs768348669 Chr.11: 104999052-104999052 on Build GRCh38
rs778430065 Chr.11: 104999017-104999017 on Build GRCh38
rs111671737 Chr.11: 104999176-104999176 on Build GRCh38
rs903244402 Chr.11: 104999087-104999087 on Build GRCh38
rs548584696 Chr.11: 104999213-104999213 on Build GRCh38
rs562335827 Chr.11: 104999048-104999049 on Build GRCh38
rs140164381 Chr.11: 104998977-104998977 on Build GRCh38
rs779604515 Chr.11: 104999037-104999037 on Build GRCh38
rs547243060 Chr.11: 104999228-104999228 on Build GRCh38
rs377448726 Chr.11: 104999074-104999075 on Build GRCh38
rs981207713 Chr.11: 104999180-104999180 on Build GRCh38
rs886774750 Chr.11: 104999361-104999361 on Build GRCh38
rs562983297 Chr.11: 104999127-104999127 on Build GRCh38
rs772991467 Chr.11: 104999208-104999208 on Build GRCh38
rs1009582537 Chr.11: 104999426-104999426 on Build GRCh38
rs540897946 Chr.11: 104999034-104999034 on Build GRCh38
rs999368692 Chr.11: 104999096-104999096 on Build GRCh38
rs566244155 Chr.11: 104999049-104999049 on Build GRCh38
rs549645594 Chr.11: 104999343-104999343 on Build GRCh38
rs746604160 Chr.11: 104999044-104999044 on Build GRCh38
rs748709161 Chr.11: 104999058-104999059 on Build GRCh38
rs45574231 Chr.11: 104999229-104999229 on Build GRCh38
rs190341219 Chr.11: 104999353-104999353 on Build GRCh38
rs760316345 Chr.11: 104999199-104999199 on Build GRCh38
rs995112829 Chr.11: 104999148-104999148 on Build GRCh38
rs200038316 Chr.11: 104998992-104998992 on Build GRCh38
rs1046168517 Chr.11: 104999316-104999316 on Build GRCh38
rs765206988 Chr.11: 104999207-104999207 on Build GRCh38
rs45537133 Chr.11: 104999404-104999404 on Build GRCh38
rs767168033 Chr.11: 104999470-104999470 on Build GRCh38
rs1003061306 Chr.11: 104999390-104999390 on Build GRCh38
rs1051337273 Chr.11: 104999100-104999100 on Build GRCh38
rs1036308242 Chr.11: 104999189-104999189 on Build GRCh38
rs546019343 Chr.11: 104999117-104999117 on Build GRCh38
rs1021043745 Chr.11: 104999139-104999139 on Build GRCh38
rs2511327 Chr.11: 104999463-104999463 on Build GRCh38
rs968924956 Chr.11: 104999451-104999451 on Build GRCh38
rs184840151 Chr.11: 104999108-104999108 on Build GRCh38
rs933432970 Chr.11: 104999221-104999221 on Build GRCh38
rs770608807 Chr.11: 104999383-104999383 on Build GRCh38
rs1025353522 Chr.11: 104999158-104999158 on Build GRCh38
rs376032189 Chr.11: 104999216-104999216 on Build GRCh38
rs776648653 Chr.11: 104999185-104999185 on Build GRCh38
rs771560676 Chr.11: 104999036-104999036 on Build GRCh38
rs539612691 Chr.11: 104999240-104999240 on Build GRCh38
rs768724161 Chr.11: 104999131-104999131 on Build GRCh38
rs531636004 Chr.11: 104999195-104999195 on Build GRCh38
rs773238221 Chr.11: 104999072-104999072 on Build GRCh38
rs942533988 Chr.11: 104999230-104999230 on Build GRCh38
rs45605936 Chr.11: 104999217-104999217 on Build GRCh38
rs762979012 Chr.11: 104999078-104999078 on Build GRCh38
rs544756402 Chr.11: 104999498-104999498 on Build GRCh38
rs766418127 Chr.11: 104999079-104999079 on Build GRCh38
rs991631269 Chr.11: 104999417-104999417 on Build GRCh38
rs1054041486 Chr.11: 104999415-104999415 on Build GRCh38
rs958062369 Chr.11: 104999109-104999109 on Build GRCh38
rs898380104 Chr.11: 104999266-104999266 on Build GRCh38
rs995144129 Chr.11: 104999332-104999332 on Build GRCh38
rs748136753 Chr.11: 104999058-104999058 on Build GRCh38
rs756607382 Chr.11: 104999014-104999014 on Build GRCh38

View Full Product Details