Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132771170-132771678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA3
Entrez Gene ID
2802
Gene Name
golgin A3
Gene Aliases
GCP170, MEA-2
UniGene
Hs.507333
Gene Chromosome Location
Chr.12: 132768909-132829131 on Build GRCh38
Transcripts
NM_005895.3 XM_005266164.4 XM_005266167.4 XM_006719736.3 XM_006719737.3 XM_011534793.2 XM_011534794.2

SNP Detail

SNP ID Chromosome Location
rs756739673 Chr.12: 132771622-132771622 on Build GRCh38
rs969590370 Chr.12: 132771199-132771199 on Build GRCh38
rs549307026 Chr.12: 132771272-132771272 on Build GRCh38
rs753727331 Chr.12: 132771602-132771602 on Build GRCh38
rs908729571 Chr.12: 132771352-132771352 on Build GRCh38
rs919882654 Chr.12: 132771373-132771373 on Build GRCh38
rs1013257300 Chr.12: 132771441-132771441 on Build GRCh38
rs892306241 Chr.12: 132771530-132771530 on Build GRCh38
rs116969103 Chr.12: 132771301-132771301 on Build GRCh38
rs1043946874 Chr.12: 132771328-132771328 on Build GRCh38
rs918553790 Chr.12: 132771271-132771271 on Build GRCh38
rs961901797 Chr.12: 132771361-132771361 on Build GRCh38
rs1028056457 Chr.12: 132771637-132771637 on Build GRCh38
rs961864309 Chr.12: 132771615-132771615 on Build GRCh38
rs984630359 Chr.12: 132771218-132771218 on Build GRCh38
rs529263331 Chr.12: 132771254-132771254 on Build GRCh38
rs573620834 Chr.12: 132771425-132771425 on Build GRCh38
rs563728609 Chr.12: 132771208-132771208 on Build GRCh38
rs986989740 Chr.12: 132771313-132771313 on Build GRCh38
rs77146606 Chr.12: 132771414-132771414 on Build GRCh38
rs537173378 Chr.12: 132771413-132771413 on Build GRCh38
rs756180491 Chr.12: 132771374-132771374 on Build GRCh38
rs1043686696 Chr.12: 132771594-132771594 on Build GRCh38
rs1016958688 Chr.12: 132771232-132771232 on Build GRCh38
rs955510408 Chr.12: 132771303-132771303 on Build GRCh38
rs1009856969 Chr.12: 132771531-132771531 on Build GRCh38
rs1044453629 Chr.12: 132771376-132771376 on Build GRCh38
rs571441229 Chr.12: 132771398-132771398 on Build GRCh38
rs572412377 Chr.12: 132771621-132771621 on Build GRCh38
rs551620434 Chr.12: 132771305-132771305 on Build GRCh38
rs753306059 Chr.12: 132771562-132771569 on Build GRCh38
rs535866073 Chr.12: 132771590-132771590 on Build GRCh38
rs974359653 Chr.12: 132771367-132771367 on Build GRCh38
rs972809212 Chr.12: 132771260-132771260 on Build GRCh38
rs141225601 Chr.12: 132771595-132771595 on Build GRCh38
rs942838921 Chr.12: 132771320-132771320 on Build GRCh38
rs967706082 Chr.12: 132771250-132771250 on Build GRCh38
rs73487155 Chr.12: 132771650-132771650 on Build GRCh38
rs996542099 Chr.12: 132771614-132771614 on Build GRCh38
rs80004790 Chr.12: 132771300-132771300 on Build GRCh38
rs948764164 Chr.12: 132771375-132771375 on Build GRCh38

View Full Product Details