Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7344444-7344716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAP1
Entrez Gene ID
9744
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 1
Gene Aliases
CENTB1
UniGene
Hs.337242
Gene Chromosome Location
Chr.17: 7336506-7354944 on Build GRCh38
Transcripts
NM_014716.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5387237 c.830G>A p.S277N Chr.17: 7344624-7344624 on Build GRCh38
1709705 c.757G>A p.E253K Chr.17: 7344551-7344551 on Build GRCh38
7193826 c.798G>A p.L266L Chr.17: 7344592-7344592 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759569442 Chr.17: 7344509-7344509 on Build GRCh38
rs11868674 Chr.17: 7344675-7344675 on Build GRCh38
rs549313700 Chr.17: 7344554-7344554 on Build GRCh38
rs549465730 Chr.17: 7344653-7344653 on Build GRCh38
rs749532458 Chr.17: 7344507-7344519 on Build GRCh38
rs938039907 Chr.17: 7344557-7344557 on Build GRCh38
rs747675012 Chr.17: 7344687-7344687 on Build GRCh38
rs920491883 Chr.17: 7344466-7344466 on Build GRCh38
rs180820837 Chr.17: 7344663-7344663 on Build GRCh38
rs911584327 Chr.17: 7344497-7344497 on Build GRCh38
rs753917000 Chr.17: 7344529-7344529 on Build GRCh38
rs113243710 Chr.17: 7344469-7344469 on Build GRCh38
rs370521995 Chr.17: 7344676-7344676 on Build GRCh38
rs750760122 Chr.17: 7344617-7344617 on Build GRCh38
rs566684350 Chr.17: 7344472-7344472 on Build GRCh38
rs770672247 Chr.17: 7344671-7344675 on Build GRCh38
rs774128190 Chr.17: 7344500-7344500 on Build GRCh38
rs538244665 Chr.17: 7344664-7344664 on Build GRCh38
rs148480413 Chr.17: 7344627-7344627 on Build GRCh38
rs761961930 Chr.17: 7344538-7344538 on Build GRCh38
rs751994762 Chr.17: 7344660-7344660 on Build GRCh38
rs982497882 Chr.17: 7344523-7344523 on Build GRCh38
rs780515268 Chr.17: 7344628-7344628 on Build GRCh38
rs949343849 Chr.17: 7344485-7344485 on Build GRCh38
rs527554527 Chr.17: 7344510-7344510 on Build GRCh38
rs1045070487 Chr.17: 7344690-7344690 on Build GRCh38

View Full Product Details