Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157341812-157342404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP2
Entrez Gene ID
26999
Gene Name
cytoplasmic FMR1 interacting protein 2
Gene Aliases
PIR121
UniGene
Hs.519702
Gene Chromosome Location
Chr.5: 157266080-157395598 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1 NM_001001343.3

Gene Symbol
FNDC9
Entrez Gene ID
408263
Gene Name
fibronectin type III domain containing 9
Gene Aliases
C5orf40
UniGene
Hs.437066
Gene Chromosome Location
Chr.5: 157341599-157345721 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1 NM_001001343.3

SNP Detail

SNP ID Chromosome Location
rs950817000 Chr.5: 157342058-157342058 on Build GRCh38
rs552462393 Chr.5: 157342291-157342291 on Build GRCh38
rs1032562323 Chr.5: 157342192-157342192 on Build GRCh38
rs187870591 Chr.5: 157342349-157342349 on Build GRCh38
rs911073924 Chr.5: 157342322-157342322 on Build GRCh38
rs1034013940 Chr.5: 157342365-157342365 on Build GRCh38
rs114291757 Chr.5: 157341859-157341859 on Build GRCh38
rs11739823 Chr.5: 157342000-157342000 on Build GRCh38
rs926446577 Chr.5: 157342102-157342102 on Build GRCh38
rs531580003 Chr.5: 157342348-157342348 on Build GRCh38
rs139462846 Chr.5: 157342098-157342098 on Build GRCh38
rs890702681 Chr.5: 157341975-157341975 on Build GRCh38
rs542419281 Chr.5: 157341881-157341881 on Build GRCh38
rs1001134978 Chr.5: 157342162-157342162 on Build GRCh38
rs746432160 Chr.5: 157342342-157342342 on Build GRCh38
rs114208084 Chr.5: 157342041-157342041 on Build GRCh38
rs747784714 Chr.5: 157342138-157342138 on Build GRCh38
rs761509862 Chr.5: 157342268-157342268 on Build GRCh38
rs114763717 Chr.5: 157342226-157342226 on Build GRCh38
rs114692865 Chr.5: 157341879-157341879 on Build GRCh38
rs564708094 Chr.5: 157342018-157342018 on Build GRCh38
rs574964234 Chr.5: 157342364-157342364 on Build GRCh38
rs937848382 Chr.5: 157342125-157342125 on Build GRCh38
rs994898930 Chr.5: 157342011-157342011 on Build GRCh38
rs540703230 Chr.5: 157341935-157341935 on Build GRCh38
rs1009902553 Chr.5: 157342228-157342228 on Build GRCh38
rs969620406 Chr.5: 157341843-157341843 on Build GRCh38
rs561278407 Chr.5: 157341919-157341919 on Build GRCh38
rs529785288 Chr.5: 157342135-157342135 on Build GRCh38
rs78141548 Chr.5: 157342071-157342071 on Build GRCh38
rs574813073 Chr.5: 157341927-157341927 on Build GRCh38
rs770139419 Chr.5: 157342352-157342352 on Build GRCh38
rs973966608 Chr.5: 157342341-157342341 on Build GRCh38
rs1053479257 Chr.5: 157341916-157341916 on Build GRCh38
rs945160064 Chr.5: 157342171-157342171 on Build GRCh38
rs989148769 Chr.5: 157342144-157342144 on Build GRCh38
rs983443023 Chr.5: 157342064-157342064 on Build GRCh38
rs911168799 Chr.5: 157341959-157341959 on Build GRCh38
rs753221060 Chr.5: 157342010-157342010 on Build GRCh38
rs1027858433 Chr.5: 157342294-157342294 on Build GRCh38
rs560323524 Chr.5: 157341938-157341938 on Build GRCh38
rs942605435 Chr.5: 157341985-157341985 on Build GRCh38
rs918023760 Chr.5: 157342380-157342380 on Build GRCh38
rs767275822 Chr.5: 157341844-157341844 on Build GRCh38
rs1017749528 Chr.5: 157342007-157342007 on Build GRCh38
rs149744494 Chr.5: 157342269-157342269 on Build GRCh38
rs982455138 Chr.5: 157342101-157342101 on Build GRCh38
rs558822901 Chr.5: 157341841-157341841 on Build GRCh38
rs183255896 Chr.5: 157341842-157341842 on Build GRCh38
rs905466948 Chr.5: 157342111-157342111 on Build GRCh38
rs963818858 Chr.5: 157342333-157342333 on Build GRCh38
rs987063549 Chr.5: 157342317-157342317 on Build GRCh38
rs774180291 Chr.5: 157342204-157342205 on Build GRCh38
rs919802829 Chr.5: 157342347-157342347 on Build GRCh38
rs74356688 Chr.5: 157341958-157341958 on Build GRCh38
rs569175871 Chr.5: 157342292-157342292 on Build GRCh38
rs566392663 Chr.5: 157342264-157342264 on Build GRCh38
rs561115110 Chr.5: 157342108-157342108 on Build GRCh38
rs754690994 Chr.5: 157342045-157342045 on Build GRCh38
rs568565635 Chr.5: 157342299-157342301 on Build GRCh38

View Full Product Details