Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160995368-160995931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F11R
Entrez Gene ID
50848
Gene Name
F11 receptor
Gene Aliases
CD321, JAM, JAM1, JAMA, JCAM, KAT, PAM-1
UniGene
Hs.517293
Gene Chromosome Location
Chr.1: 160995211-161021343 on Build GRCh38
Transcripts
NM_016946.4

SNP Detail

SNP ID Chromosome Location
rs144674867 Chr.1: 160995866-160995866 on Build GRCh38
rs67433451 Chr.1: 160995748-160995749 on Build GRCh38
rs890512428 Chr.1: 160995486-160995486 on Build GRCh38
rs564596850 Chr.1: 160995897-160995897 on Build GRCh38
rs1042765424 Chr.1: 160995692-160995692 on Build GRCh38
rs894023557 Chr.1: 160995891-160995891 on Build GRCh38
rs553472765 Chr.1: 160995562-160995562 on Build GRCh38
rs1008969814 Chr.1: 160995738-160995738 on Build GRCh38
rs570396543 Chr.1: 160995479-160995479 on Build GRCh38
rs957047305 Chr.1: 160995412-160995412 on Build GRCh38
rs1022639424 Chr.1: 160995684-160995684 on Build GRCh38
rs944111150 Chr.1: 160995774-160995774 on Build GRCh38
rs908255674 Chr.1: 160995409-160995409 on Build GRCh38
rs555335466 Chr.1: 160995858-160995858 on Build GRCh38
rs999001224 Chr.1: 160995461-160995461 on Build GRCh38
rs751203543 Chr.1: 160995752-160995753 on Build GRCh38
rs948393982 Chr.1: 160995680-160995680 on Build GRCh38
rs1013168695 Chr.1: 160995446-160995446 on Build GRCh38
rs536136418 Chr.1: 160995489-160995489 on Build GRCh38
rs575978553 Chr.1: 160995733-160995733 on Build GRCh38
rs1019749610 Chr.1: 160995549-160995549 on Build GRCh38
rs3737785 Chr.1: 160995898-160995898 on Build GRCh38
rs572336846 Chr.1: 160995749-160995750 on Build GRCh38
rs377127010 Chr.1: 160995546-160995546 on Build GRCh38
rs867892920 Chr.1: 160995749-160995749 on Build GRCh38
rs928470597 Chr.1: 160995705-160995705 on Build GRCh38
rs994765673 Chr.1: 160995841-160995841 on Build GRCh38
rs542113172 Chr.1: 160995736-160995736 on Build GRCh38
rs1057005681 Chr.1: 160995576-160995576 on Build GRCh38
rs938436906 Chr.1: 160995411-160995411 on Build GRCh38
rs972668949 Chr.1: 160995727-160995727 on Build GRCh38
rs761676923 Chr.1: 160995665-160995665 on Build GRCh38
rs202176100 Chr.1: 160995748-160995755 on Build GRCh38
rs369583450 Chr.1: 160995747-160995747 on Build GRCh38
rs61803665 Chr.1: 160995545-160995545 on Build GRCh38
rs755373694 Chr.1: 160995629-160995629 on Build GRCh38
rs372825550 Chr.1: 160995768-160995773 on Build GRCh38
rs1017595965 Chr.1: 160995495-160995495 on Build GRCh38
rs1049801072 Chr.1: 160995528-160995528 on Build GRCh38
rs948531301 Chr.1: 160995475-160995475 on Build GRCh38
rs1046896464 Chr.1: 160995403-160995409 on Build GRCh38
rs75141013 Chr.1: 160995772-160995772 on Build GRCh38
rs191524137 Chr.1: 160995476-160995476 on Build GRCh38
rs939877665 Chr.1: 160995715-160995715 on Build GRCh38
rs570192127 Chr.1: 160995622-160995622 on Build GRCh38
rs764569515 Chr.1: 160995519-160995519 on Build GRCh38
rs981617969 Chr.1: 160995698-160995698 on Build GRCh38
rs139474503 Chr.1: 160995867-160995867 on Build GRCh38
rs999761430 Chr.1: 160995581-160995581 on Build GRCh38
rs766908119 Chr.1: 160995624-160995624 on Build GRCh38
rs889829935 Chr.1: 160995529-160995529 on Build GRCh38
rs541350699 Chr.1: 160995773-160995774 on Build GRCh38
rs762002439 Chr.1: 160995491-160995491 on Build GRCh38
rs183616933 Chr.1: 160995625-160995625 on Build GRCh38
rs1018129367 Chr.1: 160995769-160995774 on Build GRCh38
rs533587448 Chr.1: 160995440-160995440 on Build GRCh38
rs555932332 Chr.1: 160995626-160995626 on Build GRCh38
rs551274044 Chr.1: 160995772-160995773 on Build GRCh38
rs898489588 Chr.1: 160995507-160995507 on Build GRCh38
rs969760699 Chr.1: 160995691-160995691 on Build GRCh38
rs113364821 Chr.1: 160995732-160995732 on Build GRCh38
rs989854339 Chr.1: 160995632-160995632 on Build GRCh38
rs982543219 Chr.1: 160995595-160995595 on Build GRCh38
rs915697740 Chr.1: 160995452-160995452 on Build GRCh38
rs568051321 Chr.1: 160995532-160995532 on Build GRCh38
rs757543437 Chr.1: 160995826-160995826 on Build GRCh38
rs765423048 Chr.1: 160995472-160995472 on Build GRCh38
rs902664744 Chr.1: 160995859-160995859 on Build GRCh38
rs112148050 Chr.1: 160995516-160995516 on Build GRCh38
rs1031310033 Chr.1: 160995405-160995405 on Build GRCh38
rs904675521 Chr.1: 160995460-160995460 on Build GRCh38
rs992574699 Chr.1: 160995902-160995903 on Build GRCh38
rs908372406 Chr.1: 160995613-160995613 on Build GRCh38
rs753239649 Chr.1: 160995848-160995848 on Build GRCh38
rs1037379851 Chr.1: 160995492-160995492 on Build GRCh38

View Full Product Details