Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200560568-200560950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF14
Entrez Gene ID
9928
Gene Name
kinesin family member 14
Gene Aliases
MKS12
UniGene
Hs.3104
Gene Chromosome Location
Chr.1: 200551497-200620791 on Build GRCh38
Transcripts
NM_001305792.1 NM_014875.2 XM_011510231.2 XM_011510232.2 XM_011510233.2 XM_011510235.2 XM_011510236.1 XM_017003005.1 XM_017003006.1 XM_017003007.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6694760 c.4134A>T p.V1378V Chr.1: 200560818-200560818 on Build GRCh38
6904850 c.4186C>G p.L1396V Chr.1: 200560766-200560766 on Build GRCh38
3976812 c.4179G>C p.G1393G Chr.1: 200560773-200560773 on Build GRCh38
4546936 c.4107G>A p.E1369E Chr.1: 200560845-200560845 on Build GRCh38
375365 c.4203C>T p.N1401N Chr.1: 200560749-200560749 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991804558 Chr.1: 200560613-200560613 on Build GRCh38
rs377335626 Chr.1: 200560748-200560748 on Build GRCh38
rs548052925 Chr.1: 200560660-200560660 on Build GRCh38
rs765352677 Chr.1: 200560833-200560833 on Build GRCh38
rs754852512 Chr.1: 200560891-200560891 on Build GRCh38
rs768224343 Chr.1: 200560764-200560764 on Build GRCh38
rs747590666 Chr.1: 200560774-200560774 on Build GRCh38
rs766920695 Chr.1: 200560697-200560697 on Build GRCh38
rs756068020 Chr.1: 200560878-200560878 on Build GRCh38
rs917857867 Chr.1: 200560645-200560645 on Build GRCh38
rs777163033 Chr.1: 200560817-200560817 on Build GRCh38
rs773854019 Chr.1: 200560800-200560800 on Build GRCh38
rs559678170 Chr.1: 200560641-200560641 on Build GRCh38
rs888020961 Chr.1: 200560616-200560616 on Build GRCh38
rs754050672 Chr.1: 200560704-200560704 on Build GRCh38
rs753019306 Chr.1: 200560836-200560836 on Build GRCh38
rs924413212 Chr.1: 200560880-200560880 on Build GRCh38
rs748890169 Chr.1: 200560761-200560761 on Build GRCh38
rs373508414 Chr.1: 200560741-200560741 on Build GRCh38
rs953627835 Chr.1: 200560747-200560747 on Build GRCh38
rs758552724 Chr.1: 200560728-200560728 on Build GRCh38
rs769048341 Chr.1: 200560858-200560858 on Build GRCh38
rs764161920 Chr.1: 200560869-200560869 on Build GRCh38
rs777303869 Chr.1: 200560917-200560917 on Build GRCh38
rs375214399 Chr.1: 200560797-200560797 on Build GRCh38
rs766796790 Chr.1: 200560815-200560815 on Build GRCh38
rs761272528 Chr.1: 200560806-200560806 on Build GRCh38
rs369729175 Chr.1: 200560711-200560711 on Build GRCh38
rs534307567 Chr.1: 200560874-200560874 on Build GRCh38
rs777479851 Chr.1: 200560910-200560910 on Build GRCh38
rs747838378 Chr.1: 200560899-200560899 on Build GRCh38
rs151196320 Chr.1: 200560826-200560826 on Build GRCh38
rs779785468 Chr.1: 200560881-200560881 on Build GRCh38
rs761110961 Chr.1: 200560682-200560682 on Build GRCh38
rs771528364 Chr.1: 200560916-200560916 on Build GRCh38
rs192944130 Chr.1: 200560652-200560652 on Build GRCh38
rs976539189 Chr.1: 200560676-200560676 on Build GRCh38
rs765316633 Chr.1: 200560719-200560719 on Build GRCh38
rs753601157 Chr.1: 200560890-200560890 on Build GRCh38
rs752685557 Chr.1: 200560727-200560727 on Build GRCh38
rs769852817 Chr.1: 200560809-200560809 on Build GRCh38
rs780040784 Chr.1: 200560904-200560904 on Build GRCh38
rs200470920 Chr.1: 200560750-200560750 on Build GRCh38
rs369871290 Chr.1: 200560617-200560617 on Build GRCh38
rs530553374 Chr.1: 200560749-200560749 on Build GRCh38
rs758401427 Chr.1: 200560856-200560856 on Build GRCh38
rs962202946 Chr.1: 200560664-200560664 on Build GRCh38
rs746138826 Chr.1: 200560659-200560659 on Build GRCh38
rs554051049 Chr.1: 200560912-200560912 on Build GRCh38
rs778418335 Chr.1: 200560897-200560897 on Build GRCh38
rs79199243 Chr.1: 200560743-200560743 on Build GRCh38
rs199503476 Chr.1: 200560822-200560822 on Build GRCh38

View Full Product Details