Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124311728-124312225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM101A
Entrez Gene ID
144347
Gene Name
family with sequence similarity 101 member A
Gene Aliases
CFM2
UniGene
Hs.432901
Gene Chromosome Location
Chr.12: 124289164-124316024 on Build GRCh38
Transcripts
NM_181709.4 NM_001204299.1

Gene Symbol
ZNF664-FAM101A
Entrez Gene ID
100533183
Gene Name
filamin-interacting protein FAM101A
Gene Aliases
-
UniGene
Hs.432901
Gene Chromosome Location
Chr.12: 123973215-124316024 on Build GRCh38
Transcripts
NM_181709.4 NM_001204299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038074 c.247A>T p.R83W Chr.12: 124311857-124311857 on Build GRCh38
3688030 c.11G>A p.R4Q Chr.12: 124311864-124311864 on Build GRCh38
6658009 c.234G>A p.P78P Chr.12: 124311844-124311844 on Build GRCh38
5374563 c.288G>A p.K96K Chr.12: 124311898-124311898 on Build GRCh38
3688031 c.254G>A p.R85Q Chr.12: 124311864-124311864 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976466566 Chr.12: 124311839-124311839 on Build GRCh38
rs967512338 Chr.12: 124312162-124312162 on Build GRCh38
rs930703688 Chr.12: 124312134-124312134 on Build GRCh38
rs142004094 Chr.12: 124311822-124311822 on Build GRCh38
rs779788414 Chr.12: 124311928-124311928 on Build GRCh38
rs969674321 Chr.12: 124312081-124312081 on Build GRCh38
rs147485247 Chr.12: 124311873-124311873 on Build GRCh38
rs748745670 Chr.12: 124311930-124311930 on Build GRCh38
rs143823493 Chr.12: 124311965-124311965 on Build GRCh38
rs768298724 Chr.12: 124311933-124311933 on Build GRCh38
rs765665513 Chr.12: 124311906-124311906 on Build GRCh38
rs112604005 Chr.12: 124311850-124311850 on Build GRCh38
rs753019779 Chr.12: 124311846-124311846 on Build GRCh38
rs564165153 Chr.12: 124311972-124311972 on Build GRCh38
rs753987856 Chr.12: 124311832-124311832 on Build GRCh38
rs919353566 Chr.12: 124312129-124312129 on Build GRCh38
rs575633819 Chr.12: 124312007-124312007 on Build GRCh38
rs965161872 Chr.12: 124311830-124311830 on Build GRCh38
rs201488470 Chr.12: 124311847-124311847 on Build GRCh38
rs1011092728 Chr.12: 124312040-124312040 on Build GRCh38
rs761736318 Chr.12: 124311803-124311803 on Build GRCh38
rs188445895 Chr.12: 124311801-124311801 on Build GRCh38
rs776296219 Chr.12: 124311779-124311779 on Build GRCh38
rs1040026864 Chr.12: 124312173-124312173 on Build GRCh38
rs745308395 Chr.12: 124311780-124311780 on Build GRCh38
rs374395268 Chr.12: 124311843-124311843 on Build GRCh38
rs986771631 Chr.12: 124311818-124311818 on Build GRCh38
rs755910193 Chr.12: 124311925-124311925 on Build GRCh38
rs749185607 Chr.12: 124311965-124311966 on Build GRCh38
rs928026272 Chr.12: 124312093-124312093 on Build GRCh38
rs374813587 Chr.12: 124311952-124311952 on Build GRCh38
rs779410124 Chr.12: 124311855-124311855 on Build GRCh38
rs778627507 Chr.12: 124311940-124311940 on Build GRCh38
rs201197146 Chr.12: 124311863-124311863 on Build GRCh38
rs758959424 Chr.12: 124311915-124311915 on Build GRCh38
rs762769806 Chr.12: 124311793-124311793 on Build GRCh38
rs912503407 Chr.12: 124311936-124311936 on Build GRCh38
rs1019706758 Chr.12: 124312200-124312200 on Build GRCh38
rs775588417 Chr.12: 124311966-124311966 on Build GRCh38
rs768764193 Chr.12: 124311799-124311799 on Build GRCh38
rs912906522 Chr.12: 124311798-124311798 on Build GRCh38
rs997976568 Chr.12: 124312018-124312018 on Build GRCh38
rs769199159 Chr.12: 124311789-124311789 on Build GRCh38
rs972149226 Chr.12: 124312128-124312128 on Build GRCh38
rs547639806 Chr.12: 124311756-124311756 on Build GRCh38
rs562057447 Chr.12: 124312188-124312188 on Build GRCh38
rs775134462 Chr.12: 124311792-124311792 on Build GRCh38
rs764684959 Chr.12: 124311971-124311971 on Build GRCh38
rs994084106 Chr.12: 124311797-124311797 on Build GRCh38
rs767785192 Chr.12: 124311813-124311813 on Build GRCh38
rs948276483 Chr.12: 124311871-124311871 on Build GRCh38
rs114153127 Chr.12: 124312027-124312027 on Build GRCh38
rs900829002 Chr.12: 124312003-124312003 on Build GRCh38
rs774438403 Chr.12: 124311861-124311861 on Build GRCh38
rs777737600 Chr.12: 124311966-124311966 on Build GRCh38
rs200683384 Chr.12: 124311927-124311927 on Build GRCh38
rs766362026 Chr.12: 124311825-124311825 on Build GRCh38
rs896984609 Chr.12: 124311762-124311762 on Build GRCh38
rs1022863882 Chr.12: 124312073-124312073 on Build GRCh38
rs942669687 Chr.12: 124312168-124312168 on Build GRCh38
rs374117703 Chr.12: 124311890-124311890 on Build GRCh38
rs748091669 Chr.12: 124311941-124311941 on Build GRCh38
rs759643850 Chr.12: 124311904-124311904 on Build GRCh38
rs753807896 Chr.12: 124311898-124311898 on Build GRCh38
rs1046167591 Chr.12: 124311991-124311991 on Build GRCh38
rs773124467 Chr.12: 124311885-124311885 on Build GRCh38
rs763429949 Chr.12: 124311970-124311970 on Build GRCh38
rs766669716 Chr.12: 124311897-124311897 on Build GRCh38
rs777509955 Chr.12: 124312038-124312038 on Build GRCh38
rs1020058848 Chr.12: 124312105-124312105 on Build GRCh38
rs960676451 Chr.12: 124312104-124312104 on Build GRCh38
rs148521228 Chr.12: 124311907-124311907 on Build GRCh38
rs772856967 Chr.12: 124311951-124311951 on Build GRCh38
rs566199178 Chr.12: 124311771-124311771 on Build GRCh38
rs772083785 Chr.12: 124311943-124311943 on Build GRCh38
rs981421828 Chr.12: 124312083-124312083 on Build GRCh38
rs777970177 Chr.12: 124311916-124311916 on Build GRCh38
rs748829711 Chr.12: 124311859-124311859 on Build GRCh38
rs768678494 Chr.12: 124311860-124311860 on Build GRCh38
rs373498775 Chr.12: 124311960-124311960 on Build GRCh38
rs369101004 Chr.12: 124311953-124311953 on Build GRCh38
rs76789740 Chr.12: 124312199-124312199 on Build GRCh38
rs759552206 Chr.12: 124311958-124311958 on Build GRCh38
rs201716233 Chr.12: 124311844-124311844 on Build GRCh38
rs1030738541 Chr.12: 124312020-124312020 on Build GRCh38
rs141889365 Chr.12: 124311926-124311926 on Build GRCh38
rs368901589 Chr.12: 124311823-124311823 on Build GRCh38
rs939903921 Chr.12: 124311769-124311769 on Build GRCh38
rs909674604 Chr.12: 124312155-124312155 on Build GRCh38
rs989829405 Chr.12: 124311796-124311796 on Build GRCh38
rs367934677 Chr.12: 124311849-124311849 on Build GRCh38
rs1048228843 Chr.12: 124312146-124312146 on Build GRCh38

View Full Product Details