Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124315620-124316125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM101A
Entrez Gene ID
144347
Gene Name
family with sequence similarity 101 member A
Gene Aliases
CFM2
UniGene
Hs.432901
Gene Chromosome Location
Chr.12: 124289164-124316024 on Build GRCh38
Transcripts
NM_181709.4 NM_001204299.1

Gene Symbol
ZNF664-FAM101A
Entrez Gene ID
100533183
Gene Name
filamin-interacting protein FAM101A
Gene Aliases
-
UniGene
Hs.432901
Gene Chromosome Location
Chr.12: 123973215-124316024 on Build GRCh38
Transcripts
NM_181709.4 NM_001204299.1

SNP Detail

SNP ID Chromosome Location
rs76232753 Chr.12: 124316053-124316053 on Build GRCh38
rs553856825 Chr.12: 124315694-124315694 on Build GRCh38
rs948102333 Chr.12: 124316051-124316051 on Build GRCh38
rs989468169 Chr.12: 124316026-124316026 on Build GRCh38
rs1018420108 Chr.12: 124315896-124315896 on Build GRCh38
rs75661121 Chr.12: 124315874-124315874 on Build GRCh38
rs983128584 Chr.12: 124316041-124316041 on Build GRCh38
rs965652971 Chr.12: 124315919-124315919 on Build GRCh38
rs916885417 Chr.12: 124315742-124315742 on Build GRCh38
rs938622242 Chr.12: 124316071-124316071 on Build GRCh38
rs929010665 Chr.12: 124315685-124315685 on Build GRCh38
rs562585479 Chr.12: 124316016-124316016 on Build GRCh38
rs777012002 Chr.12: 124315726-124315726 on Build GRCh38
rs2036305 Chr.12: 124316040-124316040 on Build GRCh38
rs12551 Chr.12: 124315772-124315772 on Build GRCh38
rs1002714079 Chr.12: 124315801-124315801 on Build GRCh38
rs76454706 Chr.12: 124315866-124315866 on Build GRCh38
rs868175719 Chr.12: 124315932-124315932 on Build GRCh38
rs28562137 Chr.12: 124315845-124315845 on Build GRCh38
rs528029208 Chr.12: 124316081-124316081 on Build GRCh38
rs905731033 Chr.12: 124315757-124315757 on Build GRCh38
rs866573671 Chr.12: 124315656-124315656 on Build GRCh38
rs964934495 Chr.12: 124315915-124315915 on Build GRCh38
rs1057414394 Chr.12: 124316075-124316075 on Build GRCh38
rs949973481 Chr.12: 124315747-124315747 on Build GRCh38
rs541837427 Chr.12: 124316042-124316042 on Build GRCh38
rs534378087 Chr.12: 124315858-124315858 on Build GRCh38
rs919819744 Chr.12: 124315770-124315770 on Build GRCh38
rs774182826 Chr.12: 124315665-124315665 on Build GRCh38
rs947236336 Chr.12: 124315767-124315767 on Build GRCh38
rs1007065658 Chr.12: 124315879-124315879 on Build GRCh38
rs979907192 Chr.12: 124315780-124315780 on Build GRCh38
rs920324909 Chr.12: 124315679-124315679 on Build GRCh38
rs927124447 Chr.12: 124316063-124316063 on Build GRCh38
rs977406805 Chr.12: 124315945-124315945 on Build GRCh38
rs923988903 Chr.12: 124315994-124315994 on Build GRCh38
rs146818287 Chr.12: 124315779-124315779 on Build GRCh38
rs1009190400 Chr.12: 124315892-124315893 on Build GRCh38
rs769713058 Chr.12: 124316025-124316025 on Build GRCh38
rs914441068 Chr.12: 124315754-124315754 on Build GRCh38
rs372587611 Chr.12: 124315798-124315798 on Build GRCh38
rs1027162468 Chr.12: 124315867-124315867 on Build GRCh38
rs911495198 Chr.12: 124315648-124315648 on Build GRCh38
rs985596369 Chr.12: 124315639-124315639 on Build GRCh38
rs374906262 Chr.12: 124315681-124315681 on Build GRCh38
rs896793838 Chr.12: 124315848-124315848 on Build GRCh38
rs748084929 Chr.12: 124315682-124315682 on Build GRCh38
rs927040548 Chr.12: 124315788-124315788 on Build GRCh38
rs1017861408 Chr.12: 124315909-124315909 on Build GRCh38

View Full Product Details