Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 57011005-57011499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB1
Entrez Gene ID
1600
Gene Name
DAB1, reelin adaptor protein
Gene Aliases
-
UniGene
Hs.477370
Gene Chromosome Location
Chr.1: 56997906-58250539 on Build GRCh38
Transcripts
NM_021080.3

Gene Symbol
LOC105378749
Entrez Gene ID
105378749
Gene Name
uncharacterized LOC105378749
Gene Aliases
-
UniGene
Hs.677235
Gene Chromosome Location
Chr.1: 56998058-57014700 on Build GRCh38
Transcripts
NM_021080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3360966 c.1534G>A p.G512S Chr.1: 57011183-57011183 on Build GRCh38
4964891 c.1643G>A p.S548N Chr.1: 57011173-57011173 on Build GRCh38
5701764 c.1646_1647CC>AT p.P549H Chr.1: 57011169-57011170 on Build GRCh38
4527432 c.1562G>A p.R521K Chr.1: 57011254-57011254 on Build GRCh38
6063620 c.1512C>A p.T504T Chr.1: 57011205-57011205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984293838 Chr.1: 57011450-57011450 on Build GRCh38
rs765476976 Chr.1: 57011279-57011279 on Build GRCh38
rs749028639 Chr.1: 57011321-57011321 on Build GRCh38
rs76041739 Chr.1: 57011443-57011443 on Build GRCh38
rs868393955 Chr.1: 57011213-57011213 on Build GRCh38
rs746282815 Chr.1: 57011420-57011423 on Build GRCh38
rs781388632 Chr.1: 57011312-57011312 on Build GRCh38
rs188733717 Chr.1: 57011290-57011290 on Build GRCh38
rs373157400 Chr.1: 57011432-57011432 on Build GRCh38
rs776957089 Chr.1: 57011264-57011264 on Build GRCh38
rs759661844 Chr.1: 57011265-57011265 on Build GRCh38
rs144040023 Chr.1: 57011057-57011057 on Build GRCh38
rs758968784 Chr.1: 57011170-57011170 on Build GRCh38
rs895268828 Chr.1: 57011206-57011206 on Build GRCh38
rs369596279 Chr.1: 57011196-57011196 on Build GRCh38
rs750218948 Chr.1: 57011122-57011122 on Build GRCh38
rs977250252 Chr.1: 57011189-57011190 on Build GRCh38
rs747317225 Chr.1: 57011174-57011174 on Build GRCh38
rs1019645163 Chr.1: 57011358-57011358 on Build GRCh38
rs773006703 Chr.1: 57011228-57011228 on Build GRCh38
rs1027997560 Chr.1: 57011437-57011437 on Build GRCh38
rs576614518 Chr.1: 57011137-57011137 on Build GRCh38
rs778286361 Chr.1: 57011173-57011173 on Build GRCh38
rs769352501 Chr.1: 57011181-57011181 on Build GRCh38
rs145981487 Chr.1: 57011160-57011160 on Build GRCh38
rs999672751 Chr.1: 57011353-57011353 on Build GRCh38
rs748863712 Chr.1: 57011195-57011195 on Build GRCh38
rs760474130 Chr.1: 57011250-57011250 on Build GRCh38
rs372825410 Chr.1: 57011106-57011106 on Build GRCh38
rs753891761 Chr.1: 57011142-57011142 on Build GRCh38
rs540725213 Chr.1: 57011429-57011429 on Build GRCh38
rs200097987 Chr.1: 57011299-57011299 on Build GRCh38
rs752756620 Chr.1: 57011282-57011282 on Build GRCh38
rs929790222 Chr.1: 57011095-57011095 on Build GRCh38
rs697580 Chr.1: 57011361-57011361 on Build GRCh38
rs1008242277 Chr.1: 57011433-57011433 on Build GRCh38
rs755123075 Chr.1: 57011146-57011146 on Build GRCh38
rs867697427 Chr.1: 57011099-57011099 on Build GRCh38
rs201557324 Chr.1: 57011260-57011260 on Build GRCh38
rs369501510 Chr.1: 57011096-57011096 on Build GRCh38
rs973489799 Chr.1: 57011041-57011041 on Build GRCh38
rs772023574 Chr.1: 57011111-57011113 on Build GRCh38
rs779718940 Chr.1: 57011186-57011186 on Build GRCh38
rs201030683 Chr.1: 57011135-57011135 on Build GRCh38
rs760815827 Chr.1: 57011125-57011125 on Build GRCh38
rs919725385 Chr.1: 57011087-57011087 on Build GRCh38
rs761716676 Chr.1: 57011101-57011101 on Build GRCh38
rs771933743 Chr.1: 57011219-57011219 on Build GRCh38
rs748154116 Chr.1: 57011205-57011205 on Build GRCh38
rs773838937 Chr.1: 57011197-57011197 on Build GRCh38
rs371169348 Chr.1: 57011309-57011309 on Build GRCh38
rs17450764 Chr.1: 57011477-57011477 on Build GRCh38
rs267598671 Chr.1: 57011201-57011201 on Build GRCh38
rs766053447 Chr.1: 57011256-57011256 on Build GRCh38
rs963469542 Chr.1: 57011036-57011036 on Build GRCh38
rs777581394 Chr.1: 57011229-57011229 on Build GRCh38
rs115842928 Chr.1: 57011032-57011032 on Build GRCh38
rs1051908048 Chr.1: 57011371-57011371 on Build GRCh38
rs892255249 Chr.1: 57011342-57011342 on Build GRCh38
rs192965465 Chr.1: 57011405-57011405 on Build GRCh38
rs144227484 Chr.1: 57011233-57011233 on Build GRCh38
rs902699993 Chr.1: 57011327-57011327 on Build GRCh38
rs1037366978 Chr.1: 57011046-57011046 on Build GRCh38
rs142236626 Chr.1: 57011159-57011159 on Build GRCh38

View Full Product Details