Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 57071439-57071936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAB1
Entrez Gene ID
1600
Gene Name
DAB1, reelin adaptor protein
Gene Aliases
-
UniGene
Hs.477370
Gene Chromosome Location
Chr.1: 56997906-58250539 on Build GRCh38
Transcripts
NM_021080.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1687773 c.500G>A p.R167K Chr.1: 57071580-57071580 on Build GRCh38
6639707 c.442G>T p.E148* Chr.1: 57071638-57071638 on Build GRCh38
5675915 c.520G>T p.A174S Chr.1: 57071560-57071560 on Build GRCh38
1687774 c.449T>G p.V150G Chr.1: 57071631-57071631 on Build GRCh38
911116 c.439-1G>T p.? Chr.1: 57071642-57071642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752641705 Chr.1: 57071551-57071551 on Build GRCh38
rs147565532 Chr.1: 57071658-57071658 on Build GRCh38
rs767701683 Chr.1: 57071533-57071533 on Build GRCh38
rs776426553 Chr.1: 57071714-57071714 on Build GRCh38
rs996164693 Chr.1: 57071887-57071887 on Build GRCh38
rs1042556560 Chr.1: 57071734-57071734 on Build GRCh38
rs772160960 Chr.1: 57071660-57071660 on Build GRCh38
rs756730844 Chr.1: 57071560-57071560 on Build GRCh38
rs751296345 Chr.1: 57071618-57071618 on Build GRCh38
rs757474068 Chr.1: 57071776-57071776 on Build GRCh38
rs1008870433 Chr.1: 57071514-57071514 on Build GRCh38
rs1009169838 Chr.1: 57071483-57071483 on Build GRCh38
rs1040630962 Chr.1: 57071857-57071857 on Build GRCh38
rs780741428 Chr.1: 57071690-57071690 on Build GRCh38
rs559969171 Chr.1: 57071724-57071724 on Build GRCh38
rs779989764 Chr.1: 57071636-57071636 on Build GRCh38
rs771479617 Chr.1: 57071766-57071769 on Build GRCh38
rs924145616 Chr.1: 57071759-57071759 on Build GRCh38
rs773378664 Chr.1: 57071662-57071662 on Build GRCh38
rs770483243 Chr.1: 57071489-57071489 on Build GRCh38
rs577919871 Chr.1: 57071673-57071673 on Build GRCh38
rs780622456 Chr.1: 57071474-57071474 on Build GRCh38
rs563971293 Chr.1: 57071693-57071693 on Build GRCh38
rs1049119800 Chr.1: 57071888-57071888 on Build GRCh38
rs900506485 Chr.1: 57071586-57071586 on Build GRCh38
rs780807305 Chr.1: 57071629-57071629 on Build GRCh38
rs769700036 Chr.1: 57071497-57071497 on Build GRCh38
rs745340919 Chr.1: 57071630-57071630 on Build GRCh38
rs760119362 Chr.1: 57071679-57071679 on Build GRCh38
rs890544407 Chr.1: 57071851-57071851 on Build GRCh38
rs1021021235 Chr.1: 57071505-57071505 on Build GRCh38
rs763828068 Chr.1: 57071501-57071501 on Build GRCh38
rs754353010 Chr.1: 57071598-57071598 on Build GRCh38
rs777596258 Chr.1: 57071480-57071480 on Build GRCh38
rs758162911 Chr.1: 57071478-57071478 on Build GRCh38
rs149047297 Chr.1: 57071635-57071635 on Build GRCh38
rs982060672 Chr.1: 57071667-57071667 on Build GRCh38
rs766514617 Chr.1: 57071664-57071664 on Build GRCh38
rs915023282 Chr.1: 57071730-57071730 on Build GRCh38
rs774011865 Chr.1: 57071655-57071655 on Build GRCh38
rs775306022 Chr.1: 57071498-57071498 on Build GRCh38
rs574405795 Chr.1: 57071710-57071710 on Build GRCh38
rs900540686 Chr.1: 57071871-57071871 on Build GRCh38
rs746590813 Chr.1: 57071488-57071488 on Build GRCh38
rs996176109 Chr.1: 57071620-57071620 on Build GRCh38
rs759508268 Chr.1: 57071465-57071465 on Build GRCh38
rs144079582 Chr.1: 57071585-57071585 on Build GRCh38
rs750455500 Chr.1: 57071543-57071543 on Build GRCh38
rs1005366801 Chr.1: 57071657-57071657 on Build GRCh38
rs779357344 Chr.1: 57071616-57071616 on Build GRCh38
rs45544638 Chr.1: 57071878-57071878 on Build GRCh38
rs200624826 Chr.1: 57071644-57071644 on Build GRCh38
rs368753522 Chr.1: 57071689-57071689 on Build GRCh38
rs528736645 Chr.1: 57071725-57071725 on Build GRCh38
rs752984243 Chr.1: 57071687-57071687 on Build GRCh38
rs750030630 Chr.1: 57071691-57071691 on Build GRCh38
rs372058506 Chr.1: 57071588-57071588 on Build GRCh38
rs890425350 Chr.1: 57071511-57071511 on Build GRCh38
rs774647910 Chr.1: 57071515-57071515 on Build GRCh38
rs201179720 Chr.1: 57071653-57071653 on Build GRCh38
rs551497701 Chr.1: 57071753-57071753 on Build GRCh38
rs1042430952 Chr.1: 57071850-57071850 on Build GRCh38
rs563662046 Chr.1: 57071516-57071516 on Build GRCh38
rs946848309 Chr.1: 57071731-57071731 on Build GRCh38
rs749645273 Chr.1: 57071477-57071477 on Build GRCh38
rs866390779 Chr.1: 57071717-57071717 on Build GRCh38
rs530493486 Chr.1: 57071903-57071903 on Build GRCh38
rs760867085 Chr.1: 57071663-57071663 on Build GRCh38
rs776004302 Chr.1: 57071491-57071491 on Build GRCh38
rs756849681 Chr.1: 57071626-57071626 on Build GRCh38
rs375883054 Chr.1: 57071493-57071493 on Build GRCh38
rs374053400 Chr.1: 57071490-57071490 on Build GRCh38
rs761873136 Chr.1: 57071518-57071518 on Build GRCh38
rs543369336 Chr.1: 57071716-57071716 on Build GRCh38
rs983621596 Chr.1: 57071901-57071901 on Build GRCh38
rs1027918207 Chr.1: 57071880-57071883 on Build GRCh38
rs755412805 Chr.1: 57071609-57071609 on Build GRCh38
rs1018015073 Chr.1: 57071862-57071862 on Build GRCh38
rs765695287 Chr.1: 57071684-57071684 on Build GRCh38
rs748248461 Chr.1: 57071490-57071493 on Build GRCh38

View Full Product Details