Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 59309991-59310495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGGY
Entrez Gene ID
55277
Gene Name
FGGY carbohydrate kinase domain containing
Gene Aliases
-
UniGene
Hs.444301
Gene Chromosome Location
Chr.1: 59296405-59762730 on Build GRCh38
Transcripts
NM_001113411.1 NM_001244714.1 NM_018291.3 NR_103473.1 XM_011541730.1 XM_011541731.1 XM_011541732.1 XM_011541736.2 XM_017001643.1 XM_017001644.1 XM_017001645.1 XM_017001646.1 XM_017001647.1 XM_017001649.1 XM_017001650.1 XM_017001651.1 XM_017001652.1 XM_017001653.1 XM_017001654.1 XM_017001655.1 XM_017001656.1 XM_017001659.1 XM_017001660.1 XM_017001661.1 XM_017001663.1 XM_017001664.1 XM_017001665.1 XM_017001666.1 XM_017001667.1 XM_017001669.1 XM_017001671.1 XM_017001672.1 XM_017001673.1 XM_017001674.1 XM_017001675.1 XM_017001676.1 XM_017001679.1

SNP Detail

SNP ID Chromosome Location
rs949397619 Chr.1: 59310196-59310196 on Build GRCh38
rs563989230 Chr.1: 59310437-59310437 on Build GRCh38
rs1047305166 Chr.1: 59310154-59310154 on Build GRCh38
rs78979671 Chr.1: 59310249-59310249 on Build GRCh38
rs780218895 Chr.1: 59310451-59310451 on Build GRCh38
rs941953943 Chr.1: 59310094-59310094 on Build GRCh38
rs919116004 Chr.1: 59310126-59310126 on Build GRCh38
rs1030886311 Chr.1: 59310376-59310376 on Build GRCh38
rs1037588283 Chr.1: 59310101-59310101 on Build GRCh38
rs910558536 Chr.1: 59310401-59310401 on Build GRCh38
rs1044088641 Chr.1: 59310192-59310192 on Build GRCh38
rs753483722 Chr.1: 59310425-59310425 on Build GRCh38
rs142768372 Chr.1: 59310426-59310426 on Build GRCh38
rs41287698 Chr.1: 59310441-59310441 on Build GRCh38
rs1032592316 Chr.1: 59310293-59310293 on Build GRCh38
rs370794070 Chr.1: 59310380-59310380 on Build GRCh38
rs957115447 Chr.1: 59310318-59310318 on Build GRCh38
rs561675099 Chr.1: 59310034-59310034 on Build GRCh38
rs905172994 Chr.1: 59310246-59310246 on Build GRCh38
rs920121592 Chr.1: 59310132-59310132 on Build GRCh38
rs193267217 Chr.1: 59310381-59310381 on Build GRCh38
rs910316799 Chr.1: 59310110-59310110 on Build GRCh38
rs116253239 Chr.1: 59310445-59310445 on Build GRCh38
rs1009723867 Chr.1: 59310369-59310369 on Build GRCh38
rs963338175 Chr.1: 59310403-59310403 on Build GRCh38
rs373478642 Chr.1: 59310044-59310044 on Build GRCh38
rs988092848 Chr.1: 59310043-59310043 on Build GRCh38
rs376007009 Chr.1: 59310078-59310078 on Build GRCh38
rs751589737 Chr.1: 59310464-59310464 on Build GRCh38
rs1046357292 Chr.1: 59310237-59310237 on Build GRCh38
rs558987917 Chr.1: 59310431-59310431 on Build GRCh38
rs943090257 Chr.1: 59310111-59310111 on Build GRCh38
rs41287696 Chr.1: 59310127-59310127 on Build GRCh38
rs559675743 Chr.1: 59310087-59310087 on Build GRCh38
rs530665438 Chr.1: 59310079-59310079 on Build GRCh38
rs910527068 Chr.1: 59310093-59310093 on Build GRCh38
rs931489650 Chr.1: 59310444-59310444 on Build GRCh38
rs370229220 Chr.1: 59310133-59310133 on Build GRCh38
rs184787969 Chr.1: 59310396-59310396 on Build GRCh38
rs904155916 Chr.1: 59310193-59310193 on Build GRCh38
rs1032421792 Chr.1: 59310259-59310259 on Build GRCh38
rs564842634 Chr.1: 59310446-59310446 on Build GRCh38
rs1012649215 Chr.1: 59310177-59310177 on Build GRCh38
rs988670828 Chr.1: 59310014-59310014 on Build GRCh38
rs34644722 Chr.1: 59310469-59310470 on Build GRCh38
rs986107557 Chr.1: 59310092-59310092 on Build GRCh38
rs923362740 Chr.1: 59310020-59310020 on Build GRCh38
rs895583908 Chr.1: 59310137-59310137 on Build GRCh38
rs973764851 Chr.1: 59310413-59310413 on Build GRCh38

View Full Product Details