Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 59512212-59512757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGGY
Entrez Gene ID
55277
Gene Name
FGGY carbohydrate kinase domain containing
Gene Aliases
-
UniGene
Hs.444301
Gene Chromosome Location
Chr.1: 59296405-59762730 on Build GRCh38
Transcripts
NM_001113411.1 NM_001244714.1 NM_018291.3 NR_103473.1 XM_011541730.1 XM_011541731.1 XM_011541732.1 XM_011541733.1 XM_011541735.1 XM_011541736.2 XM_017001643.1 XM_017001644.1 XM_017001645.1 XM_017001646.1 XM_017001647.1 XM_017001648.1 XM_017001649.1 XM_017001650.1 XM_017001651.1 XM_017001652.1 XM_017001653.1 XM_017001654.1 XM_017001655.1 XM_017001656.1 XM_017001657.1 XM_017001658.1 XM_017001659.1 XM_017001660.1 XM_017001661.1 XM_017001662.1 XM_017001663.1 XM_017001664.1 XM_017001665.1 XM_017001666.1 XM_017001667.1 XM_017001668.1 XM_017001669.1 XM_017001670.1 XM_017001671.1 XM_017001672.1 XM_017001673.1 XM_017001674.1 XM_017001675.1 XM_017001676.1 XM_017001677.1 XM_017001678.1 XM_017001679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6384267 c.392C>A p.A131E Chr.1: 59512368-59512368 on Build GRCh38
6126789 c.445G>C p.A149P Chr.1: 59512421-59512421 on Build GRCh38
5969504 c.753G>T p.G251G Chr.1: 59512393-59512393 on Build GRCh38
5053611 c.727G>A p.A243T Chr.1: 59512367-59512367 on Build GRCh38
5053612 c.391G>A p.A131T Chr.1: 59512367-59512367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779430611 Chr.1: 59512431-59512432 on Build GRCh38
rs774927933 Chr.1: 59512730-59512730 on Build GRCh38
rs913856938 Chr.1: 59512624-59512624 on Build GRCh38
rs758355421 Chr.1: 59512480-59512480 on Build GRCh38
rs763277925 Chr.1: 59512359-59512359 on Build GRCh38
rs988095442 Chr.1: 59512353-59512353 on Build GRCh38
rs374855684 Chr.1: 59512352-59512352 on Build GRCh38
rs954693193 Chr.1: 59512658-59512658 on Build GRCh38
rs373050097 Chr.1: 59512400-59512400 on Build GRCh38
rs930518934 Chr.1: 59512672-59512672 on Build GRCh38
rs987494349 Chr.1: 59512683-59512683 on Build GRCh38
rs886216483 Chr.1: 59512698-59512698 on Build GRCh38
rs1037808925 Chr.1: 59512703-59512703 on Build GRCh38
rs778054131 Chr.1: 59512287-59512289 on Build GRCh38
rs778630826 Chr.1: 59512264-59512264 on Build GRCh38
rs144759346 Chr.1: 59512676-59512676 on Build GRCh38
rs900348539 Chr.1: 59512276-59512276 on Build GRCh38
rs764390692 Chr.1: 59512375-59512375 on Build GRCh38
rs765820211 Chr.1: 59512389-59512389 on Build GRCh38
rs528061854 Chr.1: 59512444-59512444 on Build GRCh38
rs763407550 Chr.1: 59512451-59512451 on Build GRCh38
rs969185575 Chr.1: 59512248-59512248 on Build GRCh38
rs561703245 Chr.1: 59512593-59512593 on Build GRCh38
rs945361085 Chr.1: 59512241-59512241 on Build GRCh38
rs781199987 Chr.1: 59512425-59512425 on Build GRCh38
rs749729204 Chr.1: 59512622-59512622 on Build GRCh38
rs372839586 Chr.1: 59512326-59512326 on Build GRCh38
rs776717697 Chr.1: 59512334-59512334 on Build GRCh38
rs945347816 Chr.1: 59512521-59512521 on Build GRCh38
rs34880868 Chr.1: 59512351-59512352 on Build GRCh38
rs929498253 Chr.1: 59512729-59512729 on Build GRCh38
rs911899894 Chr.1: 59512629-59512629 on Build GRCh38
rs1033824299 Chr.1: 59512418-59512418 on Build GRCh38
rs144401472 Chr.1: 59512305-59512305 on Build GRCh38
rs545333982 Chr.1: 59512416-59512416 on Build GRCh38
rs778888699 Chr.1: 59512398-59512398 on Build GRCh38
rs747251536 Chr.1: 59512403-59512403 on Build GRCh38
rs774639260 Chr.1: 59512457-59512457 on Build GRCh38
rs146242082 Chr.1: 59512431-59512431 on Build GRCh38
rs560840914 Chr.1: 59512435-59512435 on Build GRCh38
rs1027363949 Chr.1: 59512333-59512333 on Build GRCh38
rs546946432 Chr.1: 59512240-59512241 on Build GRCh38
rs772473025 Chr.1: 59512283-59512283 on Build GRCh38
rs1038842949 Chr.1: 59512273-59512273 on Build GRCh38
rs573781231 Chr.1: 59512350-59512350 on Build GRCh38
rs184273812 Chr.1: 59512465-59512465 on Build GRCh38
rs747196472 Chr.1: 59512318-59512318 on Build GRCh38
rs957901990 Chr.1: 59512613-59512613 on Build GRCh38
rs562936205 Chr.1: 59512399-59512399 on Build GRCh38
rs753337880 Chr.1: 59512471-59512471 on Build GRCh38
rs920720188 Chr.1: 59512727-59512727 on Build GRCh38
rs1032560119 Chr.1: 59512517-59512517 on Build GRCh38
rs369812226 Chr.1: 59512380-59512380 on Build GRCh38
rs899344929 Chr.1: 59512422-59512422 on Build GRCh38
rs1042321730 Chr.1: 59512241-59512244 on Build GRCh38
rs778165553 Chr.1: 59512308-59512308 on Build GRCh38
rs760023503 Chr.1: 59512349-59512349 on Build GRCh38
rs1026568151 Chr.1: 59512450-59512450 on Build GRCh38
rs978111901 Chr.1: 59512595-59512595 on Build GRCh38
rs377534202 Chr.1: 59512484-59512484 on Build GRCh38
rs747921550 Chr.1: 59512275-59512275 on Build GRCh38
rs925203381 Chr.1: 59512307-59512307 on Build GRCh38
rs766050010 Chr.1: 59512466-59512466 on Build GRCh38
rs759109063 Chr.1: 59512472-59512472 on Build GRCh38
rs933318072 Chr.1: 59512381-59512381 on Build GRCh38
rs764801943 Chr.1: 59512473-59512473 on Build GRCh38
rs369356815 Chr.1: 59512278-59512278 on Build GRCh38
rs988431135 Chr.1: 59512621-59512621 on Build GRCh38
rs900295423 Chr.1: 59512669-59512669 on Build GRCh38
rs753374977 Chr.1: 59512392-59512392 on Build GRCh38
rs11207463 Chr.1: 59512376-59512376 on Build GRCh38
rs1009127964 Chr.1: 59512249-59512249 on Build GRCh38
rs550700522 Chr.1: 59512733-59512733 on Build GRCh38
rs565037525 Chr.1: 59512641-59512641 on Build GRCh38
rs757738438 Chr.1: 59512331-59512331 on Build GRCh38
rs777647429 Chr.1: 59512402-59512402 on Build GRCh38
rs769011910 Chr.1: 59512453-59512453 on Build GRCh38
rs910626408 Chr.1: 59512699-59512699 on Build GRCh38
rs770249678 Chr.1: 59512438-59512438 on Build GRCh38
rs980615323 Chr.1: 59512498-59512498 on Build GRCh38
rs754167990 Chr.1: 59512306-59512306 on Build GRCh38
rs112021965 Chr.1: 59512357-59512357 on Build GRCh38
rs752688997 Chr.1: 59512475-59512475 on Build GRCh38
rs528978799 Chr.1: 59512700-59512700 on Build GRCh38

View Full Product Details