Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 59607568-59608106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGGY
Entrez Gene ID
55277
Gene Name
FGGY carbohydrate kinase domain containing
Gene Aliases
-
UniGene
Hs.444301
Gene Chromosome Location
Chr.1: 59296405-59762730 on Build GRCh38
Transcripts
NM_001113411.1 NM_001244714.1 NM_001278224.1 NM_018291.3 NR_103473.1 XM_011541730.1 XM_011541731.1 XM_011541732.1 XM_011541733.1 XM_011541735.1 XM_011541736.2 XM_017001643.1 XM_017001644.1 XM_017001645.1 XM_017001646.1 XM_017001647.1 XM_017001648.1 XM_017001649.1 XM_017001650.1 XM_017001651.1 XM_017001652.1 XM_017001653.1 XM_017001654.1 XM_017001655.1 XM_017001656.1 XM_017001657.1 XM_017001658.1 XM_017001659.1 XM_017001660.1 XM_017001661.1 XM_017001662.1 XM_017001663.1 XM_017001664.1 XM_017001665.1 XM_017001666.1 XM_017001667.1 XM_017001668.1 XM_017001669.1 XM_017001670.1 XM_017001671.1 XM_017001672.1 XM_017001673.1 XM_017001674.1 XM_017001675.1 XM_017001676.1 XM_017001677.1 XM_017001678.1 XM_017001679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
911179 c.625G>A p.V209I Chr.1: 59607860-59607860 on Build GRCh38
911177 c.597C>T p.G199G Chr.1: 59607832-59607832 on Build GRCh38
2237447 c.996C>T p.S332S Chr.1: 59607895-59607895 on Build GRCh38
2237445 c.963A>G p.V321V Chr.1: 59607862-59607862 on Build GRCh38
4943235 c.1011G>A p.L337L Chr.1: 59607910-59607910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038513985 Chr.1: 59607760-59607761 on Build GRCh38
rs907213859 Chr.1: 59607897-59607897 on Build GRCh38
rs747580458 Chr.1: 59607900-59607900 on Build GRCh38
rs956592003 Chr.1: 59608071-59608071 on Build GRCh38
rs1035508668 Chr.1: 59607829-59607829 on Build GRCh38
rs778117432 Chr.1: 59607871-59607871 on Build GRCh38
rs780845351 Chr.1: 59607933-59607940 on Build GRCh38
rs1012210207 Chr.1: 59608031-59608031 on Build GRCh38
rs536799039 Chr.1: 59607790-59607793 on Build GRCh38
rs930437774 Chr.1: 59607994-59607994 on Build GRCh38
rs547971690 Chr.1: 59608014-59608014 on Build GRCh38
rs1012068514 Chr.1: 59607929-59607929 on Build GRCh38
rs536942617 Chr.1: 59608055-59608055 on Build GRCh38
rs907077718 Chr.1: 59608064-59608064 on Build GRCh38
rs369981835 Chr.1: 59607933-59607933 on Build GRCh38
rs201277851 Chr.1: 59607885-59607885 on Build GRCh38
rs758815710 Chr.1: 59607780-59607780 on Build GRCh38
rs571961605 Chr.1: 59607788-59607788 on Build GRCh38
rs935054471 Chr.1: 59607640-59607641 on Build GRCh38
rs960761496 Chr.1: 59607831-59607831 on Build GRCh38
rs747343466 Chr.1: 59607794-59607794 on Build GRCh38
rs34026954 Chr.1: 59607878-59607878 on Build GRCh38
rs201404620 Chr.1: 59607910-59607910 on Build GRCh38
rs768288661 Chr.1: 59607816-59607816 on Build GRCh38
rs897356374 Chr.1: 59608004-59608004 on Build GRCh38
rs765342770 Chr.1: 59607935-59607935 on Build GRCh38
rs560935352 Chr.1: 59607815-59607815 on Build GRCh38
rs867268703 Chr.1: 59607721-59607721 on Build GRCh38
rs987000982 Chr.1: 59607654-59607654 on Build GRCh38
rs758981993 Chr.1: 59607869-59607869 on Build GRCh38
rs74086245 Chr.1: 59608026-59608026 on Build GRCh38
rs372539905 Chr.1: 59607833-59607833 on Build GRCh38
rs974474036 Chr.1: 59607987-59607987 on Build GRCh38
rs769615107 Chr.1: 59607809-59607809 on Build GRCh38
rs757784738 Chr.1: 59607961-59607961 on Build GRCh38
rs752712363 Chr.1: 59607864-59607864 on Build GRCh38
rs907726054 Chr.1: 59608022-59608022 on Build GRCh38
rs768323135 Chr.1: 59607884-59607884 on Build GRCh38
rs921587077 Chr.1: 59607990-59607990 on Build GRCh38
rs773164035 Chr.1: 59607903-59607903 on Build GRCh38
rs760441119 Chr.1: 59607836-59607836 on Build GRCh38
rs967639263 Chr.1: 59607595-59607595 on Build GRCh38
rs752867937 Chr.1: 59607938-59607938 on Build GRCh38
rs758165432 Chr.1: 59608066-59608066 on Build GRCh38
rs1019796133 Chr.1: 59607956-59607956 on Build GRCh38
rs968016449 Chr.1: 59608045-59608045 on Build GRCh38
rs374982665 Chr.1: 59607789-59607789 on Build GRCh38
rs944365328 Chr.1: 59607724-59607724 on Build GRCh38
rs77543297 Chr.1: 59607678-59607678 on Build GRCh38
rs1055835665 Chr.1: 59608074-59608074 on Build GRCh38
rs149702935 Chr.1: 59607891-59607891 on Build GRCh38
rs778790229 Chr.1: 59607771-59607771 on Build GRCh38
rs550246397 Chr.1: 59607972-59607972 on Build GRCh38
rs145799253 Chr.1: 59607765-59607765 on Build GRCh38
rs890021064 Chr.1: 59607706-59607706 on Build GRCh38
rs759695196 Chr.1: 59607858-59607858 on Build GRCh38
rs1001507953 Chr.1: 59607899-59607899 on Build GRCh38
rs1056569792 Chr.1: 59608024-59608024 on Build GRCh38
rs1023550593 Chr.1: 59608037-59608037 on Build GRCh38
rs967501551 Chr.1: 59607939-59607939 on Build GRCh38
rs754063323 Chr.1: 59607857-59607857 on Build GRCh38
rs200666364 Chr.1: 59607915-59607915 on Build GRCh38
rs778265157 Chr.1: 59607786-59607786 on Build GRCh38
rs766121989 Chr.1: 59607838-59607838 on Build GRCh38
rs565354392 Chr.1: 59607895-59607895 on Build GRCh38
rs772095974 Chr.1: 59607902-59607902 on Build GRCh38
rs773100282 Chr.1: 59607834-59607834 on Build GRCh38
rs191411790 Chr.1: 59607810-59607810 on Build GRCh38
rs762497381 Chr.1: 59607854-59607855 on Build GRCh38
rs148064306 Chr.1: 59607817-59607817 on Build GRCh38
rs773680369 Chr.1: 59607823-59607823 on Build GRCh38
rs761870388 Chr.1: 59607824-59607824 on Build GRCh38
rs778553524 Chr.1: 59607896-59607896 on Build GRCh38
rs930386539 Chr.1: 59607866-59607866 on Build GRCh38
rs761859122 Chr.1: 59607954-59607954 on Build GRCh38
rs781638004 Chr.1: 59607888-59607888 on Build GRCh38
rs370201173 Chr.1: 59607911-59607911 on Build GRCh38
rs765319223 Chr.1: 59607984-59607984 on Build GRCh38
rs931628557 Chr.1: 59607648-59607648 on Build GRCh38
rs1040522979 Chr.1: 59607641-59607641 on Build GRCh38
rs753260311 Chr.1: 59607773-59607773 on Build GRCh38
rs765414131 Chr.1: 59607859-59607859 on Build GRCh38
rs755080745 Chr.1: 59607757-59607757 on Build GRCh38
rs750224343 Chr.1: 59608008-59608008 on Build GRCh38

View Full Product Details