Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62236568-62237116 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK4
Entrez Gene ID
163782
Gene Name
KN motif and ankyrin repeat domains 4
Gene Aliases
ANKRD38, dJ1078M7.1
UniGene
Hs.283398
Gene Chromosome Location
Chr.1: 62236165-62319414 on Build GRCh38
Transcripts
NM_001320269.1 NM_181712.4 XM_011540843.2 XM_011540845.2 XM_017000483.1 XM_017000484.1 XM_017000485.1

SNP Detail

SNP ID Chromosome Location
rs574905626 Chr.1: 62237084-62237084 on Build GRCh38
rs374072220 Chr.1: 62236885-62236885 on Build GRCh38
rs953441032 Chr.1: 62237061-62237061 on Build GRCh38
rs536017639 Chr.1: 62236770-62236770 on Build GRCh38
rs1036487063 Chr.1: 62236741-62236741 on Build GRCh38
rs200410886 Chr.1: 62236847-62236847 on Build GRCh38
rs530020577 Chr.1: 62236598-62236598 on Build GRCh38
rs923972488 Chr.1: 62236776-62236776 on Build GRCh38
rs930453389 Chr.1: 62237026-62237026 on Build GRCh38
rs1001085274 Chr.1: 62236901-62236901 on Build GRCh38
rs887682401 Chr.1: 62236622-62236622 on Build GRCh38
rs757602889 Chr.1: 62236687-62236687 on Build GRCh38
rs1051785806 Chr.1: 62236703-62236703 on Build GRCh38
rs36039313 Chr.1: 62236612-62236613 on Build GRCh38
rs941928868 Chr.1: 62236820-62236820 on Build GRCh38
rs938325648 Chr.1: 62236863-62236863 on Build GRCh38
rs774708830 Chr.1: 62236593-62236594 on Build GRCh38
rs1023984158 Chr.1: 62236619-62236620 on Build GRCh38
rs976209593 Chr.1: 62236784-62236784 on Build GRCh38
rs6688986 Chr.1: 62237048-62237048 on Build GRCh38
rs2481688 Chr.1: 62236965-62236965 on Build GRCh38
rs577073515 Chr.1: 62236647-62236647 on Build GRCh38
rs768085338 Chr.1: 62237063-62237063 on Build GRCh38
rs979612037 Chr.1: 62236752-62236752 on Build GRCh38
rs1036656906 Chr.1: 62236824-62236824 on Build GRCh38
rs968266614 Chr.1: 62236748-62236748 on Build GRCh38
rs967096152 Chr.1: 62236843-62236843 on Build GRCh38
rs894343253 Chr.1: 62237046-62237046 on Build GRCh38
rs985954673 Chr.1: 62236804-62236804 on Build GRCh38
rs12403485 Chr.1: 62236630-62236630 on Build GRCh38
rs2457833 Chr.1: 62236780-62236780 on Build GRCh38
rs924088801 Chr.1: 62236635-62236635 on Build GRCh38
rs1035821482 Chr.1: 62236681-62236681 on Build GRCh38
rs1023874561 Chr.1: 62236740-62236740 on Build GRCh38
rs746256996 Chr.1: 62236594-62236596 on Build GRCh38
rs7529927 Chr.1: 62236661-62236661 on Build GRCh38
rs772554857 Chr.1: 62236948-62236948 on Build GRCh38
rs576030469 Chr.1: 62237016-62237016 on Build GRCh38
rs1001790330 Chr.1: 62236704-62236740 on Build GRCh38
rs922156734 Chr.1: 62236899-62236899 on Build GRCh38
rs1045691081 Chr.1: 62237052-62237052 on Build GRCh38
rs188224056 Chr.1: 62236732-62236732 on Build GRCh38
rs956753404 Chr.1: 62236998-62236998 on Build GRCh38
rs909056202 Chr.1: 62237072-62237072 on Build GRCh38
rs186421720 Chr.1: 62237082-62237082 on Build GRCh38
rs894206715 Chr.1: 62236875-62236875 on Build GRCh38
rs923331723 Chr.1: 62236785-62236785 on Build GRCh38
rs1005924636 Chr.1: 62236631-62236631 on Build GRCh38
rs140972744 Chr.1: 62236871-62236871 on Build GRCh38
rs768652118 Chr.1: 62236595-62236596 on Build GRCh38
rs1036818440 Chr.1: 62237001-62237001 on Build GRCh38
rs1023914058 Chr.1: 62236827-62236827 on Build GRCh38
rs922187832 Chr.1: 62236669-62236669 on Build GRCh38
rs11449212 Chr.1: 62236594-62236595 on Build GRCh38
rs1002636812 Chr.1: 62236664-62236664 on Build GRCh38
rs541771042 Chr.1: 62236887-62236887 on Build GRCh38
rs972025547 Chr.1: 62237083-62237083 on Build GRCh38
rs987744980 Chr.1: 62236944-62236944 on Build GRCh38
rs960925075 Chr.1: 62236702-62236702 on Build GRCh38
rs879594046 Chr.1: 62236757-62236757 on Build GRCh38
rs35485154 Chr.1: 62236946-62236947 on Build GRCh38
rs897504885 Chr.1: 62236848-62236848 on Build GRCh38
rs931931799 Chr.1: 62236688-62236688 on Build GRCh38
rs184166424 Chr.1: 62236663-62236663 on Build GRCh38
rs1034188478 Chr.1: 62236803-62236803 on Build GRCh38
rs967546854 Chr.1: 62236624-62236624 on Build GRCh38
rs1012406461 Chr.1: 62236876-62236876 on Build GRCh38
rs538472847 Chr.1: 62236813-62236813 on Build GRCh38
rs180959703 Chr.1: 62236800-62236800 on Build GRCh38
rs976477115 Chr.1: 62237029-62237029 on Build GRCh38
rs1055565492 Chr.1: 62237032-62237032 on Build GRCh38

View Full Product Details