Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 62247267-62247774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KANK4
Entrez Gene ID
163782
Gene Name
KN motif and ankyrin repeat domains 4
Gene Aliases
ANKRD38, dJ1078M7.1
UniGene
Hs.283398
Gene Chromosome Location
Chr.1: 62236165-62319414 on Build GRCh38
Transcripts
NM_001320269.1 NM_181712.4 XM_011540843.2 XM_011540845.2 XM_017000483.1 XM_017000484.1 XM_017000485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
365962 c.2773C>T p.Q925* Chr.1: 62247582-62247582 on Build GRCh38
6437119 c.2825T>C p.V942A Chr.1: 62247530-62247530 on Build GRCh38
4385987 c.2806G>T p.A936S Chr.1: 62247549-62247549 on Build GRCh38
6419056 c.2686G>A p.G896S Chr.1: 62247669-62247669 on Build GRCh38
211462 c.2749A>G p.S917G Chr.1: 62247606-62247606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764065154 Chr.1: 62247641-62247641 on Build GRCh38
rs141425340 Chr.1: 62247492-62247492 on Build GRCh38
rs375539552 Chr.1: 62247519-62247519 on Build GRCh38
rs533419690 Chr.1: 62247363-62247363 on Build GRCh38
rs945110634 Chr.1: 62247741-62247741 on Build GRCh38
rs751766065 Chr.1: 62247513-62247513 on Build GRCh38
rs757833475 Chr.1: 62247558-62247558 on Build GRCh38
rs187338839 Chr.1: 62247580-62247580 on Build GRCh38
rs151084532 Chr.1: 62247681-62247682 on Build GRCh38
rs2941679 Chr.1: 62247552-62247552 on Build GRCh38
rs913597000 Chr.1: 62247738-62247738 on Build GRCh38
rs982382699 Chr.1: 62247410-62247410 on Build GRCh38
rs770088489 Chr.1: 62247658-62247658 on Build GRCh38
rs747964044 Chr.1: 62247644-62247644 on Build GRCh38
rs370082247 Chr.1: 62247531-62247531 on Build GRCh38
rs1026673351 Chr.1: 62247391-62247391 on Build GRCh38
rs201058418 Chr.1: 62247536-62247536 on Build GRCh38
rs371872412 Chr.1: 62247491-62247491 on Build GRCh38
rs775028931 Chr.1: 62247719-62247719 on Build GRCh38
rs371644915 Chr.1: 62247305-62247305 on Build GRCh38
rs1011561621 Chr.1: 62247292-62247292 on Build GRCh38
rs762124757 Chr.1: 62247517-62247517 on Build GRCh38
rs745524585 Chr.1: 62247674-62247674 on Build GRCh38
rs1042262253 Chr.1: 62247428-62247428 on Build GRCh38
rs551198820 Chr.1: 62247310-62247310 on Build GRCh38
rs904721599 Chr.1: 62247479-62247479 on Build GRCh38
rs908223871 Chr.1: 62247483-62247483 on Build GRCh38
rs764941560 Chr.1: 62247634-62247634 on Build GRCh38
rs746578021 Chr.1: 62247562-62247562 on Build GRCh38
rs753635835 Chr.1: 62247522-62247522 on Build GRCh38
rs952429083 Chr.1: 62247737-62247737 on Build GRCh38
rs770081998 Chr.1: 62247573-62247573 on Build GRCh38
rs749649434 Chr.1: 62247682-62247682 on Build GRCh38
rs551929353 Chr.1: 62247369-62247369 on Build GRCh38
rs759870094 Chr.1: 62247467-62247467 on Build GRCh38
rs778793919 Chr.1: 62247518-62247518 on Build GRCh38
rs569364128 Chr.1: 62247360-62247360 on Build GRCh38
rs759317740 Chr.1: 62247624-62247624 on Build GRCh38
rs750631136 Chr.1: 62247623-62247623 on Build GRCh38
rs777258038 Chr.1: 62247559-62247559 on Build GRCh38
rs780654073 Chr.1: 62247449-62247459 on Build GRCh38
rs1008677928 Chr.1: 62247665-62247665 on Build GRCh38
rs751040393 Chr.1: 62247649-62247649 on Build GRCh38
rs1026786293 Chr.1: 62247729-62247729 on Build GRCh38
rs752600367 Chr.1: 62247636-62247636 on Build GRCh38
rs368404501 Chr.1: 62247422-62247422 on Build GRCh38
rs372152032 Chr.1: 62247613-62247613 on Build GRCh38
rs975681668 Chr.1: 62247387-62247387 on Build GRCh38
rs771624435 Chr.1: 62247728-62247728 on Build GRCh38
rs936213695 Chr.1: 62247481-62247481 on Build GRCh38
rs770551628 Chr.1: 62247563-62247563 on Build GRCh38
rs749730907 Chr.1: 62247687-62247687 on Build GRCh38
rs111297754 Chr.1: 62247328-62247328 on Build GRCh38
rs10889315 Chr.1: 62247574-62247574 on Build GRCh38
rs532961011 Chr.1: 62247742-62247742 on Build GRCh38
rs1040953863 Chr.1: 62247679-62247679 on Build GRCh38
rs764283055 Chr.1: 62247500-62247500 on Build GRCh38
rs780249497 Chr.1: 62247386-62247386 on Build GRCh38
rs776963867 Chr.1: 62247451-62247451 on Build GRCh38
rs780852436 Chr.1: 62247668-62247668 on Build GRCh38
rs191987743 Chr.1: 62247661-62247661 on Build GRCh38
rs201996398 Chr.1: 62247637-62247637 on Build GRCh38
rs756434968 Chr.1: 62247676-62247676 on Build GRCh38
rs552968178 Chr.1: 62247572-62247572 on Build GRCh38
rs539117780 Chr.1: 62247725-62247725 on Build GRCh38
rs562285579 Chr.1: 62247577-62247577 on Build GRCh38
rs370309792 Chr.1: 62247597-62247597 on Build GRCh38
rs139855169 Chr.1: 62247534-62247534 on Build GRCh38
rs762022520 Chr.1: 62247614-62247614 on Build GRCh38
rs770307314 Chr.1: 62247471-62247471 on Build GRCh38
rs763559327 Chr.1: 62247579-62247579 on Build GRCh38
rs1019987278 Chr.1: 62247376-62247376 on Build GRCh38
rs575098194 Chr.1: 62247688-62247688 on Build GRCh38
rs903069623 Chr.1: 62247368-62247368 on Build GRCh38
rs952480502 Chr.1: 62247405-62247405 on Build GRCh38

View Full Product Details