Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 64470636-64471137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
UniGene
Hs.443891
Gene Chromosome Location
Chr.1: 64470793-64693058 on Build GRCh38
Transcripts
NM_020925.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6617273 c.91C>T p.L31L Chr.1: 64470835-64470835 on Build GRCh38
6247623 c.45+8G>T p.? Chr.1: 64470950-64470950 on Build GRCh38
5473750 c.134C>T p.A45V Chr.1: 64470878-64470878 on Build GRCh38
4009356 c.14C>T p.A5V Chr.1: 64470911-64470911 on Build GRCh38
6246195 c.132G>T p.E44D Chr.1: 64470876-64470876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773198194 Chr.1: 64470790-64470790 on Build GRCh38
rs949457756 Chr.1: 64470696-64470696 on Build GRCh38
rs949364975 Chr.1: 64470899-64470899 on Build GRCh38
rs752081312 Chr.1: 64470852-64470852 on Build GRCh38
rs759152643 Chr.1: 64470806-64470806 on Build GRCh38
rs749541362 Chr.1: 64470878-64470878 on Build GRCh38
rs923994552 Chr.1: 64470939-64470939 on Build GRCh38
rs963129752 Chr.1: 64470832-64470832 on Build GRCh38
rs201850964 Chr.1: 64470922-64470922 on Build GRCh38
rs199600604 Chr.1: 64470855-64470855 on Build GRCh38
rs973255135 Chr.1: 64470858-64470858 on Build GRCh38
rs1004791882 Chr.1: 64470731-64470731 on Build GRCh38
rs552021142 Chr.1: 64470673-64470673 on Build GRCh38
rs369981595 Chr.1: 64470890-64470890 on Build GRCh38
rs958755092 Chr.1: 64471101-64471101 on Build GRCh38
rs952045736 Chr.1: 64470729-64470729 on Build GRCh38
rs151183132 Chr.1: 64470996-64470996 on Build GRCh38
rs913610079 Chr.1: 64470898-64470898 on Build GRCh38
rs779229260 Chr.1: 64470994-64470994 on Build GRCh38
rs889309034 Chr.1: 64470663-64470663 on Build GRCh38
rs764485820 Chr.1: 64470924-64470924 on Build GRCh38
rs764017197 Chr.1: 64471008-64471008 on Build GRCh38
rs1020690390 Chr.1: 64471100-64471100 on Build GRCh38
rs780183706 Chr.1: 64470873-64470873 on Build GRCh38
rs917937161 Chr.1: 64470680-64470680 on Build GRCh38
rs963123333 Chr.1: 64471031-64471031 on Build GRCh38
rs544161773 Chr.1: 64471005-64471005 on Build GRCh38
rs374756415 Chr.1: 64470945-64470945 on Build GRCh38
rs775264842 Chr.1: 64470921-64470921 on Build GRCh38
rs1047783811 Chr.1: 64470997-64470997 on Build GRCh38
rs759063070 Chr.1: 64470914-64470914 on Build GRCh38
rs771768634 Chr.1: 64470774-64470774 on Build GRCh38
rs745898161 Chr.1: 64470868-64470868 on Build GRCh38
rs559647880 Chr.1: 64471063-64471063 on Build GRCh38
rs943553414 Chr.1: 64470951-64470951 on Build GRCh38
rs776324043 Chr.1: 64470802-64470802 on Build GRCh38
rs570349136 Chr.1: 64470732-64470732 on Build GRCh38
rs372382055 Chr.1: 64470874-64470874 on Build GRCh38
rs758782412 Chr.1: 64470834-64470834 on Build GRCh38
rs751828812 Chr.1: 64470931-64470931 on Build GRCh38
rs968134020 Chr.1: 64471111-64471111 on Build GRCh38
rs769491884 Chr.1: 64470915-64470915 on Build GRCh38
rs554021517 Chr.1: 64470893-64470893 on Build GRCh38
rs781686383 Chr.1: 64470857-64470857 on Build GRCh38
rs766048570 Chr.1: 64470725-64470725 on Build GRCh38
rs762260131 Chr.1: 64470934-64470934 on Build GRCh38
rs966170691 Chr.1: 64470662-64470681 on Build GRCh38
rs1000950888 Chr.1: 64470728-64470728 on Build GRCh38
rs572493829 Chr.1: 64470911-64470911 on Build GRCh38
rs531961877 Chr.1: 64471021-64471021 on Build GRCh38
rs1014762226 Chr.1: 64470738-64470738 on Build GRCh38
rs750934734 Chr.1: 64470938-64470938 on Build GRCh38
rs764420985 Chr.1: 64470845-64470845 on Build GRCh38
rs139097650 Chr.1: 64471029-64471029 on Build GRCh38
rs767954627 Chr.1: 64470936-64470936 on Build GRCh38
rs368836452 Chr.1: 64470975-64470975 on Build GRCh38
rs954329947 Chr.1: 64470793-64470793 on Build GRCh38
rs760589659 Chr.1: 64470794-64470794 on Build GRCh38
rs78942112 Chr.1: 64471013-64471013 on Build GRCh38
rs1014641404 Chr.1: 64470826-64470826 on Build GRCh38
rs902624921 Chr.1: 64470705-64470705 on Build GRCh38
rs987913157 Chr.1: 64471097-64471097 on Build GRCh38
rs1042373514 Chr.1: 64470901-64470901 on Build GRCh38
rs866270383 Chr.1: 64470976-64470976 on Build GRCh38
rs754070191 Chr.1: 64470974-64470974 on Build GRCh38
rs1034175847 Chr.1: 64471052-64471052 on Build GRCh38
rs764938792 Chr.1: 64470811-64470811 on Build GRCh38
rs1004516534 Chr.1: 64471023-64471023 on Build GRCh38
rs1004567492 Chr.1: 64470825-64470825 on Build GRCh38
rs1055132827 Chr.1: 64470677-64470677 on Build GRCh38
rs937290815 Chr.1: 64470660-64470660 on Build GRCh38
rs996854308 Chr.1: 64470739-64470739 on Build GRCh38
rs1042424446 Chr.1: 64470702-64470702 on Build GRCh38
rs765893511 Chr.1: 64470799-64470799 on Build GRCh38
rs752398856 Chr.1: 64470833-64470833 on Build GRCh38
rs780167224 Chr.1: 64470955-64470955 on Build GRCh38
rs529796571 Chr.1: 64471071-64471071 on Build GRCh38
rs960527809 Chr.1: 64471060-64471060 on Build GRCh38
rs900696596 Chr.1: 64471004-64471004 on Build GRCh38
rs1054396048 Chr.1: 64470730-64470730 on Build GRCh38
rs756198980 Chr.1: 64470869-64470869 on Build GRCh38
rs995020715 Chr.1: 64471034-64471034 on Build GRCh38
rs980576786 Chr.1: 64470801-64470801 on Build GRCh38
rs1005904424 Chr.1: 64470752-64470752 on Build GRCh38
rs1016838141 Chr.1: 64470788-64470788 on Build GRCh38
rs771889446 Chr.1: 64470886-64470886 on Build GRCh38
rs1029985452 Chr.1: 64470750-64470750 on Build GRCh38

View Full Product Details