Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 130835074-130835563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABL1
Entrez Gene ID
25
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
Gene Aliases
ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl
UniGene
Hs.431048
Gene Chromosome Location
Chr.9: 130713881-130887675 on Build GRCh38
Transcripts
NM_005157.5 NM_007313.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6917533 c.9G>C p.E3D Chr.9: 130835455-130835455 on Build GRCh38
6931840 c.28G>A p.G10S Chr.9: 130835474-130835474 on Build GRCh38
6992937 c.14G>A p.C5Y Chr.9: 130835460-130835460 on Build GRCh38
6973895 c.44A>T p.K15M Chr.9: 130835490-130835490 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765093917 Chr.9: 130835347-130835347 on Build GRCh38
rs7866590 Chr.9: 130835417-130835417 on Build GRCh38
rs566960197 Chr.9: 130835400-130835400 on Build GRCh38
rs551563970 Chr.9: 130835367-130835367 on Build GRCh38
rs867306927 Chr.9: 130835296-130835296 on Build GRCh38
rs1039019340 Chr.9: 130835509-130835509 on Build GRCh38
rs386416354 Chr.9: 130835355-130835356 on Build GRCh38
rs777942802 Chr.9: 130835358-130835358 on Build GRCh38
rs547083737 Chr.9: 130835394-130835394 on Build GRCh38
rs11433463 Chr.9: 130835432-130835433 on Build GRCh38
rs977932719 Chr.9: 130835261-130835261 on Build GRCh38
rs879071055 Chr.9: 130835431-130835431 on Build GRCh38
rs531472812 Chr.9: 130835239-130835239 on Build GRCh38
rs560518347 Chr.9: 130835134-130835134 on Build GRCh38
rs746984089 Chr.9: 130835532-130835532 on Build GRCh38
rs750562445 Chr.9: 130835455-130835455 on Build GRCh38
rs752579770 Chr.9: 130835411-130835411 on Build GRCh38
rs758715020 Chr.9: 130835356-130835356 on Build GRCh38
rs762809497 Chr.9: 130835430-130835430 on Build GRCh38
rs869252581 Chr.9: 130835354-130835355 on Build GRCh38
rs763889356 Chr.9: 130835434-130835434 on Build GRCh38
rs185078786 Chr.9: 130835368-130835368 on Build GRCh38
rs869128416 Chr.9: 130835337-130835338 on Build GRCh38
rs769177282 Chr.9: 130835423-130835423 on Build GRCh38
rs540682109 Chr.9: 130835097-130835097 on Build GRCh38
rs751784616 Chr.9: 130835413-130835413 on Build GRCh38
rs563529313 Chr.9: 130835432-130835432 on Build GRCh38
rs867708178 Chr.9: 130835475-130835475 on Build GRCh38
rs752422044 Chr.9: 130835352-130835352 on Build GRCh38
rs746399848 Chr.9: 130835388-130835388 on Build GRCh38
rs986484558 Chr.9: 130835279-130835279 on Build GRCh38
rs868536950 Chr.9: 130835150-130835150 on Build GRCh38
rs369076507 Chr.9: 130835454-130835454 on Build GRCh38
rs942473685 Chr.9: 130835306-130835306 on Build GRCh38
rs765083060 Chr.9: 130835410-130835410 on Build GRCh38
rs923881960 Chr.9: 130835268-130835268 on Build GRCh38
rs756682519 Chr.9: 130835419-130835419 on Build GRCh38
rs772021289 Chr.9: 130835349-130835367 on Build GRCh38
rs933865853 Chr.9: 130835274-130835275 on Build GRCh38
rs549037029 Chr.9: 130835215-130835215 on Build GRCh38
rs991276188 Chr.9: 130835221-130835221 on Build GRCh38
rs781302379 Chr.9: 130835415-130835415 on Build GRCh38
rs776405716 Chr.9: 130835383-130835384 on Build GRCh38
rs11433465 Chr.9: 130835335-130835336 on Build GRCh38
rs775950802 Chr.9: 130835408-130835408 on Build GRCh38
rs11433464 Chr.9: 130835353-130835354 on Build GRCh38
rs562823882 Chr.9: 130835216-130835216 on Build GRCh38
rs555855497 Chr.9: 130835428-130835428 on Build GRCh38
rs959449801 Chr.9: 130835209-130835219 on Build GRCh38
rs760366614 Chr.9: 130835404-130835404 on Build GRCh38
rs529296057 Chr.9: 130835201-130835201 on Build GRCh38
rs1027023685 Chr.9: 130835189-130835189 on Build GRCh38
rs751342661 Chr.9: 130835440-130835440 on Build GRCh38
rs756167748 Chr.9: 130835485-130835485 on Build GRCh38
rs765994887 Chr.9: 130835406-130835406 on Build GRCh38
rs910852163 Chr.9: 130835300-130835301 on Build GRCh38
rs869124264 Chr.9: 130835356-130835357 on Build GRCh38
rs763836018 Chr.9: 130835412-130835412 on Build GRCh38
rs754309439 Chr.9: 130835515-130835515 on Build GRCh38
rs770694407 Chr.9: 130835427-130835427 on Build GRCh38
rs1038562274 Chr.9: 130835315-130835315 on Build GRCh38
rs762084744 Chr.9: 130835441-130835441 on Build GRCh38
rs978342260 Chr.9: 130835545-130835545 on Build GRCh38
rs968147442 Chr.9: 130835536-130835536 on Build GRCh38
rs779357411 Chr.9: 130835425-130835425 on Build GRCh38
rs1047873308 Chr.9: 130835382-130835382 on Build GRCh38
rs968324113 Chr.9: 130835230-130835230 on Build GRCh38
rs747592287 Chr.9: 130835347-130835357 on Build GRCh38
rs775985469 Chr.9: 130835393-130835393 on Build GRCh38
rs771896444 Chr.9: 130835395-130835395 on Build GRCh38
rs781180759 Chr.9: 130835378-130835378 on Build GRCh38
rs373323350 Chr.9: 130835503-130835503 on Build GRCh38
rs1022368687 Chr.9: 130835225-130835225 on Build GRCh38
rs533688531 Chr.9: 130835377-130835377 on Build GRCh38
rs749756686 Chr.9: 130835422-130835422 on Build GRCh38
rs746677365 Chr.9: 130835426-130835426 on Build GRCh38
rs1034957408 Chr.9: 130835202-130835203 on Build GRCh38
rs995009369 Chr.9: 130835158-130835158 on Build GRCh38
rs113937878 Chr.9: 130835231-130835231 on Build GRCh38
rs34738731 Chr.9: 130835336-130835337 on Build GRCh38
rs770184274 Chr.9: 130835392-130835392 on Build GRCh38
rs1012244732 Chr.9: 130835462-130835462 on Build GRCh38
rs869245913 Chr.9: 130835433-130835434 on Build GRCh38
rs866428555 Chr.9: 130835502-130835502 on Build GRCh38
rs886676205 Chr.9: 130835190-130835190 on Build GRCh38

View Full Product Details