Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 131120551-131121042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIF1L
Entrez Gene ID
83543
Gene Name
allograft inflammatory factor 1 like
Gene Aliases
C9orf58, IBA2
UniGene
Hs.4944
Gene Chromosome Location
Chr.9: 131096476-131123152 on Build GRCh38
Transcripts
NM_001185095.1 NM_001185096.1 NM_031426.3 NR_033701.1 XM_017015177.1

SNP Detail

SNP ID Chromosome Location
rs528708221 Chr.9: 131120633-131120633 on Build GRCh38
rs994893884 Chr.9: 131120740-131120740 on Build GRCh38
rs1045791501 Chr.9: 131120659-131120659 on Build GRCh38
rs1012905522 Chr.9: 131120653-131120653 on Build GRCh38
rs764508719 Chr.9: 131120772-131120772 on Build GRCh38
rs943601951 Chr.9: 131120916-131120916 on Build GRCh38
rs561451043 Chr.9: 131120954-131120954 on Build GRCh38
rs571993951 Chr.9: 131120691-131120691 on Build GRCh38
rs1017424078 Chr.9: 131120921-131120921 on Build GRCh38
rs1038874756 Chr.9: 131120593-131120593 on Build GRCh38
rs547283478 Chr.9: 131120668-131120668 on Build GRCh38
rs1045722125 Chr.9: 131120833-131120833 on Build GRCh38
rs531977405 Chr.9: 131120947-131120947 on Build GRCh38
rs1051956055 Chr.9: 131120712-131120712 on Build GRCh38
rs563335267 Chr.9: 131120724-131120724 on Build GRCh38
rs950448917 Chr.9: 131121005-131121005 on Build GRCh38
rs959458207 Chr.9: 131120581-131120581 on Build GRCh38
rs544934898 Chr.9: 131120862-131120862 on Build GRCh38
rs141270487 Chr.9: 131120996-131120996 on Build GRCh38
rs941423364 Chr.9: 131120573-131120573 on Build GRCh38
rs983033226 Chr.9: 131121006-131121006 on Build GRCh38
rs552819941 Chr.9: 131120824-131120824 on Build GRCh38
rs933652142 Chr.9: 131120866-131120866 on Build GRCh38
rs1011299469 Chr.9: 131120718-131120718 on Build GRCh38
rs1047834999 Chr.9: 131121001-131121001 on Build GRCh38
rs528671507 Chr.9: 131120955-131120955 on Build GRCh38
rs557103173 Chr.9: 131120834-131120834 on Build GRCh38
rs577490479 Chr.9: 131120840-131120840 on Build GRCh38
rs117181550 Chr.9: 131120723-131120723 on Build GRCh38
rs756352750 Chr.9: 131120902-131120902 on Build GRCh38
rs551305193 Chr.9: 131121016-131121016 on Build GRCh38
rs1003578190 Chr.9: 131120777-131120777 on Build GRCh38
rs116190463 Chr.9: 131120965-131120965 on Build GRCh38
rs353512 Chr.9: 131120797-131120797 on Build GRCh38
rs148235918 Chr.9: 131120872-131120872 on Build GRCh38
rs563521031 Chr.9: 131120867-131120867 on Build GRCh38
rs1026420296 Chr.9: 131120760-131120760 on Build GRCh38
rs757719869 Chr.9: 131120790-131120790 on Build GRCh38
rs113017234 Chr.9: 131120571-131120571 on Build GRCh38
rs571904167 Chr.9: 131120970-131120970 on Build GRCh38
rs353513 Chr.9: 131120572-131120572 on Build GRCh38
rs999180146 Chr.9: 131120677-131120677 on Build GRCh38
rs1035345392 Chr.9: 131120781-131120781 on Build GRCh38
rs145735788 Chr.9: 131120801-131120801 on Build GRCh38
rs573921080 Chr.9: 131120911-131120911 on Build GRCh38
rs990707719 Chr.9: 131120591-131120591 on Build GRCh38
rs542716413 Chr.9: 131120927-131120927 on Build GRCh38
rs922980915 Chr.9: 131120738-131120738 on Build GRCh38
rs142039195 Chr.9: 131120773-131120773 on Build GRCh38
rs536744253 Chr.9: 131120734-131120734 on Build GRCh38
rs185107272 Chr.9: 131120711-131120711 on Build GRCh38
rs915061241 Chr.9: 131120631-131120631 on Build GRCh38
rs998752151 Chr.9: 131120841-131120841 on Build GRCh38
rs1038573483 Chr.9: 131120800-131120800 on Build GRCh38
rs923583163 Chr.9: 131120703-131120703 on Build GRCh38

View Full Product Details