Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 131122134-131122630 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIF1L
Entrez Gene ID
83543
Gene Name
allograft inflammatory factor 1 like
Gene Aliases
C9orf58, IBA2
UniGene
Hs.4944
Gene Chromosome Location
Chr.9: 131096476-131123152 on Build GRCh38
Transcripts
NM_001185095.1 NM_001185096.1 NM_031426.3 NR_033701.1 XM_017015177.1

SNP Detail

SNP ID Chromosome Location
rs775180425 Chr.9: 131122160-131122160 on Build GRCh38
rs921977012 Chr.9: 131122302-131122302 on Build GRCh38
rs76982397 Chr.9: 131122250-131122250 on Build GRCh38
rs879783111 Chr.9: 131122303-131122303 on Build GRCh38
rs947824480 Chr.9: 131122378-131122378 on Build GRCh38
rs996221731 Chr.9: 131122519-131122519 on Build GRCh38
rs774735877 Chr.9: 131122349-131122349 on Build GRCh38
rs201720167 Chr.9: 131122474-131122474 on Build GRCh38
rs148553521 Chr.9: 131122456-131122456 on Build GRCh38
rs200937805 Chr.9: 131122359-131122359 on Build GRCh38
rs536190670 Chr.9: 131122213-131122213 on Build GRCh38
rs1036234759 Chr.9: 131122224-131122224 on Build GRCh38
rs906205180 Chr.9: 131122422-131122422 on Build GRCh38
rs3832604 Chr.9: 131122474-131122475 on Build GRCh38
rs921222428 Chr.9: 131122586-131122586 on Build GRCh38
rs749578978 Chr.9: 131122199-131122199 on Build GRCh38
rs912572110 Chr.9: 131122208-131122208 on Build GRCh38
rs764530929 Chr.9: 131122275-131122275 on Build GRCh38
rs540685935 Chr.9: 131122314-131122314 on Build GRCh38
rs776473945 Chr.9: 131122491-131122491 on Build GRCh38
rs954726773 Chr.9: 131122553-131122553 on Build GRCh38
rs1003508847 Chr.9: 131122435-131122435 on Build GRCh38
rs1014828106 Chr.9: 131122589-131122589 on Build GRCh38
rs1057392671 Chr.9: 131122449-131122449 on Build GRCh38
rs1043169 Chr.9: 131122156-131122156 on Build GRCh38
rs575477298 Chr.9: 131122163-131122163 on Build GRCh38
rs969515377 Chr.9: 131122453-131122453 on Build GRCh38
rs397808826 Chr.9: 131122484-131122485 on Build GRCh38
rs188059228 Chr.9: 131122462-131122462 on Build GRCh38
rs544811187 Chr.9: 131122476-131122476 on Build GRCh38
rs988142386 Chr.9: 131122196-131122196 on Build GRCh38
rs1028920431 Chr.9: 131122538-131122538 on Build GRCh38
rs940515133 Chr.9: 131122331-131122331 on Build GRCh38
rs974709972 Chr.9: 131122294-131122294 on Build GRCh38
rs353509 Chr.9: 131122574-131122574 on Build GRCh38
rs895498157 Chr.9: 131122330-131122330 on Build GRCh38
rs956246213 Chr.9: 131122243-131122243 on Build GRCh38
rs573322404 Chr.9: 131122251-131122251 on Build GRCh38
rs34366428 Chr.9: 131122451-131122452 on Build GRCh38
rs530404465 Chr.9: 131122518-131122518 on Build GRCh38
rs763494323 Chr.9: 131122223-131122223 on Build GRCh38
rs1048162117 Chr.9: 131122286-131122286 on Build GRCh38
rs987418767 Chr.9: 131122307-131122307 on Build GRCh38
rs968805941 Chr.9: 131122270-131122270 on Build GRCh38
rs956750692 Chr.9: 131122473-131122473 on Build GRCh38
rs78531203 Chr.9: 131122488-131122489 on Build GRCh38
rs1044785777 Chr.9: 131122157-131122157 on Build GRCh38
rs935327069 Chr.9: 131122162-131122162 on Build GRCh38
rs975720955 Chr.9: 131122582-131122582 on Build GRCh38
rs920334248 Chr.9: 131122352-131122352 on Build GRCh38
rs1044711630 Chr.9: 131122395-131122395 on Build GRCh38
rs79967688 Chr.9: 131122484-131122484 on Build GRCh38

View Full Product Details