Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132978626-132979122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFI1B
Entrez Gene ID
8328
Gene Name
growth factor independent 1B transcriptional repressor
Gene Aliases
BDPLT17, ZNF163B
UniGene
Hs.553160
Gene Chromosome Location
Chr.9: 132945545-132991697 on Build GRCh38
Transcripts
NM_001135031.1 NM_004188.6 XM_011519068.2 XM_011519069.2 XM_011519070.2 XM_017015175.1 XM_017015176.1

SNP Detail

SNP ID Chromosome Location
rs1043453192 Chr.9: 132978967-132978967 on Build GRCh38
rs76396041 Chr.9: 132979004-132979004 on Build GRCh38
rs1042078964 Chr.9: 132978897-132978897 on Build GRCh38
rs1023560230 Chr.9: 132979087-132979087 on Build GRCh38
rs541204377 Chr.9: 132978877-132978877 on Build GRCh38
rs114860173 Chr.9: 132978777-132978777 on Build GRCh38
rs893694749 Chr.9: 132978893-132978893 on Build GRCh38
rs55875224 Chr.9: 132978874-132978874 on Build GRCh38
rs1018549222 Chr.9: 132978693-132978693 on Build GRCh38
rs1030417677 Chr.9: 132978954-132978954 on Build GRCh38
rs937839276 Chr.9: 132978865-132978865 on Build GRCh38
rs1025899618 Chr.9: 132979061-132979061 on Build GRCh38
rs574085205 Chr.9: 132978857-132978857 on Build GRCh38
rs540003230 Chr.9: 132978707-132978707 on Build GRCh38
rs995814180 Chr.9: 132978957-132978957 on Build GRCh38
rs1009135416 Chr.9: 132978659-132978659 on Build GRCh38
rs970023040 Chr.9: 132979097-132979097 on Build GRCh38
rs900145563 Chr.9: 132978935-132978935 on Build GRCh38
rs780188919 Chr.9: 132979015-132979015 on Build GRCh38
rs1030470228 Chr.9: 132978718-132978718 on Build GRCh38
rs967429841 Chr.9: 132978674-132978674 on Build GRCh38
rs960564300 Chr.9: 132979056-132979056 on Build GRCh38
rs558062654 Chr.9: 132978724-132978724 on Build GRCh38
rs1008123356 Chr.9: 132979020-132979020 on Build GRCh38
rs1016535670 Chr.9: 132979027-132979027 on Build GRCh38
rs903586151 Chr.9: 132978968-132978968 on Build GRCh38
rs920694420 Chr.9: 132978859-132978859 on Build GRCh38
rs997695427 Chr.9: 132978717-132978717 on Build GRCh38
rs948707480 Chr.9: 132978817-132978817 on Build GRCh38
rs963936842 Chr.9: 132979053-132979053 on Build GRCh38
rs886635494 Chr.9: 132978890-132978890 on Build GRCh38
rs900217772 Chr.9: 132978697-132978697 on Build GRCh38
rs191593809 Chr.9: 132978763-132978763 on Build GRCh38
rs1046220662 Chr.9: 132978829-132978829 on Build GRCh38
rs972117460 Chr.9: 132978773-132978773 on Build GRCh38
rs1040641270 Chr.9: 132978899-132978899 on Build GRCh38
rs756384001 Chr.9: 132978896-132978896 on Build GRCh38
rs967883211 Chr.9: 132978740-132978740 on Build GRCh38
rs999306319 Chr.9: 132978972-132978972 on Build GRCh38
rs767931356 Chr.9: 132978798-132978798 on Build GRCh38
rs886704901 Chr.9: 132978652-132978652 on Build GRCh38
rs963101216 Chr.9: 132978772-132978772 on Build GRCh38
rs572172986 Chr.9: 132979096-132979096 on Build GRCh38
rs903276767 Chr.9: 132978905-132978905 on Build GRCh38
rs750717816 Chr.9: 132978860-132978860 on Build GRCh38
rs1030860618 Chr.9: 132978758-132978758 on Build GRCh38
rs980145063 Chr.9: 132978836-132978836 on Build GRCh38
rs887922901 Chr.9: 132979058-132979058 on Build GRCh38
rs997562737 Chr.9: 132978936-132978936 on Build GRCh38
rs917245189 Chr.9: 132978809-132978809 on Build GRCh38
rs926499914 Chr.9: 132978854-132978854 on Build GRCh38
rs45526934 Chr.9: 132978745-132978745 on Build GRCh38
rs537069061 Chr.9: 132978764-132978764 on Build GRCh38

View Full Product Details