Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 132989650-132990157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFI1B
Entrez Gene ID
8328
Gene Name
growth factor independent 1B transcriptional repressor
Gene Aliases
BDPLT17, ZNF163B
UniGene
Hs.553160
Gene Chromosome Location
Chr.9: 132945545-132991697 on Build GRCh38
Transcripts
NM_001135031.1 NM_004188.6 XM_006717297.3 XM_011519068.2 XM_011519069.2 XM_011519070.2 XM_017015175.1 XM_017015176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6670505 c.693C>T p.R231R Chr.9: 132989786-132989786 on Build GRCh38
4820104 c.792G>A p.K264K Chr.9: 132989885-132989885 on Build GRCh38
4450518 c.649-1G>A p.? Chr.9: 132989741-132989741 on Build GRCh38
3655176 c.734C>G p.T245R Chr.9: 132989827-132989827 on Build GRCh38
6670507 c.767G>A p.R256H Chr.9: 132989860-132989860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913577464 Chr.9: 132989963-132989963 on Build GRCh38
rs681470 Chr.9: 132989972-132989972 on Build GRCh38
rs771432303 Chr.9: 132989719-132989719 on Build GRCh38
rs773869692 Chr.9: 132989860-132989860 on Build GRCh38
rs578136625 Chr.9: 132989683-132989683 on Build GRCh38
rs367558713 Chr.9: 132989702-132989702 on Build GRCh38
rs745670434 Chr.9: 132989829-132989829 on Build GRCh38
rs765800825 Chr.9: 132989925-132989925 on Build GRCh38
rs986311076 Chr.9: 132990122-132990122 on Build GRCh38
rs755145823 Chr.9: 132989721-132989721 on Build GRCh38
rs547130331 Chr.9: 132990001-132990001 on Build GRCh38
rs776841660 Chr.9: 132989912-132989912 on Build GRCh38
rs372779350 Chr.9: 132989879-132989879 on Build GRCh38
rs952773206 Chr.9: 132989730-132989730 on Build GRCh38
rs767105897 Chr.9: 132989763-132989763 on Build GRCh38
rs1002036423 Chr.9: 132989994-132989994 on Build GRCh38
rs766810669 Chr.9: 132989899-132989899 on Build GRCh38
rs993522466 Chr.9: 132990103-132990103 on Build GRCh38
rs772402109 Chr.9: 132989735-132989735 on Build GRCh38
rs746072908 Chr.9: 132989935-132989953 on Build GRCh38
rs754439744 Chr.9: 132989788-132989788 on Build GRCh38
rs571621302 Chr.9: 132990002-132990002 on Build GRCh38
rs58133031 Chr.9: 132990058-132990058 on Build GRCh38
rs930720301 Chr.9: 132989923-132989923 on Build GRCh38
rs944694471 Chr.9: 132989848-132989848 on Build GRCh38
rs777066878 Chr.9: 132989720-132989720 on Build GRCh38
rs945098182 Chr.9: 132989971-132989971 on Build GRCh38
rs34183728 Chr.9: 132989931-132989932 on Build GRCh38
rs747507422 Chr.9: 132989684-132989684 on Build GRCh38
rs771729851 Chr.9: 132989714-132989714 on Build GRCh38
rs545245507 Chr.9: 132989688-132989688 on Build GRCh38
rs1055407895 Chr.9: 132990020-132990020 on Build GRCh38
rs374075090 Chr.9: 132989753-132989753 on Build GRCh38
rs755994673 Chr.9: 132989852-132989852 on Build GRCh38
rs910712372 Chr.9: 132990131-132990131 on Build GRCh38
rs113875915 Chr.9: 132989981-132989981 on Build GRCh38
rs778859388 Chr.9: 132989723-132989723 on Build GRCh38
rs565702703 Chr.9: 132989908-132989911 on Build GRCh38
rs528479020 Chr.9: 132989988-132989988 on Build GRCh38
rs967855552 Chr.9: 132990079-132990079 on Build GRCh38
rs371395428 Chr.9: 132989757-132989757 on Build GRCh38
rs772909138 Chr.9: 132989897-132989897 on Build GRCh38
rs765457550 Chr.9: 132989796-132989796 on Build GRCh38
rs906389252 Chr.9: 132989714-132989714 on Build GRCh38
rs375186946 Chr.9: 132989929-132989929 on Build GRCh38
rs900680337 Chr.9: 132989685-132989685 on Build GRCh38
rs764254137 Chr.9: 132989943-132989943 on Build GRCh38
rs954807060 Chr.9: 132990114-132990114 on Build GRCh38
rs768451177 Chr.9: 132989859-132989859 on Build GRCh38
rs1053415693 Chr.9: 132989893-132989893 on Build GRCh38
rs543334869 Chr.9: 132989898-132989898 on Build GRCh38
rs748020878 Chr.9: 132989713-132989714 on Build GRCh38
rs747614393 Chr.9: 132989876-132989876 on Build GRCh38
rs368151614 Chr.9: 132989828-132989828 on Build GRCh38
rs866282944 Chr.9: 132989875-132989875 on Build GRCh38
rs142322678 Chr.9: 132989754-132989754 on Build GRCh38
rs767905800 Chr.9: 132989716-132989716 on Build GRCh38
rs1016682120 Chr.9: 132989745-132989745 on Build GRCh38
rs766642707 Chr.9: 132989786-132989786 on Build GRCh38
rs759704658 Chr.9: 132989736-132989736 on Build GRCh38
rs201349915 Chr.9: 132989784-132989784 on Build GRCh38
rs146563054 Chr.9: 132990062-132990062 on Build GRCh38
rs1035248887 Chr.9: 132990024-132990024 on Build GRCh38
rs762784268 Chr.9: 132989746-132989746 on Build GRCh38
rs750759802 Chr.9: 132989718-132989718 on Build GRCh38
rs765446969 Chr.9: 132989853-132989853 on Build GRCh38
rs8193003 Chr.9: 132990109-132990109 on Build GRCh38
rs761450242 Chr.9: 132989715-132989715 on Build GRCh38
rs760110678 Chr.9: 132989921-132989921 on Build GRCh38
rs533153661 Chr.9: 132990025-132990025 on Build GRCh38
rs757388586 Chr.9: 132989952-132989952 on Build GRCh38
rs1050720309 Chr.9: 132989671-132989671 on Build GRCh38
rs910795219 Chr.9: 132989833-132989833 on Build GRCh38
rs551245163 Chr.9: 132990030-132990030 on Build GRCh38
rs1009272733 Chr.9: 132989687-132989687 on Build GRCh38
rs555222954 Chr.9: 132990073-132990073 on Build GRCh38
rs758726712 Chr.9: 132989808-132989808 on Build GRCh38
rs539540352 Chr.9: 132989681-132989681 on Build GRCh38
rs371084332 Chr.9: 132989769-132989769 on Build GRCh38
rs896155878 Chr.9: 132990035-132990035 on Build GRCh38
rs773925352 Chr.9: 132989672-132989672 on Build GRCh38
rs986660744 Chr.9: 132989807-132989807 on Build GRCh38
rs367947845 Chr.9: 132989761-132989761 on Build GRCh38
rs751873363 Chr.9: 132989948-132989948 on Build GRCh38
rs758496657 Chr.9: 132989941-132989941 on Build GRCh38
rs962059503 Chr.9: 132990077-132990077 on Build GRCh38
rs749288375 Chr.9: 132989854-132989854 on Build GRCh38
rs752776987 Chr.9: 132989802-132989802 on Build GRCh38
rs769624853 Chr.9: 132989737-132989737 on Build GRCh38
rs201340517 Chr.9: 132989795-132989795 on Build GRCh38
rs763665503 Chr.9: 132989693-132989693 on Build GRCh38
rs375635333 Chr.9: 132989813-132989813 on Build GRCh38
rs748424899 Chr.9: 132989725-132989725 on Build GRCh38
rs755412589 Chr.9: 132989789-132989789 on Build GRCh38
rs761044764 Chr.9: 132989785-132989785 on Build GRCh38

View Full Product Details