Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133066931-133067376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEL
Entrez Gene ID
1056
Gene Name
carboxyl ester lipase
Gene Aliases
BAL, BSDL, BSSL, CELL, CEase, FAP, FAPP, LIPA, MODY8
UniGene
Hs.533258
Gene Chromosome Location
Chr.9: 133061978-133071863 on Build GRCh38
Transcripts
NM_001807.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1460765 c.851C>T p.T284I Chr.9: 133067152-133067152 on Build GRCh38
487180 c.787-1G>A p.? Chr.9: 133067087-133067087 on Build GRCh38
1248030 c.848T>G p.V283G Chr.9: 133067149-133067149 on Build GRCh38
397373 c.793G>A p.E265K Chr.9: 133067094-133067094 on Build GRCh38
6114926 c.850A>T p.T284S Chr.9: 133067151-133067151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182456386 Chr.9: 133067057-133067057 on Build GRCh38
rs748928500 Chr.9: 133067085-133067085 on Build GRCh38
rs748552732 Chr.9: 133067171-133067171 on Build GRCh38
rs373691843 Chr.9: 133067121-133067121 on Build GRCh38
rs754041656 Chr.9: 133067129-133067129 on Build GRCh38
rs776968541 Chr.9: 133067254-133067254 on Build GRCh38
rs762150574 Chr.9: 133067110-133067110 on Build GRCh38
rs781325625 Chr.9: 133067144-133067144 on Build GRCh38
rs777624006 Chr.9: 133066959-133066959 on Build GRCh38
rs540843348 Chr.9: 133067124-133067124 on Build GRCh38
rs778047659 Chr.9: 133067246-133067246 on Build GRCh38
rs369868891 Chr.9: 133067160-133067160 on Build GRCh38
rs772458342 Chr.9: 133067240-133067240 on Build GRCh38
rs768904758 Chr.9: 133067096-133067096 on Build GRCh38
rs766552475 Chr.9: 133067040-133067040 on Build GRCh38
rs759706318 Chr.9: 133067256-133067256 on Build GRCh38
rs547648173 Chr.9: 133067239-133067239 on Build GRCh38
rs775173186 Chr.9: 133066975-133066975 on Build GRCh38
rs749571469 Chr.9: 133067212-133067212 on Build GRCh38
rs779842315 Chr.9: 133067220-133067220 on Build GRCh38
rs1027973097 Chr.9: 133067206-133067207 on Build GRCh38
rs868527871 Chr.9: 133067184-133067184 on Build GRCh38
rs746951446 Chr.9: 133067053-133067053 on Build GRCh38
rs745748354 Chr.9: 133067157-133067157 on Build GRCh38
rs756182405 Chr.9: 133066996-133066996 on Build GRCh38
rs775024540 Chr.9: 133067084-133067084 on Build GRCh38
rs765533068 Chr.9: 133067193-133067193 on Build GRCh38
rs953962057 Chr.9: 133067029-133067029 on Build GRCh38
rs774639442 Chr.9: 133067099-133067099 on Build GRCh38
rs747488282 Chr.9: 133067249-133067249 on Build GRCh38
rs746257330 Chr.9: 133067175-133067175 on Build GRCh38
rs187489195 Chr.9: 133067344-133067344 on Build GRCh38
rs371637241 Chr.9: 133067050-133067050 on Build GRCh38
rs575395415 Chr.9: 133067342-133067342 on Build GRCh38
rs527649141 Chr.9: 133067306-133067306 on Build GRCh38
rs201066180 Chr.9: 133067188-133067188 on Build GRCh38
rs543159660 Chr.9: 133066981-133066981 on Build GRCh38
rs775183712 Chr.9: 133067063-133067077 on Build GRCh38
rs781267031 Chr.9: 133067066-133067066 on Build GRCh38
rs774285970 Chr.9: 133067174-133067174 on Build GRCh38
rs553188665 Chr.9: 133067354-133067354 on Build GRCh38
rs769635334 Chr.9: 133066973-133066973 on Build GRCh38
rs377579138 Chr.9: 133067203-133067203 on Build GRCh38
rs369248889 Chr.9: 133067075-133067075 on Build GRCh38
rs748906749 Chr.9: 133067161-133067161 on Build GRCh38
rs370266923 Chr.9: 133067229-133067229 on Build GRCh38
rs771529870 Chr.9: 133067250-133067250 on Build GRCh38
rs539790792 Chr.9: 133067261-133067261 on Build GRCh38
rs530887007 Chr.9: 133067179-133067179 on Build GRCh38
rs761673436 Chr.9: 133067219-133067220 on Build GRCh38
rs565872546 Chr.9: 133067259-133067259 on Build GRCh38
rs773990119 Chr.9: 133067170-133067170 on Build GRCh38
rs1024143823 Chr.9: 133067298-133067298 on Build GRCh38
rs116059127 Chr.9: 133067345-133067345 on Build GRCh38
rs755165969 Chr.9: 133067135-133067135 on Build GRCh38
rs770922315 Chr.9: 133067181-133067181 on Build GRCh38
rs926641823 Chr.9: 133067327-133067327 on Build GRCh38
rs564666083 Chr.9: 133067278-133067278 on Build GRCh38
rs746898142 Chr.9: 133066964-133066964 on Build GRCh38
rs561785461 Chr.9: 133066994-133066994 on Build GRCh38
rs372555759 Chr.9: 133067243-133067243 on Build GRCh38
rs757083662 Chr.9: 133067142-133067142 on Build GRCh38
rs572472996 Chr.9: 133067254-133067254 on Build GRCh38
rs778585448 Chr.9: 133067237-133067237 on Build GRCh38
rs754070066 Chr.9: 133067046-133067046 on Build GRCh38
rs139832194 Chr.9: 133066995-133066995 on Build GRCh38
rs766436684 Chr.9: 133067126-133067126 on Build GRCh38
rs762518268 Chr.9: 133067085-133067086 on Build GRCh38
rs763272949 Chr.9: 133066980-133066980 on Build GRCh38
rs777587552 Chr.9: 133067051-133067051 on Build GRCh38
rs575987314 Chr.9: 133066972-133066972 on Build GRCh38
rs768411918 Chr.9: 133067168-133067168 on Build GRCh38
rs567714650 Chr.9: 133067219-133067219 on Build GRCh38
rs367558907 Chr.9: 133066991-133066991 on Build GRCh38
rs375451730 Chr.9: 133067123-133067123 on Build GRCh38
rs770158285 Chr.9: 133067177-133067177 on Build GRCh38
rs200036978 Chr.9: 133066971-133066971 on Build GRCh38
rs751462980 Chr.9: 133067141-133067141 on Build GRCh38
rs771869344 Chr.9: 133066956-133066956 on Build GRCh38
rs770598389 Chr.9: 133066970-133066970 on Build GRCh38
rs754415944 Chr.9: 133067230-133067230 on Build GRCh38
rs201657030 Chr.9: 133067068-133067068 on Build GRCh38
rs763582789 Chr.9: 133067099-133067102 on Build GRCh38
rs867846320 Chr.9: 133067262-133067262 on Build GRCh38
rs537018843 Chr.9: 133067307-133067307 on Build GRCh38
rs767378268 Chr.9: 133067206-133067206 on Build GRCh38
rs759524824 Chr.9: 133067189-133067189 on Build GRCh38

View Full Product Details