Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154766910-154767408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKC1
Entrez Gene ID
1736
Gene Name
dyskerin pseudouridine synthase 1
Gene Aliases
CBF5, DKC, DKCX, NAP57, NOLA4, XAP101
UniGene
Hs.4747
Gene Chromosome Location
Chr.X: 154762742-154777689 on Build GRCh38
Transcripts
NM_001142463.2 NM_001288747.1 NM_001363.4 NR_110021.1 NR_110022.1 NR_110023.1

SNP Detail

SNP ID Chromosome Location
rs781897987 Chr.X: 154767282-154767282 on Build GRCh38
rs782639991 Chr.X: 154767333-154767333 on Build GRCh38
rs781931193 Chr.X: 154767113-154767113 on Build GRCh38
rs782429056 Chr.X: 154767073-154767073 on Build GRCh38
rs7064200 Chr.X: 154766942-154766942 on Build GRCh38
rs782273508 Chr.X: 154767080-154767080 on Build GRCh38
rs782225456 Chr.X: 154767102-154767102 on Build GRCh38
rs782422741 Chr.X: 154767174-154767174 on Build GRCh38
rs201758541 Chr.X: 154767090-154767090 on Build GRCh38
rs942619378 Chr.X: 154767141-154767141 on Build GRCh38
rs781789313 Chr.X: 154767342-154767342 on Build GRCh38
rs374548935 Chr.X: 154766933-154766933 on Build GRCh38
rs782215854 Chr.X: 154767046-154767046 on Build GRCh38
rs372412858 Chr.X: 154767105-154767105 on Build GRCh38
rs782504051 Chr.X: 154767016-154767016 on Build GRCh38
rs782735351 Chr.X: 154767187-154767187 on Build GRCh38
rs782261412 Chr.X: 154767245-154767245 on Build GRCh38
rs972134314 Chr.X: 154766944-154766944 on Build GRCh38
rs183681840 Chr.X: 154767148-154767148 on Build GRCh38
rs782159247 Chr.X: 154767035-154767035 on Build GRCh38
rs782546233 Chr.X: 154767324-154767324 on Build GRCh38
rs782336767 Chr.X: 154766968-154766969 on Build GRCh38
rs782803768 Chr.X: 154766980-154766980 on Build GRCh38
rs371421366 Chr.X: 154767363-154767363 on Build GRCh38
rs781876771 Chr.X: 154767012-154767012 on Build GRCh38
rs374771308 Chr.X: 154767364-154767364 on Build GRCh38
rs782471921 Chr.X: 154767264-154767264 on Build GRCh38
rs782571821 Chr.X: 154766961-154766961 on Build GRCh38
rs782134126 Chr.X: 154767232-154767232 on Build GRCh38
rs782765692 Chr.X: 154767242-154767242 on Build GRCh38
rs782686999 Chr.X: 154767078-154767078 on Build GRCh38
rs782622660 Chr.X: 154767042-154767042 on Build GRCh38
rs782697778 Chr.X: 154767279-154767279 on Build GRCh38
rs199422246 Chr.X: 154767020-154767020 on Build GRCh38
rs370024967 Chr.X: 154767223-154767223 on Build GRCh38
rs782193589 Chr.X: 154767368-154767368 on Build GRCh38
rs188253757 Chr.X: 154767206-154767206 on Build GRCh38

View Full Product Details