Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 62050806-62051309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARID5B
Entrez Gene ID
84159
Gene Name
AT-rich interaction domain 5B
Gene Aliases
DESRT, MRF-2, MRF2
UniGene
Hs.535297
Gene Chromosome Location
Chr.10: 61901254-62096948 on Build GRCh38
Transcripts
NM_001244638.1 NM_032199.2 XM_011540262.1 XM_017016765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4769217 c.5-7T>G p.? Chr.10: 62050881-62050881 on Build GRCh38
6980691 c.109G>A p.V37I Chr.10: 62050992-62050992 on Build GRCh38
919366 c.735G>A p.A245A Chr.10: 62050889-62050889 on Build GRCh38
3782548 c.28C>T p.R10C Chr.10: 62050911-62050911 on Build GRCh38
6951725 c.835_836insC p.V280fs*7 Chr.10: 62050989-62050990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs898795654 Chr.10: 62051018-62051018 on Build GRCh38
rs1032558328 Chr.10: 62051145-62051145 on Build GRCh38
rs765605825 Chr.10: 62051066-62051066 on Build GRCh38
rs562968164 Chr.10: 62051285-62051285 on Build GRCh38
rs781124580 Chr.10: 62051083-62051083 on Build GRCh38
rs770609104 Chr.10: 62051028-62051028 on Build GRCh38
rs755494121 Chr.10: 62050884-62050884 on Build GRCh38
rs754324583 Chr.10: 62051041-62051041 on Build GRCh38
rs756332840 Chr.10: 62050910-62050910 on Build GRCh38
rs118127014 Chr.10: 62051070-62051070 on Build GRCh38
rs375447123 Chr.10: 62050860-62050860 on Build GRCh38
rs370155560 Chr.10: 62050991-62050991 on Build GRCh38
rs200241004 Chr.10: 62051031-62051031 on Build GRCh38
rs576995601 Chr.10: 62051005-62051005 on Build GRCh38
rs777512969 Chr.10: 62051151-62051151 on Build GRCh38
rs367642225 Chr.10: 62050845-62050845 on Build GRCh38
rs367834035 Chr.10: 62051006-62051006 on Build GRCh38
rs143185576 Chr.10: 62050992-62050992 on Build GRCh38
rs752562816 Chr.10: 62051088-62051088 on Build GRCh38
rs754128323 Chr.10: 62050994-62050994 on Build GRCh38
rs777877292 Chr.10: 62050922-62050922 on Build GRCh38
rs372060091 Chr.10: 62051019-62051019 on Build GRCh38
rs990501431 Chr.10: 62051240-62051240 on Build GRCh38
rs775869189 Chr.10: 62051033-62051033 on Build GRCh38
rs748675078 Chr.10: 62051161-62051161 on Build GRCh38
rs967293182 Chr.10: 62051135-62051135 on Build GRCh38
rs987975810 Chr.10: 62051279-62051279 on Build GRCh38
rs918933657 Chr.10: 62050903-62050903 on Build GRCh38
rs776458295 Chr.10: 62050950-62050950 on Build GRCh38
rs981800327 Chr.10: 62051184-62051184 on Build GRCh38
rs780084801 Chr.10: 62050927-62050927 on Build GRCh38
rs749913903 Chr.10: 62050852-62050852 on Build GRCh38
rs541679953 Chr.10: 62051181-62051181 on Build GRCh38
rs766803815 Chr.10: 62050864-62050864 on Build GRCh38
rs747942208 Chr.10: 62050961-62050961 on Build GRCh38
rs752073681 Chr.10: 62050874-62050874 on Build GRCh38
rs761044789 Chr.10: 62051037-62051037 on Build GRCh38
rs758440176 Chr.10: 62051072-62051072 on Build GRCh38
rs992373562 Chr.10: 62050842-62050842 on Build GRCh38
rs368575697 Chr.10: 62050925-62050925 on Build GRCh38
rs746932853 Chr.10: 62050929-62050929 on Build GRCh38
rs150752704 Chr.10: 62050973-62050973 on Build GRCh38
rs769487551 Chr.10: 62051010-62051010 on Build GRCh38
rs927742433 Chr.10: 62051199-62051199 on Build GRCh38
rs748934833 Chr.10: 62051020-62051020 on Build GRCh38
rs753041204 Chr.10: 62050893-62050893 on Build GRCh38
rs796285738 Chr.10: 62051038-62051038 on Build GRCh38
rs375354880 Chr.10: 62051032-62051032 on Build GRCh38
rs757615225 Chr.10: 62050997-62050997 on Build GRCh38
rs927711471 Chr.10: 62050934-62050934 on Build GRCh38
rs770272482 Chr.10: 62051276-62051276 on Build GRCh38
rs768544889 Chr.10: 62050942-62050942 on Build GRCh38
rs769439880 Chr.10: 62050966-62050966 on Build GRCh38
rs762391579 Chr.10: 62050975-62050975 on Build GRCh38
rs907668040 Chr.10: 62051280-62051280 on Build GRCh38
rs916098904 Chr.10: 62051247-62051247 on Build GRCh38
rs138196909 Chr.10: 62050953-62050953 on Build GRCh38
rs374858461 Chr.10: 62051004-62051004 on Build GRCh38
rs760053508 Chr.10: 62050984-62050984 on Build GRCh38
rs950470323 Chr.10: 62051178-62051178 on Build GRCh38
rs201704836 Chr.10: 62050980-62050980 on Build GRCh38
rs963182272 Chr.10: 62051106-62051106 on Build GRCh38
rs771958639 Chr.10: 62051085-62051087 on Build GRCh38
rs937750656 Chr.10: 62051215-62051215 on Build GRCh38
rs146939872 Chr.10: 62050926-62050926 on Build GRCh38
rs776910978 Chr.10: 62051130-62051130 on Build GRCh38
rs757523191 Chr.10: 62051052-62051052 on Build GRCh38
rs756110928 Chr.10: 62051140-62051140 on Build GRCh38
rs959803267 Chr.10: 62050831-62050831 on Build GRCh38
rs762166155 Chr.10: 62051231-62051231 on Build GRCh38
rs764348201 Chr.10: 62050993-62050993 on Build GRCh38
rs748749476 Chr.10: 62050889-62050889 on Build GRCh38
rs764544496 Chr.10: 62051039-62051039 on Build GRCh38
rs761732949 Chr.10: 62051060-62051061 on Build GRCh38
rs921995840 Chr.10: 62051266-62051266 on Build GRCh38
rs772753680 Chr.10: 62051093-62051093 on Build GRCh38
rs774134980 Chr.10: 62050967-62050969 on Build GRCh38
rs765859847 Chr.10: 62050976-62050976 on Build GRCh38

View Full Product Details