Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 66280361-66280873 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA3
Entrez Gene ID
29119
Gene Name
catenin alpha 3
Gene Aliases
ARVD13, VR22
UniGene
Hs.21375
Gene Chromosome Location
Chr.10: 65912518-67696217 on Build GRCh38
Transcripts
NM_001127384.2 NM_013266.3 XM_017016151.1 XM_017016152.1 XM_017016153.1 XM_017016154.1 XM_017016155.1 XM_017016156.1 XM_017016157.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319733 c.1775C>T p.A592V Chr.10: 66280579-66280579 on Build GRCh38
5644518 c.1860C>G p.I620M Chr.10: 66280494-66280494 on Build GRCh38
6448695 c.1829C>G p.S610* Chr.10: 66280525-66280525 on Build GRCh38
3967110 c.1797T>G p.N599K Chr.10: 66280557-66280557 on Build GRCh38
5461506 c.1787G>A p.S596N Chr.10: 66280567-66280567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878908991 Chr.10: 66280706-66280706 on Build GRCh38
rs768797369 Chr.10: 66280579-66280579 on Build GRCh38
rs774317415 Chr.10: 66280589-66280589 on Build GRCh38
rs760552633 Chr.10: 66280617-66280617 on Build GRCh38
rs10997034 Chr.10: 66280482-66280482 on Build GRCh38
rs183252670 Chr.10: 66280772-66280772 on Build GRCh38
rs753530218 Chr.10: 66280844-66280844 on Build GRCh38
rs397952714 Chr.10: 66280838-66280839 on Build GRCh38
rs757726745 Chr.10: 66280484-66280484 on Build GRCh38
rs368000502 Chr.10: 66280615-66280615 on Build GRCh38
rs780346090 Chr.10: 66280564-66280564 on Build GRCh38
rs138314889 Chr.10: 66280531-66280531 on Build GRCh38
rs778084531 Chr.10: 66280657-66280658 on Build GRCh38
rs537042209 Chr.10: 66280828-66280829 on Build GRCh38
rs775370911 Chr.10: 66280614-66280614 on Build GRCh38
rs149470831 Chr.10: 66280730-66280730 on Build GRCh38
rs575072220 Chr.10: 66280832-66280832 on Build GRCh38
rs755658279 Chr.10: 66280474-66280474 on Build GRCh38
rs367881835 Chr.10: 66280806-66280806 on Build GRCh38
rs767260854 Chr.10: 66280523-66280523 on Build GRCh38
rs780217926 Chr.10: 66280497-66280497 on Build GRCh38
rs763834998 Chr.10: 66280621-66280621 on Build GRCh38
rs755389413 Chr.10: 66280671-66280671 on Build GRCh38
rs1056307638 Chr.10: 66280732-66280732 on Build GRCh38
rs77439535 Chr.10: 66280807-66280807 on Build GRCh38
rs377044064 Chr.10: 66280649-66280649 on Build GRCh38
rs946350265 Chr.10: 66280598-66280598 on Build GRCh38
rs4548513 Chr.10: 66280567-66280567 on Build GRCh38
rs990944705 Chr.10: 66280624-66280624 on Build GRCh38
rs375734372 Chr.10: 66280619-66280619 on Build GRCh38
rs761355084 Chr.10: 66280644-66280644 on Build GRCh38
rs754683126 Chr.10: 66280423-66280425 on Build GRCh38
rs374695438 Chr.10: 66280449-66280449 on Build GRCh38
rs190239785 Chr.10: 66280504-66280504 on Build GRCh38
rs778322726 Chr.10: 66280782-66280782 on Build GRCh38
rs924163378 Chr.10: 66280439-66280439 on Build GRCh38
rs764612743 Chr.10: 66280647-66280647 on Build GRCh38
rs184992188 Chr.10: 66280501-66280501 on Build GRCh38
rs923648581 Chr.10: 66280705-66280705 on Build GRCh38
rs759355486 Chr.10: 66280594-66280594 on Build GRCh38
rs554916620 Chr.10: 66280815-66280815 on Build GRCh38
rs772371271 Chr.10: 66280496-66280496 on Build GRCh38
rs193155648 Chr.10: 66280622-66280622 on Build GRCh38
rs978476622 Chr.10: 66280739-66280739 on Build GRCh38
rs1044973386 Chr.10: 66280759-66280759 on Build GRCh38
rs770893892 Chr.10: 66280513-66280513 on Build GRCh38
rs1006017682 Chr.10: 66280578-66280578 on Build GRCh38
rs936875153 Chr.10: 66280765-66280765 on Build GRCh38
rs1047351160 Chr.10: 66280826-66280826 on Build GRCh38
rs149081490 Chr.10: 66280472-66280472 on Build GRCh38
rs752574565 Chr.10: 66280526-66280526 on Build GRCh38
rs774209973 Chr.10: 66280518-66280518 on Build GRCh38
rs780540370 Chr.10: 66280654-66280654 on Build GRCh38
rs137910578 Chr.10: 66280554-66280554 on Build GRCh38
rs753066401 Chr.10: 66280606-66280606 on Build GRCh38
rs1054036558 Chr.10: 66280770-66280770 on Build GRCh38
rs548045837 Chr.10: 66280582-66280582 on Build GRCh38
rs532225159 Chr.10: 66280550-66280550 on Build GRCh38
rs896426280 Chr.10: 66280736-66280736 on Build GRCh38
rs138101734 Chr.10: 66280469-66280469 on Build GRCh38
rs61169201 Chr.10: 66280829-66280830 on Build GRCh38
rs367616357 Chr.10: 66280489-66280489 on Build GRCh38
rs775642323 Chr.10: 66280423-66280423 on Build GRCh38
rs762826132 Chr.10: 66280463-66280463 on Build GRCh38
rs746240423 Chr.10: 66280495-66280495 on Build GRCh38
rs187527391 Chr.10: 66280562-66280562 on Build GRCh38
rs754792055 Chr.10: 66280471-66280471 on Build GRCh38
rs758975970 Chr.10: 66280558-66280558 on Build GRCh38
rs551887944 Chr.10: 66280551-66280551 on Build GRCh38
rs747190761 Chr.10: 66280498-66280498 on Build GRCh38
rs531612019 Chr.10: 66280757-66280757 on Build GRCh38
rs760720976 Chr.10: 66280435-66280435 on Build GRCh38
rs766237431 Chr.10: 66280465-66280465 on Build GRCh38
rs753334586 Chr.10: 66280542-66280542 on Build GRCh38
rs763853559 Chr.10: 66280540-66280540 on Build GRCh38
rs182490263 Chr.10: 66280506-66280506 on Build GRCh38
rs139661235 Chr.10: 66280541-66280541 on Build GRCh38
rs751915344 Chr.10: 66280664-66280664 on Build GRCh38
rs769809500 Chr.10: 66280583-66280583 on Build GRCh38
rs866894854 Chr.10: 66280492-66280492 on Build GRCh38
rs559118359 Chr.10: 66280800-66280800 on Build GRCh38
rs997681027 Chr.10: 66280824-66280824 on Build GRCh38

View Full Product Details