Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32609802-32610201 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf59
Entrez Gene ID
56683
Gene Name
chromosome 21 open reading frame 59
Gene Aliases
C21orf48, CILD26, FBB18, Kur
UniGene
Hs.5811
Gene Chromosome Location
Chr.21: 32601674-32612608 on Build GRCh38
Transcripts
NM_021254.2 NR_036552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4472079 c.175C>T p.P59S Chr.21: 32609970-32609970 on Build GRCh38
6615645 c.290G>A p.G97E Chr.21: 32609855-32609855 on Build GRCh38
6615648 c.156C>T p.A52A Chr.21: 32609989-32609989 on Build GRCh38
4101245 c.192A>G p.G64G Chr.21: 32609953-32609953 on Build GRCh38
4101244 c.258C>T p.S86S Chr.21: 32609887-32609887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs562934216 Chr.21: 32610060-32610060 on Build GRCh38
rs745535175 Chr.21: 32610035-32610035 on Build GRCh38
rs773676621 Chr.21: 32610028-32610028 on Build GRCh38
rs754390813 Chr.21: 32609886-32609886 on Build GRCh38
rs763500013 Chr.21: 32610056-32610056 on Build GRCh38
rs759060499 Chr.21: 32609966-32609966 on Build GRCh38
rs755093345 Chr.21: 32610003-32610003 on Build GRCh38
rs777191447 Chr.21: 32610047-32610047 on Build GRCh38
rs999085167 Chr.21: 32609990-32609990 on Build GRCh38
rs112319532 Chr.21: 32610110-32610110 on Build GRCh38
rs757953127 Chr.21: 32610099-32610102 on Build GRCh38
rs139014235 Chr.21: 32609947-32609947 on Build GRCh38
rs747926523 Chr.21: 32610013-32610013 on Build GRCh38
rs568749302 Chr.21: 32609918-32609918 on Build GRCh38
rs368599489 Chr.21: 32609955-32609955 on Build GRCh38
rs143740376 Chr.21: 32609853-32609853 on Build GRCh38
rs375421240 Chr.21: 32610175-32610175 on Build GRCh38
rs770495702 Chr.21: 32610026-32610026 on Build GRCh38
rs185474701 Chr.21: 32610106-32610106 on Build GRCh38
rs147461593 Chr.21: 32609989-32609989 on Build GRCh38
rs1027185980 Chr.21: 32610112-32610112 on Build GRCh38
rs762660612 Chr.21: 32609852-32609852 on Build GRCh38
rs199853806 Chr.21: 32609976-32609976 on Build GRCh38
rs772643740 Chr.21: 32609848-32609848 on Build GRCh38
rs952879996 Chr.21: 32609864-32609864 on Build GRCh38
rs1009726909 Chr.21: 32609922-32609922 on Build GRCh38
rs761611313 Chr.21: 32609840-32609840 on Build GRCh38
rs775768955 Chr.21: 32609858-32609858 on Build GRCh38
rs751824011 Chr.21: 32609998-32609998 on Build GRCh38
rs76974938 Chr.21: 32609946-32609946 on Build GRCh38
rs780294574 Chr.21: 32609900-32609900 on Build GRCh38
rs750577375 Chr.21: 32609988-32609988 on Build GRCh38
rs1029086620 Chr.21: 32610157-32610157 on Build GRCh38
rs770246587 Chr.21: 32610050-32610050 on Build GRCh38
rs749181383 Chr.21: 32610023-32610023 on Build GRCh38
rs995843813 Chr.21: 32610069-32610069 on Build GRCh38
rs926318349 Chr.21: 32610174-32610174 on Build GRCh38
rs765823366 Chr.21: 32609986-32609986 on Build GRCh38
rs769719774 Chr.21: 32609945-32609945 on Build GRCh38
rs962616975 Chr.21: 32610133-32610133 on Build GRCh38
rs555910127 Chr.21: 32610046-32610046 on Build GRCh38
rs758749024 Chr.21: 32609897-32609897 on Build GRCh38
rs754827987 Chr.21: 32609907-32609907 on Build GRCh38
rs779235313 Chr.21: 32609948-32609948 on Build GRCh38
rs769310705 Chr.21: 32609845-32609845 on Build GRCh38
rs143039524 Chr.21: 32609984-32609984 on Build GRCh38
rs768518723 Chr.21: 32609838-32609838 on Build GRCh38
rs574138242 Chr.21: 32610055-32610055 on Build GRCh38
rs781108075 Chr.21: 32610006-32610006 on Build GRCh38
rs146402309 Chr.21: 32610084-32610084 on Build GRCh38
rs765408859 Chr.21: 32609891-32609891 on Build GRCh38
rs140986492 Chr.21: 32609928-32609928 on Build GRCh38
rs755185617 Chr.21: 32609896-32609896 on Build GRCh38
rs878855068 Chr.21: 32609895-32609895 on Build GRCh38
rs765126863 Chr.21: 32610002-32610003 on Build GRCh38
rs980851328 Chr.21: 32610153-32610153 on Build GRCh38
rs111591095 Chr.21: 32609875-32609875 on Build GRCh38
rs764324895 Chr.21: 32609876-32609876 on Build GRCh38
rs577933979 Chr.21: 32610075-32610075 on Build GRCh38
rs372418001 Chr.21: 32610018-32610018 on Build GRCh38
rs142980722 Chr.21: 32610150-32610150 on Build GRCh38
rs747161389 Chr.21: 32609906-32609906 on Build GRCh38
rs111673701 Chr.21: 32610140-32610140 on Build GRCh38
rs375347058 Chr.21: 32609887-32609887 on Build GRCh38
rs544716497 Chr.21: 32610057-32610057 on Build GRCh38
rs546097943 Chr.21: 32610076-32610076 on Build GRCh38
rs774082809 Chr.21: 32609959-32609959 on Build GRCh38
rs766323336 Chr.21: 32609992-32609992 on Build GRCh38
rs776664044 Chr.21: 32609839-32609839 on Build GRCh38
rs952973074 Chr.21: 32610122-32610122 on Build GRCh38
rs550389788 Chr.21: 32610162-32610162 on Build GRCh38
rs777658239 Chr.21: 32610022-32610022 on Build GRCh38
rs764532656 Chr.21: 32609968-32609968 on Build GRCh38
rs113830647 Chr.21: 32610129-32610129 on Build GRCh38

View Full Product Details