Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 67474391-67474966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPC
Entrez Gene ID
1060
Gene Name
centromere protein C
Gene Aliases
CENP-C, CENPC1, MIF2, hcp-4
UniGene
Hs.479867
Gene Chromosome Location
Chr.4: 67468762-67545601 on Build GRCh38
Transcripts
NM_001812.2 XM_006714064.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6626495 c.2687T>C p.F896S Chr.4: 67474962-67474962 on Build GRCh38
6056446 c.2712T>C p.H904H Chr.4: 67474937-67474937 on Build GRCh38
225544 c.2755C>T p.P919S Chr.4: 67474894-67474894 on Build GRCh38
5702981 c.2694C>A p.D898E Chr.4: 67474955-67474955 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs931778626 Chr.4: 67474628-67474628 on Build GRCh38
rs763519178 Chr.4: 67474555-67474555 on Build GRCh38
rs754228457 Chr.4: 67474780-67474780 on Build GRCh38
rs767315061 Chr.4: 67474635-67474635 on Build GRCh38
rs908246471 Chr.4: 67474704-67474704 on Build GRCh38
rs890354063 Chr.4: 67474453-67474453 on Build GRCh38
rs113515697 Chr.4: 67474591-67474591 on Build GRCh38
rs983798519 Chr.4: 67474689-67474690 on Build GRCh38
rs199562985 Chr.4: 67474881-67474881 on Build GRCh38
rs775313132 Chr.4: 67474506-67474506 on Build GRCh38
rs370870458 Chr.4: 67474930-67474930 on Build GRCh38
rs570828888 Chr.4: 67474507-67474507 on Build GRCh38
rs568997242 Chr.4: 67474443-67474443 on Build GRCh38
rs920985139 Chr.4: 67474662-67474662 on Build GRCh38
rs939955824 Chr.4: 67474548-67474548 on Build GRCh38
rs756963527 Chr.4: 67474615-67474615 on Build GRCh38
rs150009234 Chr.4: 67474590-67474590 on Build GRCh38
rs372713883 Chr.4: 67474703-67474704 on Build GRCh38
rs1035876417 Chr.4: 67474527-67474527 on Build GRCh38
rs988260181 Chr.4: 67474420-67474420 on Build GRCh38
rs940563309 Chr.4: 67474640-67474640 on Build GRCh38
rs899120788 Chr.4: 67474660-67474660 on Build GRCh38
rs780001268 Chr.4: 67474842-67474842 on Build GRCh38
rs555915232 Chr.4: 67474713-67474713 on Build GRCh38
rs755880295 Chr.4: 67474835-67474835 on Build GRCh38
rs200572892 Chr.4: 67474703-67474703 on Build GRCh38
rs202214457 Chr.4: 67474885-67474885 on Build GRCh38
rs1052003799 Chr.4: 67474630-67474630 on Build GRCh38
rs1009534449 Chr.4: 67474897-67474897 on Build GRCh38
rs186836939 Chr.4: 67474621-67474621 on Build GRCh38
rs376889812 Chr.4: 67474899-67474899 on Build GRCh38
rs373276677 Chr.4: 67474661-67474661 on Build GRCh38
rs564727768 Chr.4: 67474426-67474426 on Build GRCh38
rs952579391 Chr.4: 67474673-67474673 on Build GRCh38
rs370054315 Chr.4: 67474858-67474858 on Build GRCh38
rs201433526 Chr.4: 67474904-67474904 on Build GRCh38
rs1002096394 Chr.4: 67474513-67474513 on Build GRCh38
rs917185352 Chr.4: 67474724-67474724 on Build GRCh38
rs1045127996 Chr.4: 67474608-67474608 on Build GRCh38
rs75737176 Chr.4: 67474704-67474704 on Build GRCh38
rs968345302 Chr.4: 67474613-67474613 on Build GRCh38
rs995488596 Chr.4: 67474669-67474669 on Build GRCh38
rs755416720 Chr.4: 67474878-67474878 on Build GRCh38
rs948513141 Chr.4: 67474724-67474724 on Build GRCh38
rs375139725 Chr.4: 67474653-67474653 on Build GRCh38
rs1000838830 Chr.4: 67474620-67474620 on Build GRCh38
rs747229790 Chr.4: 67474864-67474864 on Build GRCh38
rs374627347 Chr.4: 67474500-67474500 on Build GRCh38
rs62303192 Chr.4: 67474528-67474528 on Build GRCh38
rs376426502 Chr.4: 67474910-67474910 on Build GRCh38
rs1048455132 Chr.4: 67474701-67474701 on Build GRCh38
rs773336322 Chr.4: 67474863-67474863 on Build GRCh38
rs750127517 Chr.4: 67474937-67474937 on Build GRCh38
rs749186749 Chr.4: 67474855-67474855 on Build GRCh38
rs200509560 Chr.4: 67474905-67474905 on Build GRCh38
rs960244514 Chr.4: 67474837-67474837 on Build GRCh38
rs182235678 Chr.4: 67474494-67474494 on Build GRCh38
rs939900782 Chr.4: 67474712-67474712 on Build GRCh38
rs897168522 Chr.4: 67474498-67474498 on Build GRCh38
rs751152248 Chr.4: 67474914-67474914 on Build GRCh38
rs1001647229 Chr.4: 67474751-67474751 on Build GRCh38
rs943769902 Chr.4: 67474444-67474444 on Build GRCh38
rs771183601 Chr.4: 67474870-67474870 on Build GRCh38
rs117429927 Chr.4: 67474462-67474462 on Build GRCh38
rs564533734 Chr.4: 67474879-67474879 on Build GRCh38

View Full Product Details