Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2432492-2433048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHST12
Entrez Gene ID
55501
Gene Name
carbohydrate (chondroitin 4) sulfotransferase 12
Gene Aliases
C4S-2, C4ST-2, C4ST2
UniGene
Hs.744987
Gene Chromosome Location
Chr.7: 2403489-2434607 on Build GRCh38
Transcripts
NM_001243794.1 NM_001243795.1 NM_018641.4 XM_011515443.2 XM_011515444.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6322876 c.48G>T p.V16V Chr.7: 2432687-2432687 on Build GRCh38
6248742 c.204G>T p.E68D Chr.7: 2432843-2432843 on Build GRCh38
3880314 c.223A>G p.S75G Chr.7: 2432862-2432862 on Build GRCh38
239330 c.334C>T p.R112W Chr.7: 2432973-2432973 on Build GRCh38
6894872 c.222C>G p.L74L Chr.7: 2432861-2432861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774169204 Chr.7: 2432974-2432974 on Build GRCh38
rs200264761 Chr.7: 2432948-2432948 on Build GRCh38
rs955669873 Chr.7: 2432832-2432832 on Build GRCh38
rs906965994 Chr.7: 2432742-2432742 on Build GRCh38
rs541011645 Chr.7: 2432744-2432744 on Build GRCh38
rs757418358 Chr.7: 2432708-2432708 on Build GRCh38
rs11537796 Chr.7: 2433027-2433027 on Build GRCh38
rs776736414 Chr.7: 2432915-2432915 on Build GRCh38
rs929073775 Chr.7: 2432939-2432939 on Build GRCh38
rs769271363 Chr.7: 2432811-2432811 on Build GRCh38
rs767142740 Chr.7: 2432605-2432605 on Build GRCh38
rs143942839 Chr.7: 2432774-2432774 on Build GRCh38
rs764487296 Chr.7: 2432771-2432771 on Build GRCh38
rs760696935 Chr.7: 2432822-2432822 on Build GRCh38
rs778735139 Chr.7: 2432783-2432783 on Build GRCh38
rs968004071 Chr.7: 2432527-2432527 on Build GRCh38
rs202038444 Chr.7: 2432918-2432918 on Build GRCh38
rs774975904 Chr.7: 2432903-2432903 on Build GRCh38
rs777212687 Chr.7: 2432682-2432682 on Build GRCh38
rs373646935 Chr.7: 2432602-2432602 on Build GRCh38
rs546286632 Chr.7: 2432685-2432685 on Build GRCh38
rs781074899 Chr.7: 2432722-2432722 on Build GRCh38
rs201463941 Chr.7: 2432608-2432608 on Build GRCh38
rs935890142 Chr.7: 2432518-2432518 on Build GRCh38
rs765465791 Chr.7: 2432614-2432614 on Build GRCh38
rs755678792 Chr.7: 2432676-2432676 on Build GRCh38
rs773077575 Chr.7: 2432611-2432611 on Build GRCh38
rs201387415 Chr.7: 2432980-2432980 on Build GRCh38
rs754936215 Chr.7: 2432724-2432724 on Build GRCh38
rs781398371 Chr.7: 2433017-2433017 on Build GRCh38
rs1031672043 Chr.7: 2432881-2432881 on Build GRCh38
rs766150878 Chr.7: 2432990-2432990 on Build GRCh38
rs758402020 Chr.7: 2432837-2432837 on Build GRCh38
rs929043390 Chr.7: 2432751-2432751 on Build GRCh38
rs577438892 Chr.7: 2432684-2432684 on Build GRCh38
rs1054296869 Chr.7: 2432730-2432730 on Build GRCh38
rs1023309966 Chr.7: 2433025-2433025 on Build GRCh38
rs557546220 Chr.7: 2432632-2432632 on Build GRCh38
rs951624838 Chr.7: 2432609-2432609 on Build GRCh38
rs141139719 Chr.7: 2432645-2432645 on Build GRCh38
rs772772822 Chr.7: 2432607-2432607 on Build GRCh38
rs750271419 Chr.7: 2432982-2432982 on Build GRCh38
rs772825820 Chr.7: 2432665-2432665 on Build GRCh38
rs751363984 Chr.7: 2432855-2432855 on Build GRCh38
rs780741537 Chr.7: 2432871-2432871 on Build GRCh38
rs772332771 Chr.7: 2432966-2432966 on Build GRCh38
rs758988286 Chr.7: 2433008-2433008 on Build GRCh38
rs763442722 Chr.7: 2432769-2432769 on Build GRCh38
rs774057994 Chr.7: 2432590-2432590 on Build GRCh38
rs149493991 Chr.7: 2432733-2432733 on Build GRCh38
rs762205012 Chr.7: 2432816-2432816 on Build GRCh38
rs537319044 Chr.7: 2432927-2432927 on Build GRCh38
rs773306630 Chr.7: 2432910-2432910 on Build GRCh38
rs760890293 Chr.7: 2432912-2432912 on Build GRCh38
rs573194622 Chr.7: 2432975-2432975 on Build GRCh38
rs1012659005 Chr.7: 2432563-2432563 on Build GRCh38
rs978378349 Chr.7: 2432584-2432584 on Build GRCh38
rs3735100 Chr.7: 2432820-2432820 on Build GRCh38
rs750508134 Chr.7: 2432779-2432779 on Build GRCh38
rs745394094 Chr.7: 2432795-2432795 on Build GRCh38
rs137904669 Chr.7: 2432732-2432732 on Build GRCh38
rs771248775 Chr.7: 2433011-2433011 on Build GRCh38
rs779394171 Chr.7: 2432628-2432629 on Build GRCh38
rs138895778 Chr.7: 2432638-2432638 on Build GRCh38
rs778112422 Chr.7: 2432629-2432629 on Build GRCh38
rs778123979 Chr.7: 2433009-2433009 on Build GRCh38
rs761396958 Chr.7: 2432592-2432592 on Build GRCh38
rs747611968 Chr.7: 2432896-2432898 on Build GRCh38
rs555242317 Chr.7: 2432581-2432581 on Build GRCh38
rs200895324 Chr.7: 2432892-2432892 on Build GRCh38
rs759058041 Chr.7: 2432757-2432757 on Build GRCh38
rs1047472098 Chr.7: 2432940-2432940 on Build GRCh38
rs374540711 Chr.7: 2432870-2432870 on Build GRCh38
rs1044236800 Chr.7: 2432981-2432981 on Build GRCh38
rs752131799 Chr.7: 2433004-2433004 on Build GRCh38
rs3735099 Chr.7: 2432794-2432794 on Build GRCh38
rs769991079 Chr.7: 2433021-2433021 on Build GRCh38
rs774922808 Chr.7: 2432812-2432812 on Build GRCh38
rs1022573498 Chr.7: 2432522-2432522 on Build GRCh38
rs199890527 Chr.7: 2432735-2432735 on Build GRCh38
rs561223252 Chr.7: 2432768-2432768 on Build GRCh38
rs764435891 Chr.7: 2432702-2432702 on Build GRCh38
rs754537174 Chr.7: 2432994-2432994 on Build GRCh38
rs773800932 Chr.7: 2432801-2432806 on Build GRCh38
rs17132395 Chr.7: 2432964-2432964 on Build GRCh38
rs777430276 Chr.7: 2432734-2432734 on Build GRCh38
rs571043155 Chr.7: 2432962-2432962 on Build GRCh38
rs750502987 Chr.7: 2432723-2432723 on Build GRCh38
rs758190950 Chr.7: 2432791-2432791 on Build GRCh38
rs767773477 Chr.7: 2432777-2432777 on Build GRCh38
rs752586583 Chr.7: 2432784-2432784 on Build GRCh38
rs771193781 Chr.7: 2432969-2432969 on Build GRCh38
rs753905050 Chr.7: 2432836-2432836 on Build GRCh38
rs749755603 Chr.7: 2432806-2432806 on Build GRCh38
rs553499652 Chr.7: 2432973-2432973 on Build GRCh38
rs746034097 Chr.7: 2433018-2433018 on Build GRCh38
rs750173383 Chr.7: 2432942-2432942 on Build GRCh38
rs746807012 Chr.7: 2432792-2432792 on Build GRCh38
rs747931126 Chr.7: 2432656-2432656 on Build GRCh38
rs753498159 Chr.7: 2432993-2432993 on Build GRCh38
rs763389286 Chr.7: 2432686-2432686 on Build GRCh38
rs377437782 Chr.7: 2432619-2432619 on Build GRCh38
rs766587075 Chr.7: 2432914-2432914 on Build GRCh38
rs747399350 Chr.7: 2433010-2433010 on Build GRCh38
rs772439530 Chr.7: 2432907-2432907 on Build GRCh38
rs200893373 Chr.7: 2432620-2432620 on Build GRCh38
rs142111836 Chr.7: 2432801-2432801 on Build GRCh38
rs757363492 Chr.7: 2432636-2432636 on Build GRCh38
rs759592836 Chr.7: 2432916-2432916 on Build GRCh38
rs974191414 Chr.7: 2432906-2432906 on Build GRCh38
rs769324562 Chr.7: 2432902-2432902 on Build GRCh38
rs757030475 Chr.7: 2432863-2432863 on Build GRCh38
rs894294125 Chr.7: 2432560-2432560 on Build GRCh38
rs562814984 Chr.7: 2432532-2432532 on Build GRCh38
rs17132393 Chr.7: 2432883-2432883 on Build GRCh38
rs778876663 Chr.7: 2432729-2432729 on Build GRCh38
rs757009303 Chr.7: 2432924-2432924 on Build GRCh38
rs199799991 Chr.7: 2432995-2432995 on Build GRCh38
rs747308551 Chr.7: 2432967-2432967 on Build GRCh38
rs778630466 Chr.7: 2432652-2432652 on Build GRCh38
rs775707171 Chr.7: 2433024-2433024 on Build GRCh38
rs760197641 Chr.7: 2432674-2432674 on Build GRCh38
rs149805994 Chr.7: 2432972-2432972 on Build GRCh38
rs772573270 Chr.7: 2432800-2432805 on Build GRCh38
rs756193680 Chr.7: 2432782-2432782 on Build GRCh38
rs1040334821 Chr.7: 2432780-2432780 on Build GRCh38
rs796250783 Chr.7: 2432548-2432548 on Build GRCh38
rs753446844 Chr.7: 2432951-2432951 on Build GRCh38
rs754487868 Chr.7: 2432961-2432961 on Build GRCh38
rs375672884 Chr.7: 2432661-2432661 on Build GRCh38
rs12536223 Chr.7: 2432920-2432920 on Build GRCh38
rs779767707 Chr.7: 2432891-2432891 on Build GRCh38
rs563115579 Chr.7: 2432800-2432800 on Build GRCh38
rs114841399 Chr.7: 2432817-2432817 on Build GRCh38
rs769309077 Chr.7: 2432796-2432796 on Build GRCh38
rs748471742 Chr.7: 2432635-2432636 on Build GRCh38
rs763991303 Chr.7: 2432840-2432840 on Build GRCh38
rs928477047 Chr.7: 2432931-2432931 on Build GRCh38
rs150285273 Chr.7: 2432705-2432705 on Build GRCh38
rs61729409 Chr.7: 2432952-2432952 on Build GRCh38
rs770914148 Chr.7: 2432931-2432943 on Build GRCh38
rs376187790 Chr.7: 2432755-2432755 on Build GRCh38
rs748481497 Chr.7: 2432904-2432904 on Build GRCh38
rs17793768 Chr.7: 2432882-2432882 on Build GRCh38
rs527427286 Chr.7: 2432971-2432971 on Build GRCh38
rs559930657 Chr.7: 2432985-2432985 on Build GRCh38
rs754246193 Chr.7: 2432612-2432612 on Build GRCh38
rs900044566 Chr.7: 2432808-2432808 on Build GRCh38
rs756902847 Chr.7: 2432793-2432793 on Build GRCh38
rs749112375 Chr.7: 2432644-2432644 on Build GRCh38
rs766375870 Chr.7: 2432825-2432825 on Build GRCh38

View Full Product Details