Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93991403-93991975 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BET1
Entrez Gene ID
10282
Gene Name
Bet1 golgi vesicular membrane trafficking protein
Gene Aliases
HBET1
UniGene
Hs.489132
Gene Chromosome Location
Chr.7: 93962770-94004382 on Build GRCh38
Transcripts
NR_133908.1 NR_133909.1

SNP Detail

SNP ID Chromosome Location
rs145137028 Chr.7: 93991618-93991618 on Build GRCh38
rs1045886353 Chr.7: 93991740-93991740 on Build GRCh38
rs930521744 Chr.7: 93991782-93991782 on Build GRCh38
rs953374190 Chr.7: 93991640-93991640 on Build GRCh38
rs937470655 Chr.7: 93991763-93991763 on Build GRCh38
rs116063461 Chr.7: 93991677-93991677 on Build GRCh38
rs1030073881 Chr.7: 93991914-93991914 on Build GRCh38
rs961831084 Chr.7: 93991744-93991744 on Build GRCh38
rs1023782347 Chr.7: 93991683-93991683 on Build GRCh38
rs183849465 Chr.7: 93991833-93991833 on Build GRCh38
rs981851140 Chr.7: 93991713-93991713 on Build GRCh38
rs770158447 Chr.7: 93991790-93991790 on Build GRCh38
rs111400062 Chr.7: 93991556-93991556 on Build GRCh38
rs1035973101 Chr.7: 93991844-93991844 on Build GRCh38
rs920288538 Chr.7: 93991765-93991765 on Build GRCh38
rs758188721 Chr.7: 93991826-93991826 on Build GRCh38
rs557491766 Chr.7: 93991560-93991560 on Build GRCh38
rs555224602 Chr.7: 93991505-93991506 on Build GRCh38
rs1476875 Chr.7: 93991723-93991723 on Build GRCh38
rs971717086 Chr.7: 93991761-93991761 on Build GRCh38
rs544201377 Chr.7: 93991944-93991944 on Build GRCh38
rs144182020 Chr.7: 93991591-93991591 on Build GRCh38
rs73715580 Chr.7: 93991598-93991598 on Build GRCh38
rs745719807 Chr.7: 93991505-93991505 on Build GRCh38
rs969147421 Chr.7: 93991451-93991451 on Build GRCh38
rs189598731 Chr.7: 93991838-93991838 on Build GRCh38
rs944537741 Chr.7: 93991811-93991811 on Build GRCh38
rs1041531810 Chr.7: 93991611-93991611 on Build GRCh38
rs375261171 Chr.7: 93991854-93991854 on Build GRCh38
rs773617342 Chr.7: 93991808-93991808 on Build GRCh38
rs957986117 Chr.7: 93991678-93991678 on Build GRCh38
rs756045038 Chr.7: 93991595-93991595 on Build GRCh38
rs982288685 Chr.7: 93991458-93991458 on Build GRCh38
rs894047246 Chr.7: 93991639-93991639 on Build GRCh38
rs913283073 Chr.7: 93991500-93991500 on Build GRCh38
rs1001789569 Chr.7: 93991552-93991552 on Build GRCh38
rs999480565 Chr.7: 93991621-93991621 on Build GRCh38
rs180672382 Chr.7: 93991892-93991892 on Build GRCh38
rs971937949 Chr.7: 93991502-93991502 on Build GRCh38
rs558838307 Chr.7: 93991603-93991608 on Build GRCh38
rs986464033 Chr.7: 93991803-93991803 on Build GRCh38
rs930583675 Chr.7: 93991524-93991524 on Build GRCh38
rs866642449 Chr.7: 93991701-93991701 on Build GRCh38
rs1024250949 Chr.7: 93991436-93991436 on Build GRCh38
rs547433022 Chr.7: 93991668-93991669 on Build GRCh38
rs1030995643 Chr.7: 93991627-93991627 on Build GRCh38
rs538238292 Chr.7: 93991896-93991896 on Build GRCh38
rs901257816 Chr.7: 93991853-93991853 on Build GRCh38
rs907392379 Chr.7: 93991552-93991555 on Build GRCh38
rs771838901 Chr.7: 93991465-93991465 on Build GRCh38
rs928779066 Chr.7: 93991743-93991743 on Build GRCh38
rs181603970 Chr.7: 93991491-93991491 on Build GRCh38

View Full Product Details