Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41816925-41817349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP10
Entrez Gene ID
60681
Gene Name
FK506 binding protein 10
Gene Aliases
BRKS1, FKBP65, OI11, OI6, PPIASE, hFKBP65
UniGene
Hs.463035
Gene Chromosome Location
Chr.17: 41812262-41823217 on Build GRCh38
Transcripts
NM_021939.3 XM_011525099.2 XM_011525100.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6446181 c.351C>G p.L117L Chr.17: 41817163-41817163 on Build GRCh38
4878212 c.294C>A p.L98L Chr.17: 41817106-41817106 on Build GRCh38
1162247 c.281G>T p.G94V Chr.17: 41817093-41817093 on Build GRCh38
4854022 c.311G>A p.R104Q Chr.17: 41817123-41817123 on Build GRCh38
3819588 c.324C>T p.G108G Chr.17: 41817136-41817136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542560166 Chr.17: 41817307-41817307 on Build GRCh38
rs527980524 Chr.17: 41816983-41816983 on Build GRCh38
rs782054815 Chr.17: 41817051-41817051 on Build GRCh38
rs781920419 Chr.17: 41817047-41817048 on Build GRCh38
rs200421914 Chr.17: 41817066-41817066 on Build GRCh38
rs782790783 Chr.17: 41817046-41817046 on Build GRCh38
rs571063599 Chr.17: 41817158-41817158 on Build GRCh38
rs185145831 Chr.17: 41817245-41817245 on Build GRCh38
rs181832957 Chr.17: 41817006-41817006 on Build GRCh38
rs756749818 Chr.17: 41817317-41817317 on Build GRCh38
rs533997482 Chr.17: 41817128-41817128 on Build GRCh38
rs782758369 Chr.17: 41817186-41817186 on Build GRCh38
rs377522367 Chr.17: 41817134-41817134 on Build GRCh38
rs782507882 Chr.17: 41817231-41817231 on Build GRCh38
rs782403584 Chr.17: 41817159-41817159 on Build GRCh38
rs781863642 Chr.17: 41817137-41817137 on Build GRCh38
rs782567665 Chr.17: 41817152-41817152 on Build GRCh38
rs782422781 Chr.17: 41817087-41817087 on Build GRCh38
rs781898535 Chr.17: 41817196-41817196 on Build GRCh38
rs576914393 Chr.17: 41817169-41817169 on Build GRCh38
rs142507953 Chr.17: 41817123-41817123 on Build GRCh38
rs371079363 Chr.17: 41817102-41817102 on Build GRCh38
rs782229046 Chr.17: 41817153-41817153 on Build GRCh38
rs782687260 Chr.17: 41817232-41817232 on Build GRCh38
rs782784013 Chr.17: 41817112-41817112 on Build GRCh38
rs782085398 Chr.17: 41817103-41817103 on Build GRCh38
rs992705936 Chr.17: 41816963-41816963 on Build GRCh38
rs72835683 Chr.17: 41817122-41817122 on Build GRCh38
rs782046189 Chr.17: 41817167-41817167 on Build GRCh38
rs531212095 Chr.17: 41817319-41817319 on Build GRCh38
rs782208691 Chr.17: 41817253-41817253 on Build GRCh38
rs781936464 Chr.17: 41817160-41817160 on Build GRCh38
rs782415967 Chr.17: 41817227-41817227 on Build GRCh38
rs563996805 Chr.17: 41817310-41817310 on Build GRCh38
rs373064272 Chr.17: 41817127-41817127 on Build GRCh38
rs387906960 Chr.17: 41817156-41817156 on Build GRCh38
rs117954002 Chr.17: 41817287-41817287 on Build GRCh38
rs150313949 Chr.17: 41817065-41817065 on Build GRCh38
rs782277032 Chr.17: 41817116-41817116 on Build GRCh38
rs781869482 Chr.17: 41817228-41817228 on Build GRCh38
rs190994957 Chr.17: 41816967-41816967 on Build GRCh38
rs782567281 Chr.17: 41817055-41817055 on Build GRCh38
rs782676406 Chr.17: 41817148-41817148 on Build GRCh38
rs782316942 Chr.17: 41817097-41817097 on Build GRCh38
rs137853882 Chr.17: 41817133-41817165 on Build GRCh38
rs531095764 Chr.17: 41816966-41816966 on Build GRCh38
rs782356839 Chr.17: 41817175-41817175 on Build GRCh38
rs782500474 Chr.17: 41817139-41817139 on Build GRCh38
rs922801622 Chr.17: 41817242-41817242 on Build GRCh38
rs397514674 Chr.17: 41817149-41817149 on Build GRCh38
rs925937376 Chr.17: 41817197-41817197 on Build GRCh38
rs782339006 Chr.17: 41817326-41817326 on Build GRCh38
rs782199101 Chr.17: 41817094-41817094 on Build GRCh38
rs140027863 Chr.17: 41817053-41817053 on Build GRCh38
rs138188784 Chr.17: 41817008-41817009 on Build GRCh38
rs199576301 Chr.17: 41817121-41817121 on Build GRCh38
rs138713182 Chr.17: 41817184-41817184 on Build GRCh38
rs782788849 Chr.17: 41817250-41817251 on Build GRCh38
rs144787711 Chr.17: 41817176-41817176 on Build GRCh38
rs781885521 Chr.17: 41817054-41817054 on Build GRCh38
rs782486491 Chr.17: 41817220-41817220 on Build GRCh38
rs782608468 Chr.17: 41817249-41817249 on Build GRCh38
rs782156834 Chr.17: 41817200-41817200 on Build GRCh38
rs781939779 Chr.17: 41817048-41817048 on Build GRCh38
rs141847881 Chr.17: 41817204-41817204 on Build GRCh38
rs926082153 Chr.17: 41817005-41817005 on Build GRCh38
rs568065510 Chr.17: 41817024-41817024 on Build GRCh38
rs797044559 Chr.17: 41817165-41817165 on Build GRCh38
rs368939088 Chr.17: 41817038-41817038 on Build GRCh38
rs782167452 Chr.17: 41817035-41817035 on Build GRCh38
rs201433301 Chr.17: 41817012-41817012 on Build GRCh38
rs782308183 Chr.17: 41817246-41817246 on Build GRCh38
rs782388385 Chr.17: 41817016-41817016 on Build GRCh38
rs552704842 Chr.17: 41817285-41817285 on Build GRCh38
rs137992633 Chr.17: 41817091-41817091 on Build GRCh38
rs993652435 Chr.17: 41817101-41817101 on Build GRCh38
rs782165064 Chr.17: 41817048-41817048 on Build GRCh38
rs782227439 Chr.17: 41817064-41817064 on Build GRCh38
rs782236614 Chr.17: 41817086-41817086 on Build GRCh38
rs781795993 Chr.17: 41817218-41817218 on Build GRCh38
rs190930399 Chr.17: 41817325-41817325 on Build GRCh38
rs370972136 Chr.17: 41817155-41817155 on Build GRCh38

View Full Product Details