Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42907458-42907964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC
Entrez Gene ID
2538
Gene Name
glucose-6-phosphatase catalytic subunit
Gene Aliases
G6PC1, G6PT, G6Pase, GSD1, GSD1a
UniGene
Hs.212293
Gene Chromosome Location
Chr.17: 42900797-42914433 on Build GRCh38
Transcripts
NM_000151.3 NM_001270397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3819678 c.445C>T p.R149W Chr.17: 42907627-42907627 on Build GRCh38
281278 c.433A>G p.T145A Chr.17: 42907615-42907615 on Build GRCh38
6214857 c.432G>A p.P144P Chr.17: 42907614-42907614 on Build GRCh38
4543660 c.341G>A p.G114E Chr.17: 42907523-42907523 on Build GRCh38
32952 c.347C>T p.P116L Chr.17: 42907529-42907529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765306500 Chr.17: 42907550-42907550 on Build GRCh38
rs748912020 Chr.17: 42907515-42907515 on Build GRCh38
rs757401964 Chr.17: 42907866-42907866 on Build GRCh38
rs752726795 Chr.17: 42907556-42907556 on Build GRCh38
rs776998433 Chr.17: 42907665-42907665 on Build GRCh38
rs774885342 Chr.17: 42907511-42907511 on Build GRCh38
rs161626 Chr.17: 42907778-42907778 on Build GRCh38
rs562533427 Chr.17: 42907539-42907539 on Build GRCh38
rs161625 Chr.17: 42907888-42907888 on Build GRCh38
rs757173918 Chr.17: 42907481-42907481 on Build GRCh38
rs924385591 Chr.17: 42907725-42907725 on Build GRCh38
rs968407502 Chr.17: 42907696-42907696 on Build GRCh38
rs752496230 Chr.17: 42907715-42907715 on Build GRCh38
rs773963651 Chr.17: 42907641-42907641 on Build GRCh38
rs369181197 Chr.17: 42907699-42907699 on Build GRCh38
rs762677766 Chr.17: 42907745-42907745 on Build GRCh38
rs539615669 Chr.17: 42907677-42907677 on Build GRCh38
rs527336983 Chr.17: 42907683-42907683 on Build GRCh38
rs769303030 Chr.17: 42907505-42907505 on Build GRCh38
rs777888691 Chr.17: 42907620-42907621 on Build GRCh38
rs532763969 Chr.17: 42907561-42907561 on Build GRCh38
rs181624619 Chr.17: 42907628-42907628 on Build GRCh38
rs161628 Chr.17: 42907614-42907614 on Build GRCh38
rs774055151 Chr.17: 42907527-42907527 on Build GRCh38
rs78890897 Chr.17: 42907898-42907898 on Build GRCh38
rs745770086 Chr.17: 42907617-42907617 on Build GRCh38
rs1049015719 Chr.17: 42907845-42907845 on Build GRCh38
rs956899100 Chr.17: 42907799-42907799 on Build GRCh38
rs1003569893 Chr.17: 42907635-42907635 on Build GRCh38
rs527971556 Chr.17: 42907691-42907691 on Build GRCh38
rs776533959 Chr.17: 42907485-42907485 on Build GRCh38
rs766771010 Chr.17: 42907536-42907536 on Build GRCh38
rs760024886 Chr.17: 42907666-42907666 on Build GRCh38
rs765661050 Chr.17: 42907668-42907668 on Build GRCh38
rs747830634 Chr.17: 42907648-42907648 on Build GRCh38
rs36031531 Chr.17: 42907719-42907719 on Build GRCh38
rs772500296 Chr.17: 42907726-42907726 on Build GRCh38
rs764401169 Chr.17: 42907566-42907566 on Build GRCh38
rs756018092 Chr.17: 42907621-42907621 on Build GRCh38
rs921480433 Chr.17: 42907788-42907788 on Build GRCh38
rs796136235 Chr.17: 42907605-42907605 on Build GRCh38
rs606231368 Chr.17: 42907561-42907562 on Build GRCh38
rs771236645 Chr.17: 42907660-42907660 on Build GRCh38
rs968151615 Chr.17: 42907759-42907759 on Build GRCh38
rs888630335 Chr.17: 42907865-42907865 on Build GRCh38
rs139724906 Chr.17: 42907940-42907941 on Build GRCh38
rs779548820 Chr.17: 42907627-42907627 on Build GRCh38
rs371429873 Chr.17: 42907832-42907832 on Build GRCh38
rs753163238 Chr.17: 42907492-42907493 on Build GRCh38
rs751875988 Chr.17: 42907567-42907567 on Build GRCh38
rs80356488 Chr.17: 42907562-42907563 on Build GRCh38
rs161627 Chr.17: 42907701-42907701 on Build GRCh38
rs371152892 Chr.17: 42907713-42907714 on Build GRCh38
rs906657263 Chr.17: 42907924-42907924 on Build GRCh38
rs768206272 Chr.17: 42907634-42907634 on Build GRCh38
rs104894568 Chr.17: 42907552-42907552 on Build GRCh38
rs143928802 Chr.17: 42907690-42907690 on Build GRCh38
rs1036015150 Chr.17: 42907642-42907642 on Build GRCh38
rs757083891 Chr.17: 42907571-42907571 on Build GRCh38
rs576442264 Chr.17: 42907733-42907733 on Build GRCh38
rs544563908 Chr.17: 42907523-42907523 on Build GRCh38
rs888493610 Chr.17: 42907577-42907577 on Build GRCh38
rs761007323 Chr.17: 42907534-42907534 on Build GRCh38
rs759982943 Chr.17: 42907547-42907547 on Build GRCh38
rs935614385 Chr.17: 42907483-42907483 on Build GRCh38

View Full Product Details