Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42910830-42911337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC
Entrez Gene ID
2538
Gene Name
glucose-6-phosphatase catalytic subunit
Gene Aliases
G6PC1, G6PT, G6Pase, GSD1, GSD1a
UniGene
Hs.212293
Gene Chromosome Location
Chr.17: 42900797-42914433 on Build GRCh38
Transcripts
NM_000151.3 NM_001270397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5779613 c.884G>A p.R295H Chr.17: 42911236-42911236 on Build GRCh38
4066708 c.972C>A p.V324V Chr.17: 42911324-42911324 on Build GRCh38
1710329 c.857G>A p.G286E Chr.17: 42911209-42911209 on Build GRCh38
5599583 c.932C>T p.S311F Chr.17: 42911284-42911284 on Build GRCh38
107465 c.750G>A p.W250* Chr.17: 42911102-42911102 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143321486 Chr.17: 42911295-42911295 on Build GRCh38
rs374522795 Chr.17: 42910967-42910967 on Build GRCh38
rs761361643 Chr.17: 42911174-42911174 on Build GRCh38
rs756784163 Chr.17: 42911080-42911080 on Build GRCh38
rs104894563 Chr.17: 42911235-42911235 on Build GRCh38
rs767184894 Chr.17: 42911086-42911086 on Build GRCh38
rs780317668 Chr.17: 42911248-42911248 on Build GRCh38
rs992366326 Chr.17: 42910935-42910935 on Build GRCh38
rs774448424 Chr.17: 42911274-42911274 on Build GRCh38
rs751120985 Chr.17: 42910976-42910976 on Build GRCh38
rs765059867 Chr.17: 42911037-42911037 on Build GRCh38
rs753486821 Chr.17: 42911296-42911296 on Build GRCh38
rs975103942 Chr.17: 42911201-42911201 on Build GRCh38
rs374980545 Chr.17: 42911198-42911198 on Build GRCh38
rs80356485 Chr.17: 42911076-42911076 on Build GRCh38
rs138509513 Chr.17: 42911138-42911138 on Build GRCh38
rs377622036 Chr.17: 42910878-42910878 on Build GRCh38
rs764084156 Chr.17: 42911172-42911172 on Build GRCh38
rs760981149 Chr.17: 42910915-42910915 on Build GRCh38
rs767174294 Chr.17: 42911177-42911177 on Build GRCh38
rs749665480 Chr.17: 42910998-42910998 on Build GRCh38
rs769527058 Chr.17: 42911158-42911158 on Build GRCh38
rs773356062 Chr.17: 42911011-42911011 on Build GRCh38
rs749323139 Chr.17: 42911150-42911150 on Build GRCh38
rs515726229 Chr.17: 42911017-42911017 on Build GRCh38
rs765150017 Chr.17: 42910937-42910937 on Build GRCh38
rs9905900 Chr.17: 42911078-42911078 on Build GRCh38
rs942377882 Chr.17: 42911175-42911175 on Build GRCh38
rs145172999 Chr.17: 42911046-42911046 on Build GRCh38
rs770003650 Chr.17: 42911244-42911244 on Build GRCh38
rs141352548 Chr.17: 42911294-42911294 on Build GRCh38
rs370182849 Chr.17: 42911165-42911165 on Build GRCh38
rs756944737 Chr.17: 42910986-42910986 on Build GRCh38
rs780212338 Chr.17: 42910995-42910995 on Build GRCh38
rs77814737 Chr.17: 42910936-42910936 on Build GRCh38
rs765763631 Chr.17: 42911028-42911028 on Build GRCh38
rs370472759 Chr.17: 42910874-42910874 on Build GRCh38
rs749749697 Chr.17: 42910870-42910870 on Build GRCh38
rs201961848 Chr.17: 42910962-42910962 on Build GRCh38
rs368282218 Chr.17: 42911261-42911261 on Build GRCh38
rs766030987 Chr.17: 42910916-42910916 on Build GRCh38
rs140224951 Chr.17: 42911145-42911145 on Build GRCh38
rs375212414 Chr.17: 42911236-42911236 on Build GRCh38
rs758775116 Chr.17: 42911120-42911120 on Build GRCh38
rs773267957 Chr.17: 42910895-42910895 on Build GRCh38
rs886052956 Chr.17: 42910929-42910929 on Build GRCh38
rs754345839 Chr.17: 42910934-42910934 on Build GRCh38
rs969519705 Chr.17: 42910963-42910963 on Build GRCh38
rs559563847 Chr.17: 42911109-42911109 on Build GRCh38
rs201520075 Chr.17: 42911009-42911009 on Build GRCh38
rs80356483 Chr.17: 42911161-42911161 on Build GRCh38
rs753009862 Chr.17: 42911092-42911092 on Build GRCh38
rs377077333 Chr.17: 42911156-42911156 on Build GRCh38
rs558996257 Chr.17: 42910873-42910873 on Build GRCh38
rs759306891 Chr.17: 42911029-42911029 on Build GRCh38
rs1057516858 Chr.17: 42911210-42911210 on Build GRCh38
rs993458406 Chr.17: 42911229-42911229 on Build GRCh38
rs368041878 Chr.17: 42911222-42911222 on Build GRCh38
rs887055303 Chr.17: 42910875-42910875 on Build GRCh38
rs889272586 Chr.17: 42911088-42911088 on Build GRCh38
rs762728678 Chr.17: 42911168-42911168 on Build GRCh38
rs933602270 Chr.17: 42911044-42911044 on Build GRCh38
rs863224021 Chr.17: 42911234-42911234 on Build GRCh38
rs371574035 Chr.17: 42910892-42910892 on Build GRCh38
rs772241157 Chr.17: 42910886-42910886 on Build GRCh38
rs755630386 Chr.17: 42911153-42911153 on Build GRCh38
rs531727073 Chr.17: 42911312-42911312 on Build GRCh38
rs774311350 Chr.17: 42911265-42911265 on Build GRCh38
rs529957748 Chr.17: 42911231-42911231 on Build GRCh38
rs137978566 Chr.17: 42911300-42911300 on Build GRCh38
rs771651069 Chr.17: 42911293-42911293 on Build GRCh38
rs907204295 Chr.17: 42910907-42910907 on Build GRCh38
rs781180357 Chr.17: 42911307-42911307 on Build GRCh38
rs752049826 Chr.17: 42911230-42911230 on Build GRCh38
rs756241523 Chr.17: 42911313-42911313 on Build GRCh38
rs747134498 Chr.17: 42911015-42911015 on Build GRCh38
rs768385469 Chr.17: 42911262-42911262 on Build GRCh38
rs145296477 Chr.17: 42911152-42911152 on Build GRCh38
rs80356484 Chr.17: 42911000-42911000 on Build GRCh38
rs774212157 Chr.17: 42911173-42911173 on Build GRCh38
rs140869027 Chr.17: 42911102-42911102 on Build GRCh38
rs748841842 Chr.17: 42911217-42911217 on Build GRCh38
rs779538234 Chr.17: 42911003-42911003 on Build GRCh38
rs554145213 Chr.17: 42911268-42911268 on Build GRCh38
rs534670926 Chr.17: 42911208-42911208 on Build GRCh38
rs899288649 Chr.17: 42911162-42911162 on Build GRCh38
rs750147891 Chr.17: 42911087-42911087 on Build GRCh38
rs765631320 Chr.17: 42911188-42911188 on Build GRCh38
rs111259657 Chr.17: 42911309-42911309 on Build GRCh38
rs201649392 Chr.17: 42910990-42910990 on Build GRCh38
rs372612424 Chr.17: 42911306-42911306 on Build GRCh38
rs771997505 Chr.17: 42911010-42911010 on Build GRCh38
rs753671536 Chr.17: 42910922-42910922 on Build GRCh38
rs750048146 Chr.17: 42911182-42911182 on Build GRCh38
rs748610705 Chr.17: 42911132-42911132 on Build GRCh38
rs776048625 Chr.17: 42911214-42911214 on Build GRCh38
rs779444123 Chr.17: 42911124-42911124 on Build GRCh38
rs762967975 Chr.17: 42911075-42911075 on Build GRCh38

View Full Product Details