Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42912589-42913132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PC
Entrez Gene ID
2538
Gene Name
glucose-6-phosphatase catalytic subunit
Gene Aliases
G6PC1, G6PT, G6Pase, GSD1, GSD1a
UniGene
Hs.212293
Gene Chromosome Location
Chr.17: 42900797-42914433 on Build GRCh38
Transcripts
NM_000151.3 NM_001270397.1

SNP Detail

SNP ID Chromosome Location
rs1033442792 Chr.17: 42912646-42912647 on Build GRCh38
rs559807250 Chr.17: 42912723-42912723 on Build GRCh38
rs944532944 Chr.17: 42912897-42912897 on Build GRCh38
rs552879065 Chr.17: 42912915-42912915 on Build GRCh38
rs144568330 Chr.17: 42912636-42912636 on Build GRCh38
rs1034358441 Chr.17: 42912779-42912779 on Build GRCh38
rs886663344 Chr.17: 42912739-42912739 on Build GRCh38
rs397735497 Chr.17: 42912653-42912654 on Build GRCh38
rs944611985 Chr.17: 42913100-42913100 on Build GRCh38
rs1001624548 Chr.17: 42913034-42913034 on Build GRCh38
rs558231884 Chr.17: 42912937-42912937 on Build GRCh38
rs907455211 Chr.17: 42913023-42913023 on Build GRCh38
rs201722515 Chr.17: 42912980-42912987 on Build GRCh38
rs932822722 Chr.17: 42912780-42912780 on Build GRCh38
rs966575521 Chr.17: 42912953-42912953 on Build GRCh38
rs934332561 Chr.17: 42912756-42912756 on Build GRCh38
rs959212634 Chr.17: 42912658-42912658 on Build GRCh38
rs980278332 Chr.17: 42912764-42912764 on Build GRCh38
rs148453821 Chr.17: 42912880-42912880 on Build GRCh38
rs377751565 Chr.17: 42913101-42913101 on Build GRCh38
rs534589177 Chr.17: 42912967-42912967 on Build GRCh38
rs7210335 Chr.17: 42913090-42913090 on Build GRCh38
rs994462869 Chr.17: 42912734-42912734 on Build GRCh38
rs921369844 Chr.17: 42912767-42912767 on Build GRCh38
rs1024977369 Chr.17: 42912999-42912999 on Build GRCh38
rs1005529625 Chr.17: 42912745-42912745 on Build GRCh38
rs957460607 Chr.17: 42912791-42912791 on Build GRCh38
rs946091500 Chr.17: 42912830-42912830 on Build GRCh38
rs1008760378 Chr.17: 42912716-42912716 on Build GRCh38
rs1027330315 Chr.17: 42912735-42912735 on Build GRCh38
rs200704026 Chr.17: 42912640-42912640 on Build GRCh38
rs976779865 Chr.17: 42912763-42912763 on Build GRCh38
rs976750374 Chr.17: 42913046-42913046 on Build GRCh38
rs572871169 Chr.17: 42912898-42912898 on Build GRCh38
rs911757369 Chr.17: 42913098-42913098 on Build GRCh38
rs921779184 Chr.17: 42913103-42913103 on Build GRCh38
rs957429317 Chr.17: 42913054-42913054 on Build GRCh38
rs765883953 Chr.17: 42913078-42913078 on Build GRCh38
rs556943624 Chr.17: 42913066-42913066 on Build GRCh38
rs545658086 Chr.17: 42912631-42912631 on Build GRCh38
rs111309126 Chr.17: 42912940-42912940 on Build GRCh38
rs113536060 Chr.17: 42912826-42912826 on Build GRCh38
rs886093149 Chr.17: 42912960-42912960 on Build GRCh38
rs397717945 Chr.17: 42912979-42912986 on Build GRCh38
rs374021788 Chr.17: 42912939-42912939 on Build GRCh38
rs138968865 Chr.17: 42912978-42912978 on Build GRCh38
rs537388227 Chr.17: 42913031-42913031 on Build GRCh38
rs990081454 Chr.17: 42912822-42912822 on Build GRCh38
rs763493186 Chr.17: 42912859-42912859 on Build GRCh38
rs1020395572 Chr.17: 42913068-42913068 on Build GRCh38
rs546236128 Chr.17: 42913083-42913083 on Build GRCh38
rs989746795 Chr.17: 42912699-42912699 on Build GRCh38
rs11436510 Chr.17: 42912639-42912640 on Build GRCh38
rs1054862308 Chr.17: 42912936-42912936 on Build GRCh38
rs549833149 Chr.17: 42912730-42912730 on Build GRCh38
rs161620 Chr.17: 42912720-42912720 on Build GRCh38
rs568149292 Chr.17: 42913097-42913097 on Build GRCh38
rs890265916 Chr.17: 42912701-42912701 on Build GRCh38
rs888701395 Chr.17: 42912805-42912805 on Build GRCh38
rs548787621 Chr.17: 42912750-42912750 on Build GRCh38
rs1034494396 Chr.17: 42913042-42913042 on Build GRCh38
rs201313100 Chr.17: 42912632-42912633 on Build GRCh38
rs886052962 Chr.17: 42912739-42912739 on Build GRCh38
rs971966934 Chr.17: 42912752-42912752 on Build GRCh38
rs940217664 Chr.17: 42913092-42913092 on Build GRCh38

View Full Product Details