Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33575206-33575697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAK1
Entrez Gene ID
578
Gene Name
BCL2 antagonist/killer 1
Gene Aliases
BAK, BAK-LIKE, BCL2L7, CDN1
UniGene
Hs.485139
Gene Chromosome Location
Chr.6: 33572546-33580293 on Build GRCh38
Transcripts
NM_001188.3 XM_011514779.2 XM_011514780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5405045 c.284C>T p.T95I Chr.6: 33575364-33575364 on Build GRCh38
5031469 c.207-10G>A p.? Chr.6: 33575451-33575451 on Build GRCh38
4976940 c.234C>T p.L78L Chr.6: 33575414-33575414 on Build GRCh38
6453638 c.250G>A p.D84N Chr.6: 33575398-33575398 on Build GRCh38
4968541 c.235G>A p.A79T Chr.6: 33575413-33575413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779411450 Chr.6: 33575234-33575234 on Build GRCh38
rs766540475 Chr.6: 33575347-33575347 on Build GRCh38
rs1050604168 Chr.6: 33575507-33575507 on Build GRCh38
rs1051913 Chr.6: 33575405-33575405 on Build GRCh38
rs766561404 Chr.6: 33575422-33575422 on Build GRCh38
rs770571379 Chr.6: 33575389-33575389 on Build GRCh38
rs769426436 Chr.6: 33575401-33575401 on Build GRCh38
rs755256768 Chr.6: 33575427-33575427 on Build GRCh38
rs373619041 Chr.6: 33575335-33575335 on Build GRCh38
rs1027257190 Chr.6: 33575499-33575499 on Build GRCh38
rs210139 Chr.6: 33575632-33575632 on Build GRCh38
rs775106891 Chr.6: 33575460-33575460 on Build GRCh38
rs763976645 Chr.6: 33575417-33575417 on Build GRCh38
rs765284813 Chr.6: 33575428-33575428 on Build GRCh38
rs915665710 Chr.6: 33575268-33575268 on Build GRCh38
rs199754748 Chr.6: 33575448-33575448 on Build GRCh38
rs890590689 Chr.6: 33575523-33575523 on Build GRCh38
rs778171445 Chr.6: 33575385-33575385 on Build GRCh38
rs371320117 Chr.6: 33575356-33575356 on Build GRCh38
rs961775059 Chr.6: 33575609-33575609 on Build GRCh38
rs756326673 Chr.6: 33575343-33575343 on Build GRCh38
rs961670280 Chr.6: 33575341-33575341 on Build GRCh38
rs934355567 Chr.6: 33575644-33575644 on Build GRCh38
rs563033017 Chr.6: 33575269-33575269 on Build GRCh38
rs34017174 Chr.6: 33575431-33575432 on Build GRCh38
rs748646933 Chr.6: 33575378-33575378 on Build GRCh38
rs568665592 Chr.6: 33575458-33575458 on Build GRCh38
rs761535491 Chr.6: 33575421-33575421 on Build GRCh38
rs981235602 Chr.6: 33575626-33575626 on Build GRCh38
rs764414042 Chr.6: 33575486-33575486 on Build GRCh38
rs748450309 Chr.6: 33575235-33575235 on Build GRCh38
rs752817502 Chr.6: 33575429-33575429 on Build GRCh38
rs563897863 Chr.6: 33575355-33575355 on Build GRCh38
rs551156164 Chr.6: 33575593-33575593 on Build GRCh38
rs541570437 Chr.6: 33575264-33575264 on Build GRCh38
rs754815562 Chr.6: 33575473-33575473 on Build GRCh38
rs892929984 Chr.6: 33575398-33575398 on Build GRCh38
rs759142830 Chr.6: 33575322-33575322 on Build GRCh38
rs1049800577 Chr.6: 33575407-33575407 on Build GRCh38
rs781732557 Chr.6: 33575446-33575446 on Build GRCh38
rs918121246 Chr.6: 33575542-33575542 on Build GRCh38
rs535663497 Chr.6: 33575496-33575496 on Build GRCh38
rs5745590 Chr.6: 33575611-33575627 on Build GRCh38
rs747344615 Chr.6: 33575439-33575439 on Build GRCh38
rs776171225 Chr.6: 33575309-33575309 on Build GRCh38
rs528797429 Chr.6: 33575437-33575437 on Build GRCh38
rs917081722 Chr.6: 33575668-33575668 on Build GRCh38
rs746711568 Chr.6: 33575386-33575386 on Build GRCh38
rs5745591 Chr.6: 33575598-33575600 on Build GRCh38
rs375888869 Chr.6: 33575430-33575430 on Build GRCh38
rs745314875 Chr.6: 33575399-33575399 on Build GRCh38
rs775198844 Chr.6: 33575404-33575404 on Build GRCh38
rs5745592 Chr.6: 33575441-33575441 on Build GRCh38
rs561276 Chr.6: 33575339-33575339 on Build GRCh38
rs368302168 Chr.6: 33575444-33575444 on Build GRCh38
rs200462463 Chr.6: 33575303-33575303 on Build GRCh38
rs758884894 Chr.6: 33575382-33575382 on Build GRCh38
rs762960717 Chr.6: 33575413-33575413 on Build GRCh38
rs770531908 Chr.6: 33575292-33575292 on Build GRCh38
rs758307806 Chr.6: 33575401-33575401 on Build GRCh38
rs776437416 Chr.6: 33575395-33575395 on Build GRCh38
rs748778111 Chr.6: 33575463-33575463 on Build GRCh38
rs1015992071 Chr.6: 33575461-33575461 on Build GRCh38
rs201243778 Chr.6: 33575595-33575597 on Build GRCh38
rs991398067 Chr.6: 33575659-33575659 on Build GRCh38
rs372657768 Chr.6: 33575492-33575492 on Build GRCh38
rs990957567 Chr.6: 33575527-33575527 on Build GRCh38
rs377341171 Chr.6: 33575353-33575353 on Build GRCh38
rs754059376 Chr.6: 33575426-33575426 on Build GRCh38
rs112843934 Chr.6: 33575524-33575524 on Build GRCh38
rs781638668 Chr.6: 33575269-33575270 on Build GRCh38
rs958886445 Chr.6: 33575646-33575646 on Build GRCh38
rs958587510 Chr.6: 33575368-33575368 on Build GRCh38
rs991176922 Chr.6: 33575226-33575226 on Build GRCh38
rs373778311 Chr.6: 33575340-33575340 on Build GRCh38
rs768589296 Chr.6: 33575479-33575479 on Build GRCh38
rs763052569 Chr.6: 33575337-33575337 on Build GRCh38
rs772427718 Chr.6: 33575246-33575246 on Build GRCh38
rs769110361 Chr.6: 33575231-33575231 on Build GRCh38
rs971120269 Chr.6: 33575583-33575583 on Build GRCh38
rs869244137 Chr.6: 33575600-33575602 on Build GRCh38

View Full Product Details